Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 23641023-23641519 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCTN5
Entrez Gene ID
84516
Gene Name
dynactin subunit 5
Gene Aliases
-
UniGene
Hs.435941
Gene Chromosome Location
Chr.16: 23641366-23676207 on Build GRCh38
Transcripts
NM_001199011.1 NM_001199743.1 NM_032486.3 NR_037573.1 NM_024675.3 XM_011545946.2 XM_011545947.2 XM_011545948.2 XM_017023671.1 XM_017023672.1 XM_017023673.1

Gene Symbol
PALB2
Entrez Gene ID
79728
Gene Name
partner and localizer of BRCA2
Gene Aliases
FANCN, PNCA3
UniGene
Hs.444664
Gene Chromosome Location
Chr.16: 23603162-23641357 on Build GRCh38
Transcripts
NM_001199011.1 NM_001199743.1 NM_032486.3 NR_037573.1 NM_024675.3 XM_011545946.2 XM_011545947.2 XM_011545948.2 XM_017023671.1 XM_017023672.1 XM_017023673.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6144082 c.5A>G p.D2G Chr.16: 23641153-23641153 on Build GRCh38
6928689 c.32G>T p.C11F Chr.16: 23641126-23641126 on Build GRCh38
6428813 c.47A>G p.K16R Chr.16: 23641111-23641111 on Build GRCh38
21729 c.25C>T p.L9F Chr.16: 23641133-23641133 on Build GRCh38
6960271 c.34G>T p.E12* Chr.16: 23641124-23641124 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375809781 Chr.16: 23641428-23641428 on Build GRCh38
rs138200248 Chr.16: 23641315-23641315 on Build GRCh38
rs766487850 Chr.16: 23641134-23641134 on Build GRCh38
rs1017830508 Chr.16: 23641219-23641219 on Build GRCh38
rs876659643 Chr.16: 23641129-23641129 on Build GRCh38
rs559280197 Chr.16: 23641294-23641294 on Build GRCh38
rs772854530 Chr.16: 23641331-23641331 on Build GRCh38
rs897865845 Chr.16: 23641174-23641174 on Build GRCh38
rs763284273 Chr.16: 23641496-23641496 on Build GRCh38
rs730881884 Chr.16: 23641115-23641115 on Build GRCh38
rs759520257 Chr.16: 23641094-23641094 on Build GRCh38
rs150390726 Chr.16: 23641135-23641135 on Build GRCh38
rs876658438 Chr.16: 23641143-23641143 on Build GRCh38
rs515726092 Chr.16: 23641132-23641132 on Build GRCh38
rs750522322 Chr.16: 23641063-23641063 on Build GRCh38
rs942422168 Chr.16: 23641307-23641307 on Build GRCh38
rs748971865 Chr.16: 23641175-23641175 on Build GRCh38
rs771913164 Chr.16: 23641329-23641329 on Build GRCh38
rs1046992072 Chr.16: 23641369-23641369 on Build GRCh38
rs905325642 Chr.16: 23641386-23641386 on Build GRCh38
rs745444171 Chr.16: 23641119-23641121 on Build GRCh38
rs890546426 Chr.16: 23641475-23641475 on Build GRCh38
rs587781662 Chr.16: 23641132-23641132 on Build GRCh38
rs886051832 Chr.16: 23641306-23641306 on Build GRCh38
rs876659744 Chr.16: 23641123-23641123 on Build GRCh38
rs1017776612 Chr.16: 23641490-23641490 on Build GRCh38
rs876659843 Chr.16: 23641106-23641106 on Build GRCh38
rs1057517585 Chr.16: 23641155-23641155 on Build GRCh38
rs773924164 Chr.16: 23641181-23641181 on Build GRCh38
rs769435363 Chr.16: 23641082-23641082 on Build GRCh38
rs758500749 Chr.16: 23641128-23641128 on Build GRCh38
rs587776405 Chr.16: 23641110-23641110 on Build GRCh38
rs515726119 Chr.16: 23641083-23641083 on Build GRCh38
rs377085677 Chr.16: 23641145-23641145 on Build GRCh38
rs915451951 Chr.16: 23641268-23641268 on Build GRCh38
rs200523571 Chr.16: 23641188-23641188 on Build GRCh38
rs539371156 Chr.16: 23641285-23641285 on Build GRCh38
rs1007749423 Chr.16: 23641218-23641218 on Build GRCh38
rs376865021 Chr.16: 23641206-23641206 on Build GRCh38
rs888298194 Chr.16: 23641220-23641220 on Build GRCh38
rs988399385 Chr.16: 23641266-23641266 on Build GRCh38
rs876660586 Chr.16: 23641136-23641136 on Build GRCh38
rs941950824 Chr.16: 23641420-23641420 on Build GRCh38
rs768839390 Chr.16: 23641071-23641071 on Build GRCh38
rs1032418992 Chr.16: 23641280-23641280 on Build GRCh38
rs888323089 Chr.16: 23641370-23641370 on Build GRCh38
rs766570991 Chr.16: 23641497-23641497 on Build GRCh38
rs587781902 Chr.16: 23641159-23641159 on Build GRCh38
rs947155109 Chr.16: 23641138-23641138 on Build GRCh38
rs515726056 Chr.16: 23641182-23641182 on Build GRCh38
rs770451617 Chr.16: 23641089-23641089 on Build GRCh38
rs1038099508 Chr.16: 23641167-23641167 on Build GRCh38
rs765520187 Chr.16: 23641124-23641124 on Build GRCh38
rs760609798 Chr.16: 23641122-23641122 on Build GRCh38
rs786202325 Chr.16: 23641149-23641149 on Build GRCh38
rs756519825 Chr.16: 23641156-23641156 on Build GRCh38
rs779448230 Chr.16: 23641087-23641088 on Build GRCh38
rs748854938 Chr.16: 23641107-23641107 on Build GRCh38
rs770278498 Chr.16: 23641178-23641178 on Build GRCh38
rs556490497 Chr.16: 23641202-23641202 on Build GRCh38
rs879254144 Chr.16: 23641157-23641157 on Build GRCh38
rs587782462 Chr.16: 23641140-23641140 on Build GRCh38
rs1040725416 Chr.16: 23641175-23641176 on Build GRCh38
rs573641202 Chr.16: 23641458-23641458 on Build GRCh38
rs1009961310 Chr.16: 23641412-23641412 on Build GRCh38
rs748522775 Chr.16: 23641081-23641081 on Build GRCh38
rs587782483 Chr.16: 23641148-23641148 on Build GRCh38
rs968965905 Chr.16: 23641254-23641254 on Build GRCh38
rs956978455 Chr.16: 23641266-23641266 on Build GRCh38
rs878855123 Chr.16: 23641151-23641151 on Build GRCh38
rs956720208 Chr.16: 23641393-23641393 on Build GRCh38
rs956420088 Chr.16: 23641298-23641298 on Build GRCh38
rs1007697120 Chr.16: 23641481-23641481 on Build GRCh38
rs926493297 Chr.16: 23641408-23641408 on Build GRCh38
rs963816451 Chr.16: 23641474-23641474 on Build GRCh38
rs1044869769 Chr.16: 23641385-23641385 on Build GRCh38
rs868244456 Chr.16: 23641376-23641376 on Build GRCh38
rs765927659 Chr.16: 23641465-23641465 on Build GRCh38
rs1035014687 Chr.16: 23641392-23641392 on Build GRCh38
rs911005977 Chr.16: 23641287-23641287 on Build GRCh38
rs567706422 Chr.16: 23641146-23641146 on Build GRCh38
rs786202029 Chr.16: 23641118-23641118 on Build GRCh38
rs980582134 Chr.16: 23641396-23641396 on Build GRCh38
rs8053188 Chr.16: 23641204-23641204 on Build GRCh38
rs552824227 Chr.16: 23641448-23641448 on Build GRCh38
rs45619737 Chr.16: 23641147-23641147 on Build GRCh38
rs1052258495 Chr.16: 23641463-23641463 on Build GRCh38
rs876658288 Chr.16: 23641120-23641120 on Build GRCh38
rs929661968 Chr.16: 23641495-23641495 on Build GRCh38
rs1047365776 Chr.16: 23641213-23641213 on Build GRCh38
rs749298208 Chr.16: 23641131-23641131 on Build GRCh38
rs968246597 Chr.16: 23641282-23641282 on Build GRCh38
rs1023723550 Chr.16: 23641242-23641242 on Build GRCh38
rs747570989 Chr.16: 23641070-23641070 on Build GRCh38
rs758587524 Chr.16: 23641067-23641067 on Build GRCh38
rs515726118 Chr.16: 23641109-23641109 on Build GRCh38
rs876660538 Chr.16: 23641119-23641119 on Build GRCh38
rs1005775272 Chr.16: 23641226-23641226 on Build GRCh38
rs515726054 Chr.16: 23641351-23641351 on Build GRCh38
rs730881875 Chr.16: 23641162-23641162 on Build GRCh38
rs373698818 Chr.16: 23641302-23641302 on Build GRCh38
rs377315640 Chr.16: 23641164-23641164 on Build GRCh38
rs963683062 Chr.16: 23641068-23641068 on Build GRCh38
rs180177140 Chr.16: 23641265-23641265 on Build GRCh38
rs773976779 Chr.16: 23641187-23641187 on Build GRCh38
rs373287455 Chr.16: 23641121-23641121 on Build GRCh38
rs781632286 Chr.16: 23641075-23641075 on Build GRCh38
rs876659063 Chr.16: 23641123-23641123 on Build GRCh38
rs515726055 Chr.16: 23641384-23641384 on Build GRCh38
rs180177144 Chr.16: 23641333-23641333 on Build GRCh38
rs979068942 Chr.16: 23641271-23641271 on Build GRCh38
rs180177141 Chr.16: 23641203-23641203 on Build GRCh38
rs879254062 Chr.16: 23641145-23641146 on Build GRCh38
rs1005393230 Chr.16: 23641380-23641380 on Build GRCh38
rs750720488 Chr.16: 23641126-23641126 on Build GRCh38
rs876659679 Chr.16: 23641108-23641108 on Build GRCh38
rs1003567466 Chr.16: 23641388-23641388 on Build GRCh38
rs748775669 Chr.16: 23641084-23641084 on Build GRCh38
rs201765609 Chr.16: 23641090-23641090 on Build GRCh38
rs775325716 Chr.16: 23641193-23641193 on Build GRCh38
rs515726058 Chr.16: 23641255-23641255 on Build GRCh38
rs190626072 Chr.16: 23641103-23641103 on Build GRCh38
rs929336251 Chr.16: 23641461-23641461 on Build GRCh38
rs1003534559 Chr.16: 23641259-23641259 on Build GRCh38

View Full Product Details