Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 23642972-23643514 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCTN5
Entrez Gene ID
84516
Gene Name
dynactin subunit 5
Gene Aliases
-
UniGene
Hs.435941
Gene Chromosome Location
Chr.16: 23641366-23676207 on Build GRCh38
Transcripts
NM_001199011.1 NM_001199743.1 NM_032486.3 NR_037573.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5631146 c.86G>C p.C29S Chr.16: 23642992-23642992 on Build GRCh38
4940766 c.69T>C p.S23S Chr.16: 23642975-23642975 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs546015081 Chr.16: 23643390-23643390 on Build GRCh38
rs199556350 Chr.16: 23643186-23643187 on Build GRCh38
rs896279007 Chr.16: 23643255-23643255 on Build GRCh38
rs762280184 Chr.16: 23643265-23643265 on Build GRCh38
rs946177829 Chr.16: 23643256-23643256 on Build GRCh38
rs141294424 Chr.16: 23642996-23642996 on Build GRCh38
rs770525009 Chr.16: 23643144-23643144 on Build GRCh38
rs386789819 Chr.16: 23643195-23643197 on Build GRCh38
rs944367638 Chr.16: 23643168-23643168 on Build GRCh38
rs56776509 Chr.16: 23643289-23643289 on Build GRCh38
rs774818550 Chr.16: 23643245-23643245 on Build GRCh38
rs776593520 Chr.16: 23643453-23643453 on Build GRCh38
rs755872761 Chr.16: 23643053-23643053 on Build GRCh38
rs1053442294 Chr.16: 23643336-23643336 on Build GRCh38
rs369807 Chr.16: 23643203-23643203 on Build GRCh38
rs56737329 Chr.16: 23643049-23643049 on Build GRCh38
rs951716780 Chr.16: 23643092-23643092 on Build GRCh38
rs577089793 Chr.16: 23643353-23643353 on Build GRCh38
rs565711839 Chr.16: 23643227-23643227 on Build GRCh38
rs958943936 Chr.16: 23643226-23643226 on Build GRCh38
rs965663077 Chr.16: 23643444-23643444 on Build GRCh38
rs371336670 Chr.16: 23643207-23643208 on Build GRCh38
rs751086676 Chr.16: 23643032-23643032 on Build GRCh38
rs1002897789 Chr.16: 23643046-23643046 on Build GRCh38
rs546594210 Chr.16: 23643066-23643066 on Build GRCh38
rs1030169292 Chr.16: 23643187-23643187 on Build GRCh38
rs1038304367 Chr.16: 23643202-23643202 on Build GRCh38
rs1002845529 Chr.16: 23643317-23643317 on Build GRCh38
rs921633602 Chr.16: 23643483-23643483 on Build GRCh38
rs931720179 Chr.16: 23643484-23643484 on Build GRCh38
rs920142421 Chr.16: 23643071-23643071 on Build GRCh38
rs1054793888 Chr.16: 23643253-23643253 on Build GRCh38
rs893008518 Chr.16: 23643277-23643277 on Build GRCh38
rs55844967 Chr.16: 23643207-23643207 on Build GRCh38
rs865958510 Chr.16: 23643332-23643332 on Build GRCh38
rs557162972 Chr.16: 23643352-23643352 on Build GRCh38
rs56139098 Chr.16: 23643201-23643201 on Build GRCh38
rs890310912 Chr.16: 23643461-23643461 on Build GRCh38
rs768841757 Chr.16: 23643060-23643060 on Build GRCh38
rs201102012 Chr.16: 23643201-23643203 on Build GRCh38
rs747068474 Chr.16: 23643073-23643073 on Build GRCh38
rs528459331 Chr.16: 23643075-23643075 on Build GRCh38
rs550728629 Chr.16: 23643197-23643197 on Build GRCh38
rs1041645117 Chr.16: 23643280-23643280 on Build GRCh38
rs56077886 Chr.16: 23643279-23643279 on Build GRCh38
rs1019896450 Chr.16: 23643442-23643442 on Build GRCh38
rs531146083 Chr.16: 23643195-23643195 on Build GRCh38
rs760653407 Chr.16: 23643454-23643454 on Build GRCh38
rs894558213 Chr.16: 23643382-23643382 on Build GRCh38
rs59061540 Chr.16: 23643231-23643231 on Build GRCh38
rs995646143 Chr.16: 23643384-23643384 on Build GRCh38
rs975755008 Chr.16: 23643452-23643452 on Build GRCh38
rs1015397610 Chr.16: 23643192-23643192 on Build GRCh38
rs546352137 Chr.16: 23643358-23643358 on Build GRCh38
rs146108494 Chr.16: 23643051-23643051 on Build GRCh38
rs535457560 Chr.16: 23643230-23643230 on Build GRCh38
rs983005152 Chr.16: 23643419-23643419 on Build GRCh38
rs201032498 Chr.16: 23643194-23643195 on Build GRCh38
rs374046641 Chr.16: 23643074-23643074 on Build GRCh38
rs761687132 Chr.16: 23643070-23643071 on Build GRCh38
rs780913865 Chr.16: 23643174-23643174 on Build GRCh38
rs386789820 Chr.16: 23643201-23643202 on Build GRCh38
rs558204608 Chr.16: 23643470-23643470 on Build GRCh38
rs703769 Chr.16: 23643188-23643188 on Build GRCh38
rs749164288 Chr.16: 23643061-23643061 on Build GRCh38
rs954986649 Chr.16: 23643187-23643187 on Build GRCh38
rs951608980 Chr.16: 23643405-23643405 on Build GRCh38
rs386789821 Chr.16: 23643201-23643203 on Build GRCh38
rs963411247 Chr.16: 23643203-23643204 on Build GRCh38
rs548903720 Chr.16: 23643093-23643093 on Build GRCh38
rs989938756 Chr.16: 23643485-23643485 on Build GRCh38
rs772276721 Chr.16: 23643064-23643064 on Build GRCh38
rs754588165 Chr.16: 23643042-23643042 on Build GRCh38
rs779922842 Chr.16: 23643026-23643026 on Build GRCh38

View Full Product Details