Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 24255788-24256282 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CACNG3
Entrez Gene ID
10368
Gene Name
calcium voltage-gated channel auxiliary subunit gamma 3
Gene Aliases
-
UniGene
Hs.7235
Gene Chromosome Location
Chr.16: 24255553-24362416 on Build GRCh38
Transcripts
NM_006539.3

SNP Detail

SNP ID Chromosome Location
rs1054486056 Chr.16: 24256023-24256023 on Build GRCh38
rs888254447 Chr.16: 24255904-24255904 on Build GRCh38
rs1040812156 Chr.16: 24255842-24255842 on Build GRCh38
rs958625706 Chr.16: 24255917-24255917 on Build GRCh38
rs531363440 Chr.16: 24256072-24256072 on Build GRCh38
rs866253188 Chr.16: 24255907-24255907 on Build GRCh38
rs944722156 Chr.16: 24255837-24255837 on Build GRCh38
rs968310693 Chr.16: 24256192-24256192 on Build GRCh38
rs956416612 Chr.16: 24255946-24255946 on Build GRCh38
rs912652763 Chr.16: 24255960-24255960 on Build GRCh38
rs572338290 Chr.16: 24255877-24255877 on Build GRCh38
rs905026025 Chr.16: 24256118-24256118 on Build GRCh38
rs527880748 Chr.16: 24256165-24256165 on Build GRCh38
rs995179848 Chr.16: 24256115-24256115 on Build GRCh38
rs921347078 Chr.16: 24256197-24256197 on Build GRCh38
rs944755282 Chr.16: 24256024-24256024 on Build GRCh38
rs906970382 Chr.16: 24256075-24256075 on Build GRCh38
rs994452642 Chr.16: 24255867-24255868 on Build GRCh38
rs886904074 Chr.16: 24256247-24256247 on Build GRCh38
rs910628864 Chr.16: 24256019-24256019 on Build GRCh38
rs116450476 Chr.16: 24255900-24255900 on Build GRCh38
rs138438483 Chr.16: 24256155-24256155 on Build GRCh38
rs879431023 Chr.16: 24256244-24256244 on Build GRCh38
rs944084461 Chr.16: 24255965-24255965 on Build GRCh38
rs554105254 Chr.16: 24255928-24255928 on Build GRCh38
rs374896764 Chr.16: 24255933-24255933 on Build GRCh38
rs910659886 Chr.16: 24255827-24255827 on Build GRCh38
rs951293267 Chr.16: 24256173-24256173 on Build GRCh38
rs914142486 Chr.16: 24256258-24256258 on Build GRCh38
rs936669020 Chr.16: 24256005-24256005 on Build GRCh38
rs1004374009 Chr.16: 24256251-24256251 on Build GRCh38
rs11646957 Chr.16: 24255988-24255988 on Build GRCh38
rs540459044 Chr.16: 24255926-24255926 on Build GRCh38
rs372369248 Chr.16: 24256205-24256205 on Build GRCh38
rs763696065 Chr.16: 24255847-24255847 on Build GRCh38
rs7186210 Chr.16: 24256090-24256090 on Build GRCh38
rs1027350271 Chr.16: 24256120-24256120 on Build GRCh38
rs975494460 Chr.16: 24256195-24256195 on Build GRCh38
rs968169254 Chr.16: 24255994-24255994 on Build GRCh38
rs1004453779 Chr.16: 24256181-24256181 on Build GRCh38
rs1040465358 Chr.16: 24256040-24256040 on Build GRCh38
rs958315666 Chr.16: 24256233-24256233 on Build GRCh38
rs566641675 Chr.16: 24256112-24256112 on Build GRCh38
rs542951804 Chr.16: 24255985-24255985 on Build GRCh38
rs1020490058 Chr.16: 24255889-24255889 on Build GRCh38
rs1019381391 Chr.16: 24256188-24256188 on Build GRCh38
rs544763555 Chr.16: 24256079-24256079 on Build GRCh38
rs1009216937 Chr.16: 24256177-24256177 on Build GRCh38
rs914152625 Chr.16: 24256046-24256046 on Build GRCh38
rs982627838 Chr.16: 24255952-24255952 on Build GRCh38
rs897834885 Chr.16: 24255865-24255865 on Build GRCh38
rs1005379012 Chr.16: 24256097-24256097 on Build GRCh38
rs965444624 Chr.16: 24256190-24256190 on Build GRCh38
rs945544936 Chr.16: 24256050-24256050 on Build GRCh38
rs147303074 Chr.16: 24255911-24255911 on Build GRCh38

View Full Product Details