Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 29744071-29744564 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C16orf54
Entrez Gene ID
283897
Gene Name
chromosome 16 open reading frame 54
Gene Aliases
-
UniGene
Hs.331095
Gene Chromosome Location
Chr.16: 29742463-29746019 on Build GRCh38
Transcripts
NM_175900.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3110846 c.566G>T p.R189M Chr.16: 29744386-29744386 on Build GRCh38
213684 c.632C>G p.S211* Chr.16: 29744320-29744320 on Build GRCh38
1609179 c.487G>A p.A163T Chr.16: 29744465-29744465 on Build GRCh38
1609178 c.611G>A p.R204Q Chr.16: 29744341-29744341 on Build GRCh38
5584604 c.578G>A p.G193E Chr.16: 29744374-29744374 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1047192975 Chr.16: 29744470-29744470 on Build GRCh38
rs762218040 Chr.16: 29744289-29744289 on Build GRCh38
rs764700658 Chr.16: 29744331-29744331 on Build GRCh38
rs753800773 Chr.16: 29744325-29744325 on Build GRCh38
rs545636576 Chr.16: 29744208-29744208 on Build GRCh38
rs559469333 Chr.16: 29744436-29744436 on Build GRCh38
rs957256763 Chr.16: 29744472-29744472 on Build GRCh38
rs778501279 Chr.16: 29744243-29744243 on Build GRCh38
rs776454986 Chr.16: 29744420-29744420 on Build GRCh38
rs931490524 Chr.16: 29744138-29744138 on Build GRCh38
rs543740743 Chr.16: 29744520-29744520 on Build GRCh38
rs761730713 Chr.16: 29744428-29744428 on Build GRCh38
rs975367643 Chr.16: 29744479-29744479 on Build GRCh38
rs981449979 Chr.16: 29744368-29744368 on Build GRCh38
rs931494384 Chr.16: 29744424-29744424 on Build GRCh38
rs770135296 Chr.16: 29744268-29744268 on Build GRCh38
rs772533837 Chr.16: 29744442-29744442 on Build GRCh38
rs569754860 Chr.16: 29744529-29744529 on Build GRCh38
rs771166622 Chr.16: 29744296-29744296 on Build GRCh38
rs866981402 Chr.16: 29744435-29744435 on Build GRCh38
rs141257864 Chr.16: 29744176-29744176 on Build GRCh38
rs762540464 Chr.16: 29744458-29744458 on Build GRCh38
rs774614027 Chr.16: 29744297-29744297 on Build GRCh38
rs908310717 Chr.16: 29744500-29744500 on Build GRCh38
rs772533275 Chr.16: 29744335-29744335 on Build GRCh38
rs373101692 Chr.16: 29744532-29744532 on Build GRCh38
rs775079638 Chr.16: 29744306-29744306 on Build GRCh38
rs201665846 Chr.16: 29744299-29744299 on Build GRCh38
rs978485481 Chr.16: 29744377-29744377 on Build GRCh38
rs943755188 Chr.16: 29744509-29744509 on Build GRCh38
rs112310246 Chr.16: 29744255-29744255 on Build GRCh38
rs745963912 Chr.16: 29744405-29744405 on Build GRCh38
rs748615663 Chr.16: 29744260-29744260 on Build GRCh38
rs768406355 Chr.16: 29744419-29744419 on Build GRCh38
rs756524975 Chr.16: 29744367-29744367 on Build GRCh38
rs775549533 Chr.16: 29744305-29744305 on Build GRCh38
rs749333124 Chr.16: 29744288-29744288 on Build GRCh38
rs200313763 Chr.16: 29744380-29744380 on Build GRCh38
rs758114934 Chr.16: 29744253-29744253 on Build GRCh38
rs533076739 Chr.16: 29744441-29744441 on Build GRCh38
rs779355314 Chr.16: 29744466-29744466 on Build GRCh38
rs899766599 Chr.16: 29744225-29744225 on Build GRCh38
rs563935958 Chr.16: 29744217-29744217 on Build GRCh38
rs752949285 Chr.16: 29744353-29744353 on Build GRCh38
rs1015561894 Chr.16: 29744224-29744224 on Build GRCh38
rs779764093 Chr.16: 29744346-29744346 on Build GRCh38
rs958142216 Chr.16: 29744286-29744286 on Build GRCh38
rs551608693 Chr.16: 29744466-29744466 on Build GRCh38
rs754320429 Chr.16: 29744528-29744528 on Build GRCh38
rs756785016 Chr.16: 29744242-29744242 on Build GRCh38
rs748977284 Chr.16: 29744175-29744175 on Build GRCh38
rs779130047 Chr.16: 29744397-29744397 on Build GRCh38
rs199652256 Chr.16: 29744414-29744414 on Build GRCh38
rs750011567 Chr.16: 29744341-29744341 on Build GRCh38
rs764532004 Chr.16: 29744493-29744493 on Build GRCh38
rs35764197 Chr.16: 29744381-29744381 on Build GRCh38
rs750050863 Chr.16: 29744250-29744250 on Build GRCh38
rs190639748 Chr.16: 29744342-29744342 on Build GRCh38
rs1007864508 Chr.16: 29744205-29744205 on Build GRCh38
rs778439639 Chr.16: 29744280-29744280 on Build GRCh38
rs762981985 Chr.16: 29744328-29744328 on Build GRCh38
rs746359702 Chr.16: 29744251-29744251 on Build GRCh38
rs898697809 Chr.16: 29744137-29744137 on Build GRCh38
rs746738252 Chr.16: 29744415-29744415 on Build GRCh38
rs1057422787 Chr.16: 29744133-29744133 on Build GRCh38
rs763967256 Chr.16: 29744320-29744320 on Build GRCh38
rs754718868 Chr.16: 29744483-29744483 on Build GRCh38
rs760531312 Chr.16: 29744312-29744312 on Build GRCh38
rs774723249 Chr.16: 29744215-29744215 on Build GRCh38
rs757897274 Chr.16: 29744387-29744387 on Build GRCh38
rs765900368 Chr.16: 29744465-29744465 on Build GRCh38
rs1047280460 Chr.16: 29744161-29744161 on Build GRCh38
rs753289692 Chr.16: 29744236-29744236 on Build GRCh38

View Full Product Details