Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 29858313-29858856 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDIPT
Entrez Gene ID
10423
Gene Name
CDP-diacylglycerol--inositol 3-phosphatidyltransferase
Gene Aliases
PIS, PIS1
UniGene
Hs.121549
Gene Chromosome Location
Chr.16: 29858356-29863288 on Build GRCh38
Transcripts
NM_001286585.1 NM_001286586.1 NM_006319.4 XM_005255038.4

SNP Detail

SNP ID Chromosome Location
rs151156392 Chr.16: 29858666-29858666 on Build GRCh38
rs998634527 Chr.16: 29858538-29858538 on Build GRCh38
rs968896301 Chr.16: 29858503-29858503 on Build GRCh38
rs899065172 Chr.16: 29858530-29858530 on Build GRCh38
rs879072426 Chr.16: 29858468-29858468 on Build GRCh38
rs942114779 Chr.16: 29858338-29858338 on Build GRCh38
rs1801999 Chr.16: 29858652-29858652 on Build GRCh38
rs886235488 Chr.16: 29858616-29858616 on Build GRCh38
rs992455970 Chr.16: 29858642-29858642 on Build GRCh38
rs950253897 Chr.16: 29858407-29858407 on Build GRCh38
rs969957537 Chr.16: 29858671-29858671 on Build GRCh38
rs558271185 Chr.16: 29858507-29858507 on Build GRCh38
rs750121213 Chr.16: 29858412-29858412 on Build GRCh38
rs559233181 Chr.16: 29858539-29858539 on Build GRCh38
rs536667573 Chr.16: 29858376-29858376 on Build GRCh38
rs1036796995 Chr.16: 29858368-29858368 on Build GRCh38
rs950241060 Chr.16: 29858756-29858756 on Build GRCh38
rs1036455744 Chr.16: 29858722-29858722 on Build GRCh38
rs542627925 Chr.16: 29858823-29858823 on Build GRCh38
rs984391725 Chr.16: 29858673-29858673 on Build GRCh38
rs773596784 Chr.16: 29858664-29858664 on Build GRCh38
rs937633299 Chr.16: 29858512-29858512 on Build GRCh38
rs1019421226 Chr.16: 29858646-29858646 on Build GRCh38
rs889395054 Chr.16: 29858816-29858816 on Build GRCh38
rs1007746407 Chr.16: 29858829-29858829 on Build GRCh38
rs577508050 Chr.16: 29858647-29858647 on Build GRCh38
rs1802000 Chr.16: 29858428-29858428 on Build GRCh38
rs1026141119 Chr.16: 29858603-29858603 on Build GRCh38
rs373488119 Chr.16: 29858337-29858337 on Build GRCh38
rs563204139 Chr.16: 29858679-29858679 on Build GRCh38
rs1031811138 Chr.16: 29858545-29858545 on Build GRCh38
rs534275311 Chr.16: 29858432-29858432 on Build GRCh38
rs1054825938 Chr.16: 29858524-29858524 on Build GRCh38
rs779317304 Chr.16: 29858496-29858496 on Build GRCh38
rs17842263 Chr.16: 29858541-29858541 on Build GRCh38
rs933360275 Chr.16: 29858735-29858735 on Build GRCh38
rs913429314 Chr.16: 29858385-29858385 on Build GRCh38
rs748515993 Chr.16: 29858602-29858602 on Build GRCh38
rs554640761 Chr.16: 29858381-29858381 on Build GRCh38
rs994797169 Chr.16: 29858557-29858557 on Build GRCh38
rs879088352 Chr.16: 29858717-29858717 on Build GRCh38
rs1032997341 Chr.16: 29858704-29858704 on Build GRCh38
rs144315734 Chr.16: 29858352-29858352 on Build GRCh38
rs939818849 Chr.16: 29858707-29858707 on Build GRCh38
rs889360789 Chr.16: 29858408-29858408 on Build GRCh38
rs919292780 Chr.16: 29858725-29858725 on Build GRCh38
rs8335 Chr.16: 29858427-29858427 on Build GRCh38
rs190880274 Chr.16: 29858724-29858724 on Build GRCh38
rs897837870 Chr.16: 29858369-29858369 on Build GRCh38
rs772265377 Chr.16: 29858806-29858806 on Build GRCh38

View Full Product Details