Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 30888668-30889189 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCL7C
Entrez Gene ID
9274
Gene Name
BCL tumor suppressor 7C
Gene Aliases
-
UniGene
Hs.658547
Gene Chromosome Location
Chr.16: 30834041-30894550 on Build GRCh38
Transcripts
NM_001286526.1 NM_004765.3 XM_011545980.2 XM_017023885.1 XM_017023886.1 XM_017023887.1 XM_017023888.1 NR_110940.1 NR_110941.1 NR_110942.1

Gene Symbol
MIR762HG
Entrez Gene ID
101928736
Gene Name
MIR762 host gene
Gene Aliases
-
UniGene
Hs.303197
Gene Chromosome Location
Chr.16: 30875460-30895220 on Build GRCh38
Transcripts
NM_001286526.1 NM_004765.3 XM_011545980.2 XM_017023885.1 XM_017023886.1 XM_017023887.1 XM_017023888.1 NR_110940.1 NR_110941.1 NR_110942.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4983432 c.508C>G p.P170A Chr.16: 30888880-30888880 on Build GRCh38
6252084 c.443-3C>A p.? Chr.16: 30888948-30888948 on Build GRCh38
1588417 c.475G>A p.E159K Chr.16: 30888913-30888913 on Build GRCh38
970016 c.475G>A p.E159K Chr.16: 30888913-30888913 on Build GRCh38
4418204 c.474C>T p.D158D Chr.16: 30888914-30888914 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs967607232 Chr.16: 30888952-30888952 on Build GRCh38
rs571833350 Chr.16: 30889110-30889110 on Build GRCh38
rs563030831 Chr.16: 30888758-30888758 on Build GRCh38
rs1053471654 Chr.16: 30889109-30889109 on Build GRCh38
rs769432689 Chr.16: 30888969-30888969 on Build GRCh38
rs370439394 Chr.16: 30888834-30888834 on Build GRCh38
rs746817696 Chr.16: 30888926-30888926 on Build GRCh38
rs760359864 Chr.16: 30888833-30888833 on Build GRCh38
rs146577588 Chr.16: 30889011-30889011 on Build GRCh38
rs774941477 Chr.16: 30888974-30888974 on Build GRCh38
rs530455035 Chr.16: 30888781-30888781 on Build GRCh38
rs775530024 Chr.16: 30889162-30889162 on Build GRCh38
rs180795071 Chr.16: 30889141-30889141 on Build GRCh38
rs899558016 Chr.16: 30889039-30889045 on Build GRCh38
rs1000551303 Chr.16: 30889043-30889043 on Build GRCh38
rs1046965933 Chr.16: 30888950-30888950 on Build GRCh38
rs778012799 Chr.16: 30888877-30888877 on Build GRCh38
rs956970931 Chr.16: 30888761-30888761 on Build GRCh38
rs937068484 Chr.16: 30889019-30889019 on Build GRCh38
rs757354488 Chr.16: 30888899-30888899 on Build GRCh38
rs147859188 Chr.16: 30888941-30888941 on Build GRCh38
rs753063628 Chr.16: 30888865-30888865 on Build GRCh38
rs559687110 Chr.16: 30888940-30888940 on Build GRCh38
rs776248501 Chr.16: 30888835-30888835 on Build GRCh38
rs767937649 Chr.16: 30888860-30888860 on Build GRCh38
rs777819577 Chr.16: 30888809-30888809 on Build GRCh38
rs1000901449 Chr.16: 30888746-30888746 on Build GRCh38
rs760997942 Chr.16: 30888984-30888984 on Build GRCh38
rs1005003745 Chr.16: 30888807-30888807 on Build GRCh38
rs528386433 Chr.16: 30888965-30888965 on Build GRCh38
rs766943248 Chr.16: 30888842-30888842 on Build GRCh38
rs1039458727 Chr.16: 30889018-30889018 on Build GRCh38
rs749438291 Chr.16: 30888811-30888811 on Build GRCh38
rs767978633 Chr.16: 30888980-30888980 on Build GRCh38
rs747508341 Chr.16: 30888760-30888760 on Build GRCh38
rs756355385 Chr.16: 30888875-30888875 on Build GRCh38
rs569370880 Chr.16: 30889163-30889163 on Build GRCh38
rs370874249 Chr.16: 30888995-30888995 on Build GRCh38
rs761456804 Chr.16: 30888840-30888840 on Build GRCh38
rs779892732 Chr.16: 30888918-30888918 on Build GRCh38
rs577490311 Chr.16: 30888765-30888765 on Build GRCh38
rs771487951 Chr.16: 30888799-30888799 on Build GRCh38
rs988372679 Chr.16: 30888762-30888762 on Build GRCh38
rs937013058 Chr.16: 30888721-30888721 on Build GRCh38
rs375920333 Chr.16: 30888836-30888836 on Build GRCh38
rs759753344 Chr.16: 30888851-30888851 on Build GRCh38
rs3751871 Chr.16: 30888914-30888914 on Build GRCh38
rs774943019 Chr.16: 30888825-30888825 on Build GRCh38
rs753894773 Chr.16: 30888892-30888892 on Build GRCh38
rs776443654 Chr.16: 30888936-30888936 on Build GRCh38
rs758321179 Chr.16: 30888917-30888917 on Build GRCh38
rs370756870 Chr.16: 30888872-30888872 on Build GRCh38
rs996200112 Chr.16: 30888845-30888845 on Build GRCh38
rs778873411 Chr.16: 30888815-30888815 on Build GRCh38
rs995021242 Chr.16: 30888782-30888782 on Build GRCh38
rs376387404 Chr.16: 30888790-30888790 on Build GRCh38
rs778785561 Chr.16: 30888904-30888904 on Build GRCh38
rs368587974 Chr.16: 30888953-30888953 on Build GRCh38
rs768400764 Chr.16: 30888934-30888934 on Build GRCh38
rs115537999 Chr.16: 30888813-30888813 on Build GRCh38
rs775895055 Chr.16: 30888981-30888981 on Build GRCh38
rs770225848 Chr.16: 30889045-30889045 on Build GRCh38
rs764253898 Chr.16: 30888992-30888992 on Build GRCh38
rs752141705 Chr.16: 30888843-30888843 on Build GRCh38
rs745606036 Chr.16: 30888819-30888819 on Build GRCh38
rs1055400785 Chr.16: 30889029-30889029 on Build GRCh38
rs760075619 Chr.16: 30888977-30888977 on Build GRCh38
rs532329634 Chr.16: 30889131-30889131 on Build GRCh38
rs919578864 Chr.16: 30889083-30889083 on Build GRCh38

View Full Product Details