Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 30927645-30928146 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXL19
Entrez Gene ID
54620
Gene Name
F-box and leucine rich repeat protein 19
Gene Aliases
CXXC11, Fbl19, JHDM1C
UniGene
Hs.152149
Gene Chromosome Location
Chr.16: 30923055-30948783 on Build GRCh38
Transcripts
NM_001099784.2 NM_001282351.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7323422 c.524G>C p.S175T Chr.16: 30927800-30927800 on Build GRCh38
5510807 c.510C>T p.G170G Chr.16: 30927786-30927786 on Build GRCh38
3690939 c.591C>T p.G197G Chr.16: 30927867-30927867 on Build GRCh38
4578960 c.606_607insC p.E206fs*17 Chr.16: 30927882-30927883 on Build GRCh38
7284700 c.569C>T p.P190L Chr.16: 30927845-30927845 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778574166 Chr.16: 30927865-30927865 on Build GRCh38
rs571696165 Chr.16: 30927894-30927894 on Build GRCh38
rs977612703 Chr.16: 30928034-30928035 on Build GRCh38
rs993570948 Chr.16: 30927953-30927953 on Build GRCh38
rs750305215 Chr.16: 30927907-30927907 on Build GRCh38
rs765904924 Chr.16: 30927725-30927725 on Build GRCh38
rs140893916 Chr.16: 30927983-30927984 on Build GRCh38
rs777618399 Chr.16: 30927667-30927667 on Build GRCh38
rs999992527 Chr.16: 30928059-30928059 on Build GRCh38
rs764049236 Chr.16: 30927890-30927890 on Build GRCh38
rs774683996 Chr.16: 30927962-30927962 on Build GRCh38
rs749016312 Chr.16: 30927722-30927722 on Build GRCh38
rs369148968 Chr.16: 30927943-30927943 on Build GRCh38
rs938848811 Chr.16: 30928038-30928038 on Build GRCh38
rs1017468232 Chr.16: 30928037-30928037 on Build GRCh38
rs561899181 Chr.16: 30928120-30928120 on Build GRCh38
rs964554755 Chr.16: 30928047-30928047 on Build GRCh38
rs201252774 Chr.16: 30927867-30927867 on Build GRCh38
rs1046628201 Chr.16: 30927837-30927837 on Build GRCh38
rs925706551 Chr.16: 30927800-30927800 on Build GRCh38
rs1004809724 Chr.16: 30928109-30928110 on Build GRCh38
rs770412838 Chr.16: 30927832-30927840 on Build GRCh38
rs931326447 Chr.16: 30928091-30928091 on Build GRCh38
rs759805388 Chr.16: 30927771-30927771 on Build GRCh38
rs768296373 Chr.16: 30927982-30927983 on Build GRCh38
rs777252690 Chr.16: 30927664-30927668 on Build GRCh38
rs988558207 Chr.16: 30928085-30928085 on Build GRCh38
rs759905521 Chr.16: 30927969-30927969 on Build GRCh38
rs779861596 Chr.16: 30927908-30927908 on Build GRCh38
rs770363065 Chr.16: 30927883-30927883 on Build GRCh38
rs372521780 Chr.16: 30928081-30928081 on Build GRCh38
rs574467573 Chr.16: 30927699-30927699 on Build GRCh38
rs779700307 Chr.16: 30927879-30927879 on Build GRCh38
rs530179637 Chr.16: 30928062-30928062 on Build GRCh38
rs915969002 Chr.16: 30928079-30928079 on Build GRCh38
rs916898921 Chr.16: 30927819-30927819 on Build GRCh38
rs772215491 Chr.16: 30927976-30927976 on Build GRCh38
rs199881118 Chr.16: 30927912-30927912 on Build GRCh38
rs775720790 Chr.16: 30927985-30927985 on Build GRCh38
rs763236103 Chr.16: 30927748-30927748 on Build GRCh38
rs773876689 Chr.16: 30927675-30927675 on Build GRCh38
rs946318195 Chr.16: 30928023-30928023 on Build GRCh38
rs760694345 Chr.16: 30927993-30927993 on Build GRCh38
rs1027602681 Chr.16: 30927784-30927784 on Build GRCh38
rs751753002 Chr.16: 30927761-30927761 on Build GRCh38
rs866071756 Chr.16: 30927932-30927932 on Build GRCh38
rs1055920196 Chr.16: 30928042-30928042 on Build GRCh38
rs374725615 Chr.16: 30927720-30927720 on Build GRCh38
rs780803515 Chr.16: 30927923-30927923 on Build GRCh38
rs771888038 Chr.16: 30927870-30927870 on Build GRCh38
rs553383049 Chr.16: 30927882-30927882 on Build GRCh38
rs764054392 Chr.16: 30927994-30927994 on Build GRCh38
rs745385709 Chr.16: 30927882-30927883 on Build GRCh38
rs543267978 Chr.16: 30928112-30928112 on Build GRCh38
rs201873980 Chr.16: 30927843-30927843 on Build GRCh38
rs766585656 Chr.16: 30927749-30927749 on Build GRCh38
rs200068009 Chr.16: 30927839-30927847 on Build GRCh38
rs752713444 Chr.16: 30927889-30927889 on Build GRCh38
rs564041353 Chr.16: 30927979-30927979 on Build GRCh38
rs367877670 Chr.16: 30927777-30927777 on Build GRCh38
rs988896137 Chr.16: 30927807-30927807 on Build GRCh38
rs901715069 Chr.16: 30928115-30928115 on Build GRCh38
rs749574753 Chr.16: 30927802-30927802 on Build GRCh38
rs887078303 Chr.16: 30928107-30928107 on Build GRCh38
rs749093651 Chr.16: 30927673-30927673 on Build GRCh38
rs767784472 Chr.16: 30927888-30927888 on Build GRCh38
rs778538602 Chr.16: 30927902-30927902 on Build GRCh38
rs773916734 Chr.16: 30927883-30927883 on Build GRCh38
rs771935370 Chr.16: 30927906-30927907 on Build GRCh38
rs1036648584 Chr.16: 30928110-30928110 on Build GRCh38
rs773572642 Chr.16: 30927729-30927729 on Build GRCh38
rs752839993 Chr.16: 30927812-30927812 on Build GRCh38
rs753737723 Chr.16: 30927893-30927893 on Build GRCh38
rs368613889 Chr.16: 30927885-30927885 on Build GRCh38
rs375367725 Chr.16: 30927722-30927722 on Build GRCh38
rs201806396 Chr.16: 30927684-30927684 on Build GRCh38
rs757029811 Chr.16: 30927851-30927851 on Build GRCh38
rs964502926 Chr.16: 30927766-30927766 on Build GRCh38
rs377021646 Chr.16: 30927835-30927835 on Build GRCh38
rs775186468 Chr.16: 30927713-30927713 on Build GRCh38
rs1048445483 Chr.16: 30928105-30928106 on Build GRCh38
rs915969507 Chr.16: 30927866-30927866 on Build GRCh38
rs777585176 Chr.16: 30927822-30927822 on Build GRCh38
rs369197931 Chr.16: 30928011-30928011 on Build GRCh38
rs770558884 Chr.16: 30927674-30927674 on Build GRCh38

View Full Product Details