Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 31489813-31490307 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C16orf58
Entrez Gene ID
64755
Gene Name
chromosome 16 open reading frame 58
Gene Aliases
RUS
UniGene
Hs.499102
Gene Chromosome Location
Chr.16: 31489464-31508419 on Build GRCh38
Transcripts
NM_022744.3 XM_017023570.1 NM_003041.3 NR_130783.1 XM_006721072.3

Gene Symbol
SLC5A2
Entrez Gene ID
6524
Gene Name
solute carrier family 5 member 2
Gene Aliases
SGLT2
UniGene
Hs.709195
Gene Chromosome Location
Chr.16: 31482535-31490770 on Build GRCh38
Transcripts
NM_022744.3 XM_017023570.1 NM_003041.3 NR_130783.1 XM_006721072.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5493428 c.1673G>A p.R558H Chr.16: 31490111-31490111 on Build GRCh38
6783092 c.1702G>C p.E568Q Chr.16: 31490140-31490140 on Build GRCh38
5821688 c.1781T>A p.M594K Chr.16: 31490219-31490219 on Build GRCh38
4531085 c.1758G>A p.Q586Q Chr.16: 31490196-31490196 on Build GRCh38
3509282 c.1704G>A p.E568E Chr.16: 31490142-31490142 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775131993 Chr.16: 31490129-31490129 on Build GRCh38
rs143696144 Chr.16: 31489848-31489848 on Build GRCh38
rs775002648 Chr.16: 31490212-31490212 on Build GRCh38
rs777705985 Chr.16: 31490106-31490106 on Build GRCh38
rs374416729 Chr.16: 31490275-31490275 on Build GRCh38
rs779865507 Chr.16: 31490265-31490265 on Build GRCh38
rs775983861 Chr.16: 31490143-31490143 on Build GRCh38
rs143874850 Chr.16: 31490229-31490229 on Build GRCh38
rs746724730 Chr.16: 31490196-31490196 on Build GRCh38
rs765301127 Chr.16: 31490247-31490247 on Build GRCh38
rs776410459 Chr.16: 31490009-31490009 on Build GRCh38
rs886051963 Chr.16: 31490242-31490242 on Build GRCh38
rs371419116 Chr.16: 31490055-31490055 on Build GRCh38
rs74015546 Chr.16: 31489860-31489860 on Build GRCh38
rs982206127 Chr.16: 31490043-31490043 on Build GRCh38
rs999016308 Chr.16: 31489943-31489943 on Build GRCh38
rs1043649078 Chr.16: 31489917-31489917 on Build GRCh38
rs181079724 Chr.16: 31490147-31490147 on Build GRCh38
rs754037246 Chr.16: 31490239-31490239 on Build GRCh38
rs753032417 Chr.16: 31490158-31490158 on Build GRCh38
rs972155479 Chr.16: 31490040-31490040 on Build GRCh38
rs376197037 Chr.16: 31490084-31490084 on Build GRCh38
rs1012566854 Chr.16: 31489974-31489974 on Build GRCh38
rs758309955 Chr.16: 31490250-31490250 on Build GRCh38
rs1011247665 Chr.16: 31490033-31490033 on Build GRCh38
rs766818678 Chr.16: 31490080-31490080 on Build GRCh38
rs756642401 Chr.16: 31490077-31490077 on Build GRCh38
rs148186423 Chr.16: 31490232-31490232 on Build GRCh38
rs559072132 Chr.16: 31490061-31490061 on Build GRCh38
rs768049276 Chr.16: 31490223-31490223 on Build GRCh38
rs751006963 Chr.16: 31490066-31490066 on Build GRCh38
rs756222271 Chr.16: 31490094-31490094 on Build GRCh38
rs147052910 Chr.16: 31490113-31490113 on Build GRCh38
rs61661590 Chr.16: 31490024-31490024 on Build GRCh38
rs768270732 Chr.16: 31490142-31490142 on Build GRCh38
rs376265028 Chr.16: 31490276-31490276 on Build GRCh38
rs778201008 Chr.16: 31490208-31490209 on Build GRCh38
rs757315090 Chr.16: 31490168-31490168 on Build GRCh38
rs767926604 Chr.16: 31490157-31490157 on Build GRCh38
rs895500954 Chr.16: 31489952-31489952 on Build GRCh38
rs780719227 Chr.16: 31490272-31490272 on Build GRCh38
rs532838657 Chr.16: 31490081-31490081 on Build GRCh38
rs772673392 Chr.16: 31490281-31490281 on Build GRCh38
rs779812189 Chr.16: 31490184-31490184 on Build GRCh38
rs781385283 Chr.16: 31490058-31490059 on Build GRCh38
rs754752147 Chr.16: 31490267-31490267 on Build GRCh38
rs764251134 Chr.16: 31490234-31490234 on Build GRCh38
rs771033838 Chr.16: 31490004-31490004 on Build GRCh38
rs902896872 Chr.16: 31489920-31489920 on Build GRCh38
rs745742859 Chr.16: 31490178-31490178 on Build GRCh38
rs748199004 Chr.16: 31490071-31490087 on Build GRCh38
rs374900599 Chr.16: 31489962-31489962 on Build GRCh38
rs752061835 Chr.16: 31490151-31490151 on Build GRCh38
rs746975838 Chr.16: 31489891-31489891 on Build GRCh38
rs147022245 Chr.16: 31490133-31490133 on Build GRCh38
rs762542751 Chr.16: 31490137-31490137 on Build GRCh38
rs889392805 Chr.16: 31489890-31489890 on Build GRCh38
rs865816262 Chr.16: 31489994-31489994 on Build GRCh38
rs768164745 Chr.16: 31490203-31490203 on Build GRCh38
rs552837231 Chr.16: 31490085-31490085 on Build GRCh38
rs756273705 Chr.16: 31490166-31490166 on Build GRCh38
rs373371384 Chr.16: 31490064-31490064 on Build GRCh38
rs759992398 Chr.16: 31490156-31490156 on Build GRCh38
rs763557593 Chr.16: 31490146-31490146 on Build GRCh38
rs745687701 Chr.16: 31490122-31490122 on Build GRCh38
rs149525864 Chr.16: 31490211-31490211 on Build GRCh38
rs138861487 Chr.16: 31490128-31490128 on Build GRCh38
rs776308203 Chr.16: 31490210-31490210 on Build GRCh38
rs375719916 Chr.16: 31490035-31490035 on Build GRCh38
rs759434471 Chr.16: 31490027-31490027 on Build GRCh38
rs757291294 Chr.16: 31490240-31490240 on Build GRCh38
rs778838521 Chr.16: 31490170-31490170 on Build GRCh38
rs531524294 Chr.16: 31489957-31489957 on Build GRCh38
rs760115223 Chr.16: 31490213-31490213 on Build GRCh38
rs147697689 Chr.16: 31490130-31490130 on Build GRCh38
rs74394621 Chr.16: 31489836-31489836 on Build GRCh38
rs749300905 Chr.16: 31490110-31490110 on Build GRCh38
rs767191886 Chr.16: 31490056-31490056 on Build GRCh38
rs771925284 Chr.16: 31490181-31490181 on Build GRCh38
rs758817910 Chr.16: 31490077-31490077 on Build GRCh38
rs930887549 Chr.16: 31489834-31489834 on Build GRCh38
rs528819794 Chr.16: 31490036-31490036 on Build GRCh38
rs370100883 Chr.16: 31490266-31490266 on Build GRCh38
rs369673274 Chr.16: 31490111-31490111 on Build GRCh38
rs564291548 Chr.16: 31489892-31489892 on Build GRCh38
rs1035297390 Chr.16: 31489948-31489948 on Build GRCh38

View Full Product Details