Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 31490207-31490707 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C16orf58
Entrez Gene ID
64755
Gene Name
chromosome 16 open reading frame 58
Gene Aliases
RUS
UniGene
Hs.499102
Gene Chromosome Location
Chr.16: 31489464-31508419 on Build GRCh38
Transcripts
NM_022744.3 XM_017023570.1 NM_003041.3 NR_130783.1 XM_006721072.3

Gene Symbol
SLC5A2
Entrez Gene ID
6524
Gene Name
solute carrier family 5 member 2
Gene Aliases
SGLT2
UniGene
Hs.709195
Gene Chromosome Location
Chr.16: 31482535-31490770 on Build GRCh38
Transcripts
NM_022744.3 XM_017023570.1 NM_003041.3 NR_130783.1 XM_006721072.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6998650 c.1823G>A p.R608H Chr.16: 31490339-31490339 on Build GRCh38
471679 c.1862G>C p.G621A Chr.16: 31490378-31490378 on Build GRCh38
6576790 c.1800G>T p.Q600H Chr.16: 31490316-31490316 on Build GRCh38
1749590 c.1805C>T p.P602L Chr.16: 31490321-31490321 on Build GRCh38
7293674 c.1889A>G p.Q630R Chr.16: 31490405-31490405 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752505450 Chr.16: 31490404-31490404 on Build GRCh38
rs1036733905 Chr.16: 31490547-31490547 on Build GRCh38
rs978810285 Chr.16: 31490457-31490457 on Build GRCh38
rs756916093 Chr.16: 31490456-31490456 on Build GRCh38
rs752629971 Chr.16: 31490448-31490448 on Build GRCh38
rs777588302 Chr.16: 31490409-31490409 on Build GRCh38
rs775638719 Chr.16: 31490498-31490498 on Build GRCh38
rs765301127 Chr.16: 31490247-31490247 on Build GRCh38
rs200862764 Chr.16: 31490332-31490332 on Build GRCh38
rs768964115 Chr.16: 31490377-31490377 on Build GRCh38
rs371778694 Chr.16: 31490479-31490479 on Build GRCh38
rs933896617 Chr.16: 31490567-31490567 on Build GRCh38
rs774574743 Chr.16: 31490409-31490410 on Build GRCh38
rs753477563 Chr.16: 31490411-31490411 on Build GRCh38
rs761287089 Chr.16: 31490399-31490399 on Build GRCh38
rs532877244 Chr.16: 31490675-31490675 on Build GRCh38
rs755803144 Chr.16: 31490361-31490361 on Build GRCh38
rs368071047 Chr.16: 31490468-31490468 on Build GRCh38
rs777461500 Chr.16: 31490366-31490366 on Build GRCh38
rs758309955 Chr.16: 31490250-31490250 on Build GRCh38
rs771257793 Chr.16: 31490405-31490407 on Build GRCh38
rs144399155 Chr.16: 31490406-31490406 on Build GRCh38
rs141376808 Chr.16: 31490528-31490528 on Build GRCh38
rs750471836 Chr.16: 31490317-31490317 on Build GRCh38
rs145716653 Chr.16: 31490464-31490464 on Build GRCh38
rs778150076 Chr.16: 31490578-31490578 on Build GRCh38
rs557690622 Chr.16: 31490442-31490442 on Build GRCh38
rs61741237 Chr.16: 31490401-31490401 on Build GRCh38
rs1000598313 Chr.16: 31490618-31490618 on Build GRCh38
rs770034327 Chr.16: 31490384-31490384 on Build GRCh38
rs376265028 Chr.16: 31490276-31490276 on Build GRCh38
rs371220506 Chr.16: 31490300-31490300 on Build GRCh38
rs201577425 Chr.16: 31490491-31490491 on Build GRCh38
rs780719227 Chr.16: 31490272-31490272 on Build GRCh38
rs766964066 Chr.16: 31490466-31490466 on Build GRCh38
rs754752147 Chr.16: 31490267-31490267 on Build GRCh38
rs1031681131 Chr.16: 31490625-31490625 on Build GRCh38
rs368649902 Chr.16: 31490322-31490322 on Build GRCh38
rs376318525 Chr.16: 31490367-31490367 on Build GRCh38
rs905122501 Chr.16: 31490320-31490320 on Build GRCh38
rs201073991 Chr.16: 31490360-31490360 on Build GRCh38
rs576141726 Chr.16: 31490475-31490475 on Build GRCh38
rs1017578103 Chr.16: 31490444-31490444 on Build GRCh38
rs61742739 Chr.16: 31490477-31490477 on Build GRCh38
rs867270650 Chr.16: 31490605-31490605 on Build GRCh38
rs578168680 Chr.16: 31490362-31490362 on Build GRCh38
rs61746400 Chr.16: 31490321-31490321 on Build GRCh38
rs749691010 Chr.16: 31490579-31490579 on Build GRCh38
rs149843540 Chr.16: 31490437-31490437 on Build GRCh38
rs139986790 Chr.16: 31490431-31490431 on Build GRCh38
rs748585579 Chr.16: 31490473-31490473 on Build GRCh38
rs757291294 Chr.16: 31490240-31490240 on Build GRCh38
rs764033463 Chr.16: 31490509-31490509 on Build GRCh38
rs778330418 Chr.16: 31490460-31490460 on Build GRCh38
rs745592398 Chr.16: 31490418-31490419 on Build GRCh38
rs777035063 Chr.16: 31490378-31490378 on Build GRCh38
rs146166287 Chr.16: 31490417-31490417 on Build GRCh38
rs370100883 Chr.16: 31490266-31490266 on Build GRCh38
rs924779569 Chr.16: 31490681-31490681 on Build GRCh38
rs748536340 Chr.16: 31490421-31490421 on Build GRCh38
rs749662937 Chr.16: 31490372-31490376 on Build GRCh38
rs962089914 Chr.16: 31490341-31490341 on Build GRCh38
rs976338121 Chr.16: 31490657-31490657 on Build GRCh38
rs374416729 Chr.16: 31490275-31490275 on Build GRCh38
rs755934984 Chr.16: 31490593-31490593 on Build GRCh38
rs146209441 Chr.16: 31490338-31490338 on Build GRCh38
rs535031004 Chr.16: 31490393-31490393 on Build GRCh38
rs779865507 Chr.16: 31490265-31490265 on Build GRCh38
rs759427025 Chr.16: 31490445-31490445 on Build GRCh38
rs886051963 Chr.16: 31490242-31490242 on Build GRCh38
rs200935087 Chr.16: 31490484-31490484 on Build GRCh38
rs774466733 Chr.16: 31490443-31490443 on Build GRCh38
rs893453236 Chr.16: 31490462-31490462 on Build GRCh38
rs368278064 Chr.16: 31490416-31490416 on Build GRCh38
rs754037246 Chr.16: 31490239-31490239 on Build GRCh38
rs776022013 Chr.16: 31490287-31490287 on Build GRCh38
rs775869188 Chr.16: 31490373-31490373 on Build GRCh38
rs767517946 Chr.16: 31490447-31490447 on Build GRCh38
rs148186423 Chr.16: 31490232-31490232 on Build GRCh38
rs770514306 Chr.16: 31490372-31490372 on Build GRCh38
rs776541459 Chr.16: 31490516-31490516 on Build GRCh38
rs372404252 Chr.16: 31490418-31490418 on Build GRCh38
rs765350396 Chr.16: 31490304-31490304 on Build GRCh38
rs764455043 Chr.16: 31490292-31490292 on Build GRCh38
rs1027771675 Chr.16: 31490596-31490596 on Build GRCh38
rs760556743 Chr.16: 31490449-31490449 on Build GRCh38
rs772673392 Chr.16: 31490281-31490281 on Build GRCh38
rs535589155 Chr.16: 31490575-31490575 on Build GRCh38
rs972178516 Chr.16: 31490382-31490382 on Build GRCh38
rs760963048 Chr.16: 31490288-31490288 on Build GRCh38
rs761475754 Chr.16: 31490521-31490521 on Build GRCh38
rs751517591 Chr.16: 31490396-31490396 on Build GRCh38
rs764251134 Chr.16: 31490234-31490234 on Build GRCh38
rs199886583 Chr.16: 31490568-31490568 on Build GRCh38
rs1034845678 Chr.16: 31490546-31490546 on Build GRCh38
rs184367787 Chr.16: 31490583-31490583 on Build GRCh38
rs373179527 Chr.16: 31490545-31490545 on Build GRCh38
rs776789822 Chr.16: 31490296-31490296 on Build GRCh38
rs148924550 Chr.16: 31490467-31490467 on Build GRCh38
rs886051964 Chr.16: 31490411-31490413 on Build GRCh38
rs375802106 Chr.16: 31490508-31490508 on Build GRCh38
rs764966449 Chr.16: 31490526-31490526 on Build GRCh38
rs763032828 Chr.16: 31490394-31490394 on Build GRCh38
rs767260986 Chr.16: 31490402-31490402 on Build GRCh38
rs966660737 Chr.16: 31490630-31490630 on Build GRCh38
rs763978448 Chr.16: 31490455-31490455 on Build GRCh38
rs753259266 Chr.16: 31490556-31490556 on Build GRCh38
rs747550379 Chr.16: 31490374-31490374 on Build GRCh38
rs778096784 Chr.16: 31490476-31490476 on Build GRCh38
rs544637169 Chr.16: 31490638-31490638 on Build GRCh38
rs751455529 Chr.16: 31490326-31490326 on Build GRCh38
rs1017605901 Chr.16: 31490621-31490621 on Build GRCh38
rs774392134 Chr.16: 31490481-31490481 on Build GRCh38
rs114517938 Chr.16: 31490598-31490598 on Build GRCh38
rs756788287 Chr.16: 31490412-31490412 on Build GRCh38
rs77183830 Chr.16: 31490510-31490510 on Build GRCh38
rs369855362 Chr.16: 31490559-31490559 on Build GRCh38

View Full Product Details