Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 46802568-46803074 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C16orf87
Entrez Gene ID
388272
Gene Name
chromosome 16 open reading frame 87
Gene Aliases
-
UniGene
Hs.406551
Gene Chromosome Location
Chr.16: 46802047-46831162 on Build GRCh38
Transcripts
NM_001001436.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7098084 c.434G>T p.R145I Chr.16: 46802983-46802983 on Build GRCh38
4878606 c.414C>T p.V138V Chr.16: 46803003-46803003 on Build GRCh38
7136486 c.462C>G p.L154L Chr.16: 46802955-46802955 on Build GRCh38
7136487 c.444C>T p.I148I Chr.16: 46802973-46802973 on Build GRCh38
1629901 c.402C>A p.F134L Chr.16: 46803015-46803015 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs981844601 Chr.16: 46802911-46802911 on Build GRCh38
rs184173949 Chr.16: 46802634-46802634 on Build GRCh38
rs773116283 Chr.16: 46802929-46802929 on Build GRCh38
rs377742960 Chr.16: 46802948-46802948 on Build GRCh38
rs143952575 Chr.16: 46802881-46802881 on Build GRCh38
rs931287274 Chr.16: 46802640-46802640 on Build GRCh38
rs1035302788 Chr.16: 46802971-46802971 on Build GRCh38
rs747451014 Chr.16: 46802959-46802959 on Build GRCh38
rs774170179 Chr.16: 46802906-46802906 on Build GRCh38
rs1012743684 Chr.16: 46803002-46803002 on Build GRCh38
rs375594311 Chr.16: 46803000-46803000 on Build GRCh38
rs1041589373 Chr.16: 46802874-46802874 on Build GRCh38
rs893498038 Chr.16: 46802841-46802841 on Build GRCh38
rs113056950 Chr.16: 46802802-46802802 on Build GRCh38
rs28447050 Chr.16: 46802830-46802830 on Build GRCh38
rs774636981 Chr.16: 46803008-46803008 on Build GRCh38
rs529566336 Chr.16: 46802787-46802788 on Build GRCh38
rs559127082 Chr.16: 46802639-46802639 on Build GRCh38
rs999767422 Chr.16: 46802803-46802803 on Build GRCh38
rs755068527 Chr.16: 46802943-46802943 on Build GRCh38
rs1010546447 Chr.16: 46802851-46802851 on Build GRCh38
rs978162001 Chr.16: 46802591-46802591 on Build GRCh38
rs1035045803 Chr.16: 46802613-46802613 on Build GRCh38
rs375842624 Chr.16: 46802686-46802686 on Build GRCh38
rs1023639823 Chr.16: 46803014-46803014 on Build GRCh38
rs767222337 Chr.16: 46802926-46802926 on Build GRCh38
rs777990199 Chr.16: 46802953-46802953 on Build GRCh38
rs758716790 Chr.16: 46802952-46802952 on Build GRCh38
rs930951020 Chr.16: 46802686-46802687 on Build GRCh38
rs952108950 Chr.16: 46803026-46803026 on Build GRCh38
rs1011498797 Chr.16: 46802857-46802857 on Build GRCh38
rs191926883 Chr.16: 46802616-46802616 on Build GRCh38
rs982366963 Chr.16: 46802614-46802614 on Build GRCh38
rs756089920 Chr.16: 46802995-46802995 on Build GRCh38
rs940590774 Chr.16: 46802627-46802627 on Build GRCh38
rs556235675 Chr.16: 46802811-46802811 on Build GRCh38
rs368374110 Chr.16: 46803003-46803003 on Build GRCh38
rs942767378 Chr.16: 46802687-46802687 on Build GRCh38
rs1041613201 Chr.16: 46802796-46802796 on Build GRCh38
rs768298577 Chr.16: 46803015-46803015 on Build GRCh38
rs867238818 Chr.16: 46802853-46802853 on Build GRCh38
rs971970593 Chr.16: 46802628-46802628 on Build GRCh38
rs544455080 Chr.16: 46802884-46802884 on Build GRCh38
rs760506131 Chr.16: 46802932-46802932 on Build GRCh38
rs753874419 Chr.16: 46802942-46802942 on Build GRCh38
rs752863704 Chr.16: 46802951-46802951 on Build GRCh38
rs969552359 Chr.16: 46802887-46802887 on Build GRCh38
rs773989833 Chr.16: 46803045-46803045 on Build GRCh38
rs908054330 Chr.16: 46802687-46802687 on Build GRCh38
rs1048369148 Chr.16: 46802642-46802642 on Build GRCh38
rs188277260 Chr.16: 46802912-46802912 on Build GRCh38
rs766343233 Chr.16: 46802937-46802937 on Build GRCh38

View Full Product Details