Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 46956049-46956567 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJA2
Entrez Gene ID
10294
Gene Name
DnaJ heat shock protein family (Hsp40) member A2
Gene Aliases
CPR3, DJ3, DJA2, DNAJ, DNJ3, HIRIP4, PRO3015, RDJ2
UniGene
Hs.368078
Gene Chromosome Location
Chr.16: 46955362-46973714 on Build GRCh38
Transcripts
NM_005880.3

SNP Detail

SNP ID Chromosome Location
rs747233946 Chr.16: 46956437-46956437 on Build GRCh38
rs74019104 Chr.16: 46956438-46956438 on Build GRCh38
rs1000870635 Chr.16: 46956183-46956184 on Build GRCh38
rs912297305 Chr.16: 46956310-46956310 on Build GRCh38
rs142894389 Chr.16: 46956482-46956482 on Build GRCh38
rs536057049 Chr.16: 46956476-46956476 on Build GRCh38
rs907281555 Chr.16: 46956498-46956498 on Build GRCh38
rs1034170590 Chr.16: 46956198-46956198 on Build GRCh38
rs1012346226 Chr.16: 46956255-46956255 on Build GRCh38
rs571450068 Chr.16: 46956407-46956407 on Build GRCh38
rs767668312 Chr.16: 46956437-46956438 on Build GRCh38
rs569711853 Chr.16: 46956121-46956121 on Build GRCh38
rs986504538 Chr.16: 46956309-46956309 on Build GRCh38
rs929621754 Chr.16: 46956303-46956303 on Build GRCh38
rs535003952 Chr.16: 46956359-46956359 on Build GRCh38
rs970758231 Chr.16: 46956287-46956287 on Build GRCh38
rs572630337 Chr.16: 46956502-46956502 on Build GRCh38
rs918219571 Chr.16: 46956302-46956302 on Build GRCh38
rs75094037 Chr.16: 46956450-46956452 on Build GRCh38
rs57637732 Chr.16: 46956343-46956343 on Build GRCh38
rs979885359 Chr.16: 46956272-46956272 on Build GRCh38
rs900742150 Chr.16: 46956175-46956175 on Build GRCh38
rs944854252 Chr.16: 46956134-46956134 on Build GRCh38
rs983671333 Chr.16: 46956293-46956293 on Build GRCh38
rs398029261 Chr.16: 46956452-46956452 on Build GRCh38
rs34990708 Chr.16: 46956323-46956323 on Build GRCh38
rs532778455 Chr.16: 46956328-46956328 on Build GRCh38
rs764201526 Chr.16: 46956104-46956108 on Build GRCh38
rs973553626 Chr.16: 46956358-46956358 on Build GRCh38
rs201781640 Chr.16: 46956453-46956453 on Build GRCh38
rs892078953 Chr.16: 46956396-46956396 on Build GRCh38
rs953222369 Chr.16: 46956393-46956393 on Build GRCh38
rs556984584 Chr.16: 46956452-46956452 on Build GRCh38
rs971491543 Chr.16: 46956259-46956259 on Build GRCh38
rs890146151 Chr.16: 46956119-46956119 on Build GRCh38
rs1034162608 Chr.16: 46956513-46956513 on Build GRCh38
rs1010240846 Chr.16: 46956241-46956242 on Build GRCh38
rs982801725 Chr.16: 46956471-46956471 on Build GRCh38
rs138409843 Chr.16: 46956135-46956135 on Build GRCh38
rs547046691 Chr.16: 46956386-46956386 on Build GRCh38
rs925855138 Chr.16: 46956522-46956522 on Build GRCh38
rs549474123 Chr.16: 46956131-46956131 on Build GRCh38
rs35686106 Chr.16: 46956166-46956167 on Build GRCh38
rs936229168 Chr.16: 46956365-46956365 on Build GRCh38
rs1055126017 Chr.16: 46956390-46956390 on Build GRCh38
rs575367186 Chr.16: 46956470-46956470 on Build GRCh38
rs35534956 Chr.16: 46956438-46956438 on Build GRCh38
rs72812457 Chr.16: 46956128-46956128 on Build GRCh38
rs577918644 Chr.16: 46956439-46956439 on Build GRCh38
rs891795800 Chr.16: 46956220-46956220 on Build GRCh38
rs1039176837 Chr.16: 46956344-46956344 on Build GRCh38
rs539837621 Chr.16: 46956539-46956539 on Build GRCh38
rs941295717 Chr.16: 46956088-46956088 on Build GRCh38
rs922134971 Chr.16: 46956366-46956366 on Build GRCh38
rs1032991759 Chr.16: 46956184-46956184 on Build GRCh38
rs895208282 Chr.16: 46956236-46956236 on Build GRCh38
rs1010188394 Chr.16: 46956446-46956446 on Build GRCh38
rs200182056 Chr.16: 46956460-46956460 on Build GRCh38
rs766693677 Chr.16: 46956284-46956284 on Build GRCh38

View Full Product Details