Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 46956819-46957322 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJA2
Entrez Gene ID
10294
Gene Name
DnaJ heat shock protein family (Hsp40) member A2
Gene Aliases
CPR3, DJ3, DJA2, DNAJ, DNJ3, HIRIP4, PRO3015, RDJ2
UniGene
Hs.368078
Gene Chromosome Location
Chr.16: 46955362-46973714 on Build GRCh38
Transcripts
NM_005880.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6839058 c.1108G>C p.E370Q Chr.16: 46957160-46957160 on Build GRCh38
3509722 c.1070C>T p.S357F Chr.16: 46957198-46957198 on Build GRCh38
6864120 c.1071T>C p.S357S Chr.16: 46957197-46957197 on Build GRCh38
970799 c.1060C>A p.L354I Chr.16: 46957208-46957208 on Build GRCh38
970798 c.1177G>T p.D393Y Chr.16: 46957091-46957091 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1008169302 Chr.16: 46956968-46956968 on Build GRCh38
rs957045430 Chr.16: 46956989-46956989 on Build GRCh38
rs776054413 Chr.16: 46957228-46957228 on Build GRCh38
rs779768447 Chr.16: 46956951-46956951 on Build GRCh38
rs751394241 Chr.16: 46957258-46957258 on Build GRCh38
rs531616417 Chr.16: 46957247-46957247 on Build GRCh38
rs764018420 Chr.16: 46957128-46957128 on Build GRCh38
rs991599545 Chr.16: 46957059-46957059 on Build GRCh38
rs749619418 Chr.16: 46957026-46957026 on Build GRCh38
rs192293812 Chr.16: 46956872-46956872 on Build GRCh38
rs12836 Chr.16: 46956869-46956869 on Build GRCh38
rs375218877 Chr.16: 46956969-46956969 on Build GRCh38
rs915697500 Chr.16: 46956862-46956862 on Build GRCh38
rs757101179 Chr.16: 46957264-46957264 on Build GRCh38
rs781464561 Chr.16: 46956981-46956981 on Build GRCh38
rs775955552 Chr.16: 46957022-46957022 on Build GRCh38
rs200288545 Chr.16: 46957105-46957105 on Build GRCh38
rs930658535 Chr.16: 46957280-46957280 on Build GRCh38
rs908391216 Chr.16: 46957152-46957152 on Build GRCh38
rs981662210 Chr.16: 46957256-46957256 on Build GRCh38
rs746067266 Chr.16: 46956982-46956982 on Build GRCh38
rs751421502 Chr.16: 46957129-46957129 on Build GRCh38
rs953797075 Chr.16: 46956939-46956939 on Build GRCh38
rs770161090 Chr.16: 46957020-46957020 on Build GRCh38
rs542651256 Chr.16: 46956853-46956853 on Build GRCh38
rs759151550 Chr.16: 46957101-46957101 on Build GRCh38
rs755208295 Chr.16: 46957253-46957253 on Build GRCh38
rs967072497 Chr.16: 46956950-46956950 on Build GRCh38
rs762707693 Chr.16: 46957253-46957253 on Build GRCh38
rs899164889 Chr.16: 46956920-46956920 on Build GRCh38
rs781374497 Chr.16: 46957170-46957170 on Build GRCh38
rs372263272 Chr.16: 46957191-46957191 on Build GRCh38
rs745975632 Chr.16: 46957177-46957177 on Build GRCh38
rs758118666 Chr.16: 46957124-46957124 on Build GRCh38
rs751867588 Chr.16: 46957229-46957232 on Build GRCh38
rs749578018 Chr.16: 46957192-46957192 on Build GRCh38
rs772601096 Chr.16: 46957219-46957219 on Build GRCh38
rs748577877 Chr.16: 46957217-46957217 on Build GRCh38
rs951248964 Chr.16: 46956958-46956958 on Build GRCh38
rs538659082 Chr.16: 46956912-46956912 on Build GRCh38
rs114487971 Chr.16: 46956947-46956947 on Build GRCh38
rs369435063 Chr.16: 46957251-46957251 on Build GRCh38
rs184637288 Chr.16: 46957270-46957270 on Build GRCh38
rs995730138 Chr.16: 46956944-46956944 on Build GRCh38
rs915829591 Chr.16: 46957060-46957060 on Build GRCh38
rs774703657 Chr.16: 46957083-46957083 on Build GRCh38
rs571576125 Chr.16: 46956881-46956881 on Build GRCh38
rs1029238397 Chr.16: 46956938-46956938 on Build GRCh38
rs764789212 Chr.16: 46957250-46957250 on Build GRCh38
rs547008191 Chr.16: 46956879-46956879 on Build GRCh38
rs1050879866 Chr.16: 46957286-46957286 on Build GRCh38
rs995053060 Chr.16: 46956929-46956929 on Build GRCh38
rs376667429 Chr.16: 46957100-46957100 on Build GRCh38
rs769147675 Chr.16: 46957069-46957069 on Build GRCh38
rs762444482 Chr.16: 46957084-46957084 on Build GRCh38
rs765100242 Chr.16: 46957024-46957025 on Build GRCh38
rs907230914 Chr.16: 46956861-46956861 on Build GRCh38
rs765773216 Chr.16: 46957088-46957088 on Build GRCh38
rs977159622 Chr.16: 46956966-46956966 on Build GRCh38
rs756336225 Chr.16: 46957178-46957178 on Build GRCh38
rs554388659 Chr.16: 46957050-46957050 on Build GRCh38
rs187678402 Chr.16: 46956844-46956844 on Build GRCh38
rs1032600763 Chr.16: 46956987-46956987 on Build GRCh38
rs752435428 Chr.16: 46957119-46957119 on Build GRCh38
rs757270298 Chr.16: 46957165-46957165 on Build GRCh38
rs774815723 Chr.16: 46957196-46957196 on Build GRCh38
rs550817486 Chr.16: 46956945-46956945 on Build GRCh38
rs1055718645 Chr.16: 46956856-46956856 on Build GRCh38
rs1004982832 Chr.16: 46956896-46956896 on Build GRCh38
rs369681262 Chr.16: 46957257-46957257 on Build GRCh38
rs768894461 Chr.16: 46957193-46957193 on Build GRCh38

View Full Product Details