Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 48176842-48177338 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCC11
Entrez Gene ID
85320
Gene Name
ATP binding cassette subfamily C member 11
Gene Aliases
EWWD, MRP8, WW
UniGene
Hs.652267
Gene Chromosome Location
Chr.16: 48164842-48249973 on Build GRCh38
Transcripts
NM_032583.3 NM_033151.3 NM_145186.2 XM_011523397.1 XM_011523398.2 XM_017023795.1 XM_017023796.1 XM_017023797.1 XM_017023798.1 XM_017023799.1 XM_017023800.1 XM_017023801.1 XM_017023802.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1324433 c.3527G>A p.R1176K Chr.16: 48176935-48176935 on Build GRCh38
7353604 c.3427T>C p.F1143L Chr.16: 48177035-48177035 on Build GRCh38
3818028 c.3466A>C p.T1156P Chr.16: 48176996-48176996 on Build GRCh38
3509834 c.3472C>T p.L1158F Chr.16: 48176990-48176990 on Build GRCh38
4754578 c.3497G>A p.R1166H Chr.16: 48176965-48176965 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs184691369 Chr.16: 48177271-48177271 on Build GRCh38
rs746672566 Chr.16: 48177051-48177051 on Build GRCh38
rs907387624 Chr.16: 48177156-48177156 on Build GRCh38
rs930604849 Chr.16: 48176872-48176872 on Build GRCh38
rs943995904 Chr.16: 48177201-48177201 on Build GRCh38
rs763480791 Chr.16: 48177121-48177121 on Build GRCh38
rs374295166 Chr.16: 48176965-48176965 on Build GRCh38
rs781765054 Chr.16: 48176883-48176883 on Build GRCh38
rs146149363 Chr.16: 48176966-48176966 on Build GRCh38
rs567931559 Chr.16: 48177208-48177208 on Build GRCh38
rs768300581 Chr.16: 48176888-48176888 on Build GRCh38
rs746761722 Chr.16: 48176963-48176963 on Build GRCh38
rs747668188 Chr.16: 48176955-48176955 on Build GRCh38
rs761918114 Chr.16: 48177089-48177089 on Build GRCh38
rs773337191 Chr.16: 48177117-48177117 on Build GRCh38
rs769775213 Chr.16: 48176969-48176969 on Build GRCh38
rs943960695 Chr.16: 48176991-48176991 on Build GRCh38
rs370633599 Chr.16: 48176943-48176943 on Build GRCh38
rs764283805 Chr.16: 48176927-48176927 on Build GRCh38
rs754325857 Chr.16: 48177116-48177116 on Build GRCh38
rs777435399 Chr.16: 48177050-48177050 on Build GRCh38
rs747770549 Chr.16: 48176895-48176895 on Build GRCh38
rs192098195 Chr.16: 48177195-48177195 on Build GRCh38
rs756893355 Chr.16: 48177147-48177147 on Build GRCh38
rs867614952 Chr.16: 48177228-48177228 on Build GRCh38
rs771748765 Chr.16: 48176957-48176957 on Build GRCh38
rs910400237 Chr.16: 48176907-48176907 on Build GRCh38
rs778543613 Chr.16: 48176947-48176947 on Build GRCh38
rs778118523 Chr.16: 48176930-48176930 on Build GRCh38
rs761123406 Chr.16: 48177103-48177103 on Build GRCh38
rs780946898 Chr.16: 48177060-48177060 on Build GRCh38
rs985493202 Chr.16: 48177192-48177192 on Build GRCh38
rs774455418 Chr.16: 48176984-48176984 on Build GRCh38
rs774915707 Chr.16: 48177149-48177150 on Build GRCh38
rs576765645 Chr.16: 48177267-48177267 on Build GRCh38
rs143002804 Chr.16: 48176958-48176958 on Build GRCh38
rs753522724 Chr.16: 48177227-48177227 on Build GRCh38
rs146373123 Chr.16: 48177086-48177086 on Build GRCh38
rs76500528 Chr.16: 48177215-48177215 on Build GRCh38
rs1053479240 Chr.16: 48177044-48177044 on Build GRCh38
rs1049739149 Chr.16: 48176970-48176970 on Build GRCh38
rs763093209 Chr.16: 48177174-48177174 on Build GRCh38
rs374297650 Chr.16: 48176994-48176994 on Build GRCh38
rs774525466 Chr.16: 48177087-48177087 on Build GRCh38
rs906207879 Chr.16: 48177281-48177281 on Build GRCh38
rs762193031 Chr.16: 48176985-48176985 on Build GRCh38
rs141607865 Chr.16: 48176993-48176993 on Build GRCh38
rs760097903 Chr.16: 48177282-48177282 on Build GRCh38
rs757572924 Chr.16: 48176932-48176932 on Build GRCh38
rs769721654 Chr.16: 48177068-48177068 on Build GRCh38
rs1027095922 Chr.16: 48177175-48177175 on Build GRCh38
rs756948208 Chr.16: 48177056-48177056 on Build GRCh38
rs748745969 Chr.16: 48176885-48176885 on Build GRCh38
rs556521347 Chr.16: 48177283-48177283 on Build GRCh38
rs1034727987 Chr.16: 48177170-48177170 on Build GRCh38
rs745724144 Chr.16: 48177067-48177067 on Build GRCh38
rs771670319 Chr.16: 48176897-48176897 on Build GRCh38
rs749337499 Chr.16: 48177073-48177073 on Build GRCh38
rs200025381 Chr.16: 48176874-48176874 on Build GRCh38
rs200083765 Chr.16: 48177154-48177154 on Build GRCh38
rs894873152 Chr.16: 48177049-48177049 on Build GRCh38
rs767908449 Chr.16: 48177094-48177094 on Build GRCh38
rs775300874 Chr.16: 48177071-48177071 on Build GRCh38
rs767963296 Chr.16: 48176987-48176987 on Build GRCh38
rs907490070 Chr.16: 48177112-48177112 on Build GRCh38
rs908709926 Chr.16: 48176905-48176905 on Build GRCh38
rs773044957 Chr.16: 48176899-48176899 on Build GRCh38
rs1054843158 Chr.16: 48177272-48177272 on Build GRCh38
rs763217587 Chr.16: 48176919-48176919 on Build GRCh38
rs768645355 Chr.16: 48177079-48177079 on Build GRCh38
rs766459488 Chr.16: 48177230-48177230 on Build GRCh38
rs1027346116 Chr.16: 48177191-48177191 on Build GRCh38
rs552190839 Chr.16: 48177102-48177102 on Build GRCh38
rs74869498 Chr.16: 48176931-48176931 on Build GRCh38
rs936473292 Chr.16: 48177019-48177019 on Build GRCh38
rs773706636 Chr.16: 48177098-48177098 on Build GRCh38
rs202078215 Chr.16: 48176903-48176903 on Build GRCh38
rs757861134 Chr.16: 48177038-48177038 on Build GRCh38
rs367800571 Chr.16: 48177136-48177136 on Build GRCh38
rs568719377 Chr.16: 48177108-48177108 on Build GRCh38
rs866202316 Chr.16: 48177177-48177177 on Build GRCh38
rs752187256 Chr.16: 48177017-48177017 on Build GRCh38
rs372658697 Chr.16: 48177160-48177160 on Build GRCh38
rs948949027 Chr.16: 48177298-48177298 on Build GRCh38
rs148224575 Chr.16: 48176975-48176975 on Build GRCh38
rs1042653391 Chr.16: 48177013-48177013 on Build GRCh38
rs750995040 Chr.16: 48176940-48176940 on Build GRCh38
rs537952184 Chr.16: 48177171-48177171 on Build GRCh38
rs770735735 Chr.16: 48176964-48176964 on Build GRCh38
rs771273792 Chr.16: 48177064-48177064 on Build GRCh38
rs759564176 Chr.16: 48176904-48176904 on Build GRCh38
rs763012072 Chr.16: 48176981-48176981 on Build GRCh38
rs559146548 Chr.16: 48177244-48177244 on Build GRCh38
rs778668533 Chr.16: 48176894-48176894 on Build GRCh38
rs185651615 Chr.16: 48176900-48176900 on Build GRCh38
rs1023057976 Chr.16: 48177076-48177076 on Build GRCh38
rs781743926 Chr.16: 48176938-48176938 on Build GRCh38
rs975948138 Chr.16: 48177245-48177245 on Build GRCh38
rs780893647 Chr.16: 48177158-48177158 on Build GRCh38
rs111327948 Chr.16: 48177240-48177240 on Build GRCh38

View Full Product Details