Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 48187123-48187533 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCC11
Entrez Gene ID
85320
Gene Name
ATP binding cassette subfamily C member 11
Gene Aliases
EWWD, MRP8, WW
UniGene
Hs.652267
Gene Chromosome Location
Chr.16: 48164842-48249973 on Build GRCh38
Transcripts
NM_032583.3 NM_033151.3 NM_145186.2 XM_011523397.1 XM_011523398.2 XM_017023795.1 XM_017023796.1 XM_017023797.1 XM_017023798.1 XM_017023799.1 XM_017023800.1 XM_017023801.1 XM_017023802.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2833010 c.2851G>A p.V951I Chr.16: 48187283-48187283 on Build GRCh38
3716937 c.2798T>A p.L933H Chr.16: 48187336-48187336 on Build GRCh38
1563104 c.2770G>A p.A924T Chr.16: 48187364-48187364 on Build GRCh38
970970 c.2850C>T p.A950A Chr.16: 48187284-48187284 on Build GRCh38
3927547 c.2931T>C p.Y977Y Chr.16: 48187203-48187203 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773761656 Chr.16: 48187421-48187421 on Build GRCh38
rs1000018855 Chr.16: 48187181-48187181 on Build GRCh38
rs536562760 Chr.16: 48187446-48187446 on Build GRCh38
rs368875106 Chr.16: 48187365-48187365 on Build GRCh38
rs1042003419 Chr.16: 48187152-48187152 on Build GRCh38
rs774785973 Chr.16: 48187305-48187305 on Build GRCh38
rs751246833 Chr.16: 48187452-48187452 on Build GRCh38
rs201181092 Chr.16: 48187283-48187283 on Build GRCh38
rs779781211 Chr.16: 48187354-48187365 on Build GRCh38
rs150094014 Chr.16: 48187271-48187271 on Build GRCh38
rs375180799 Chr.16: 48187420-48187420 on Build GRCh38
rs775682969 Chr.16: 48187287-48187287 on Build GRCh38
rs766978109 Chr.16: 48187438-48187438 on Build GRCh38
rs758242372 Chr.16: 48187183-48187184 on Build GRCh38
rs573436780 Chr.16: 48187453-48187453 on Build GRCh38
rs1055640422 Chr.16: 48187409-48187409 on Build GRCh38
rs750350095 Chr.16: 48187153-48187169 on Build GRCh38
rs750997339 Chr.16: 48187329-48187329 on Build GRCh38
rs754467204 Chr.16: 48187192-48187192 on Build GRCh38
rs753338581 Chr.16: 48187191-48187191 on Build GRCh38
rs553284869 Chr.16: 48187447-48187447 on Build GRCh38
rs1038169311 Chr.16: 48187308-48187308 on Build GRCh38
rs768086033 Chr.16: 48187328-48187328 on Build GRCh38
rs189467834 Chr.16: 48187341-48187341 on Build GRCh38
rs745915376 Chr.16: 48187279-48187279 on Build GRCh38
rs758894044 Chr.16: 48187289-48187289 on Build GRCh38
rs757983517 Chr.16: 48187360-48187360 on Build GRCh38
rs559803411 Chr.16: 48187297-48187297 on Build GRCh38
rs34926101 Chr.16: 48187148-48187149 on Build GRCh38
rs751888897 Chr.16: 48187371-48187371 on Build GRCh38
rs746872740 Chr.16: 48187257-48187257 on Build GRCh38
rs1015209183 Chr.16: 48187504-48187504 on Build GRCh38
rs752082488 Chr.16: 48187444-48187444 on Build GRCh38
rs138453043 Chr.16: 48187364-48187364 on Build GRCh38
rs113674902 Chr.16: 48187281-48187281 on Build GRCh38
rs41280943 Chr.16: 48187226-48187226 on Build GRCh38
rs371869786 Chr.16: 48187412-48187412 on Build GRCh38
rs770790668 Chr.16: 48187270-48187270 on Build GRCh38
rs79875006 Chr.16: 48187350-48187350 on Build GRCh38
rs780947119 Chr.16: 48187456-48187456 on Build GRCh38
rs370716853 Chr.16: 48187158-48187158 on Build GRCh38
rs55713504 Chr.16: 48187321-48187321 on Build GRCh38
rs777311991 Chr.16: 48187483-48187483 on Build GRCh38
rs761071932 Chr.16: 48187430-48187430 on Build GRCh38
rs763067200 Chr.16: 48187290-48187290 on Build GRCh38
rs769822967 Chr.16: 48187381-48187381 on Build GRCh38
rs778586275 Chr.16: 48187197-48187197 on Build GRCh38
rs765294261 Chr.16: 48187206-48187206 on Build GRCh38
rs369803247 Chr.16: 48187474-48187474 on Build GRCh38
rs868853392 Chr.16: 48187189-48187189 on Build GRCh38
rs764631134 Chr.16: 48187345-48187345 on Build GRCh38
rs925444516 Chr.16: 48187149-48187149 on Build GRCh38
rs904380193 Chr.16: 48187169-48187169 on Build GRCh38
rs966893173 Chr.16: 48187384-48187384 on Build GRCh38
rs374816951 Chr.16: 48187254-48187254 on Build GRCh38
rs762227083 Chr.16: 48187416-48187416 on Build GRCh38
rs951088516 Chr.16: 48187284-48187284 on Build GRCh38
rs904435056 Chr.16: 48187407-48187407 on Build GRCh38
rs139927866 Chr.16: 48187382-48187382 on Build GRCh38
rs143346912 Chr.16: 48187408-48187408 on Build GRCh38
rs922058695 Chr.16: 48187324-48187324 on Build GRCh38
rs747724821 Chr.16: 48187200-48187200 on Build GRCh38
rs1015571308 Chr.16: 48187293-48187293 on Build GRCh38
rs138369671 Chr.16: 48187273-48187273 on Build GRCh38
rs374188649 Chr.16: 48187196-48187196 on Build GRCh38
rs377303801 Chr.16: 48187150-48187150 on Build GRCh38
rs983737712 Chr.16: 48187286-48187286 on Build GRCh38

View Full Product Details