Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 50287940-50288437 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADCY7
Entrez Gene ID
113
Gene Name
adenylate cyclase 7
Gene Aliases
AC7
UniGene
Hs.513578
Gene Chromosome Location
Chr.16: 50246158-50318135 on Build GRCh38
Transcripts
NM_001114.4 NM_001286057.1 XM_011522835.2 XM_011522836.2 XM_011522837.2 XM_011522838.2 XM_011522839.2 XM_011522840.2 XM_011522842.2 XM_017022895.1 XM_017022896.1 XM_017022897.1 XM_017022898.1 XM_017022899.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6227666 c.17G>A p.R6H Chr.16: 50288196-50288196 on Build GRCh38
6002021 c.142G>A p.A48T Chr.16: 50288321-50288321 on Build GRCh38
7354546 c.151A>G p.I51V Chr.16: 50288330-50288330 on Build GRCh38
7322608 c.143C>A p.A48D Chr.16: 50288322-50288322 on Build GRCh38
3948573 c.30C>T p.N10N Chr.16: 50288209-50288209 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201090410 Chr.16: 50288306-50288306 on Build GRCh38
rs370314909 Chr.16: 50288091-50288091 on Build GRCh38
rs775075222 Chr.16: 50288396-50288396 on Build GRCh38
rs77518645 Chr.16: 50288262-50288262 on Build GRCh38
rs775634360 Chr.16: 50288157-50288157 on Build GRCh38
rs982870279 Chr.16: 50288277-50288277 on Build GRCh38
rs764318741 Chr.16: 50288202-50288204 on Build GRCh38
rs1034181278 Chr.16: 50288354-50288354 on Build GRCh38
rs968545803 Chr.16: 50287987-50287987 on Build GRCh38
rs1014225993 Chr.16: 50288012-50288012 on Build GRCh38
rs551578722 Chr.16: 50288118-50288118 on Build GRCh38
rs367714919 Chr.16: 50288317-50288317 on Build GRCh38
rs945972138 Chr.16: 50288363-50288363 on Build GRCh38
rs932235843 Chr.16: 50288313-50288313 on Build GRCh38
rs77503487 Chr.16: 50288398-50288398 on Build GRCh38
rs544344084 Chr.16: 50288003-50288003 on Build GRCh38
rs373051027 Chr.16: 50288105-50288105 on Build GRCh38
rs751338460 Chr.16: 50288402-50288402 on Build GRCh38
rs376341329 Chr.16: 50288114-50288114 on Build GRCh38
rs531003571 Chr.16: 50288405-50288405 on Build GRCh38
rs958634025 Chr.16: 50288016-50288016 on Build GRCh38
rs983617687 Chr.16: 50288069-50288069 on Build GRCh38
rs1025148390 Chr.16: 50288215-50288215 on Build GRCh38
rs769023727 Chr.16: 50288307-50288307 on Build GRCh38
rs757195749 Chr.16: 50288130-50288130 on Build GRCh38
rs762361791 Chr.16: 50288164-50288164 on Build GRCh38
rs772608421 Chr.16: 50288367-50288367 on Build GRCh38
rs374349100 Chr.16: 50288156-50288156 on Build GRCh38
rs199714954 Chr.16: 50288244-50288244 on Build GRCh38
rs745950378 Chr.16: 50288274-50288274 on Build GRCh38
rs148243317 Chr.16: 50288088-50288088 on Build GRCh38
rs1025200585 Chr.16: 50288031-50288031 on Build GRCh38
rs144769944 Chr.16: 50288305-50288305 on Build GRCh38
rs557064062 Chr.16: 50288163-50288163 on Build GRCh38
rs904592690 Chr.16: 50288336-50288336 on Build GRCh38
rs925748743 Chr.16: 50288378-50288378 on Build GRCh38
rs1042951308 Chr.16: 50288151-50288151 on Build GRCh38
rs950847905 Chr.16: 50288035-50288035 on Build GRCh38
rs765896604 Chr.16: 50288239-50288239 on Build GRCh38
rs139493357 Chr.16: 50288179-50288179 on Build GRCh38
rs745977969 Chr.16: 50288149-50288149 on Build GRCh38
rs1028742584 Chr.16: 50288290-50288290 on Build GRCh38
rs200293174 Chr.16: 50288145-50288145 on Build GRCh38
rs763780785 Chr.16: 50288401-50288401 on Build GRCh38
rs537134395 Chr.16: 50288161-50288161 on Build GRCh38
rs757159174 Chr.16: 50288255-50288255 on Build GRCh38
rs895114920 Chr.16: 50288196-50288196 on Build GRCh38
rs758047984 Chr.16: 50288224-50288224 on Build GRCh38
rs945856466 Chr.16: 50288150-50288150 on Build GRCh38
rs759080085 Chr.16: 50288383-50288383 on Build GRCh38
rs764927746 Chr.16: 50288195-50288195 on Build GRCh38
rs561124542 Chr.16: 50288004-50288004 on Build GRCh38
rs11644926 Chr.16: 50287967-50287967 on Build GRCh38
rs371889230 Chr.16: 50288318-50288318 on Build GRCh38
rs751373586 Chr.16: 50288245-50288245 on Build GRCh38
rs1042132967 Chr.16: 50288327-50288327 on Build GRCh38
rs752356585 Chr.16: 50288216-50288216 on Build GRCh38
rs367883473 Chr.16: 50288209-50288209 on Build GRCh38
rs112244195 Chr.16: 50288359-50288359 on Build GRCh38
rs765710324 Chr.16: 50288365-50288365 on Build GRCh38
rs756799073 Chr.16: 50288348-50288348 on Build GRCh38
rs963481999 Chr.16: 50288093-50288093 on Build GRCh38
rs916956666 Chr.16: 50288066-50288066 on Build GRCh38
rs548963798 Chr.16: 50288095-50288095 on Build GRCh38
rs764582125 Chr.16: 50288384-50288384 on Build GRCh38
rs909610076 Chr.16: 50288278-50288278 on Build GRCh38
rs28514808 Chr.16: 50288289-50288289 on Build GRCh38
rs1000314822 Chr.16: 50288338-50288338 on Build GRCh38
rs565165162 Chr.16: 50287979-50287979 on Build GRCh38
rs544253864 Chr.16: 50288379-50288379 on Build GRCh38
rs143102567 Chr.16: 50288002-50288002 on Build GRCh38
rs1010212571 Chr.16: 50288323-50288323 on Build GRCh38
rs61731918 Chr.16: 50288258-50288258 on Build GRCh38
rs200299180 Chr.16: 50288238-50288238 on Build GRCh38
rs774869842 Chr.16: 50288162-50288162 on Build GRCh38
rs867361478 Chr.16: 50288183-50288183 on Build GRCh38
rs369539863 Chr.16: 50288193-50288193 on Build GRCh38

View Full Product Details