Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 50319839-50320341 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRD7
Entrez Gene ID
29117
Gene Name
bromodomain containing 7
Gene Aliases
BP75, CELTIX1, NAG4
UniGene
Hs.437894
Gene Chromosome Location
Chr.16: 50317563-50368969 on Build GRCh38
Transcripts
NM_001173984.2 NM_013263.4 XM_011523046.2 XM_011523047.2 XM_011523048.2 XM_011523049.2 XM_011523050.2 XM_017023179.1 XM_017023180.1 XM_017023181.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
190399 c.1834C>T p.R612* Chr.16: 50319953-50319953 on Build GRCh38
5504203 c.1677C>A p.A559A Chr.16: 50320327-50320327 on Build GRCh38
6885256 c.1867G>T p.V623F Chr.16: 50319920-50319920 on Build GRCh38
6612741 c.1866C>T p.P622P Chr.16: 50319921-50319921 on Build GRCh38
5384951 c.1853C>T p.S618F Chr.16: 50319934-50319934 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369956085 Chr.16: 50320226-50320226 on Build GRCh38
rs770797884 Chr.16: 50320301-50320301 on Build GRCh38
rs796065051 Chr.16: 50319953-50319953 on Build GRCh38
rs757240104 Chr.16: 50320256-50320256 on Build GRCh38
rs757663481 Chr.16: 50320258-50320258 on Build GRCh38
rs750355136 Chr.16: 50319922-50319922 on Build GRCh38
rs759525571 Chr.16: 50320304-50320304 on Build GRCh38
rs138232073 Chr.16: 50319963-50319963 on Build GRCh38
rs373480103 Chr.16: 50320093-50320093 on Build GRCh38
rs1021999078 Chr.16: 50320163-50320163 on Build GRCh38
rs746419735 Chr.16: 50320274-50320274 on Build GRCh38
rs946948332 Chr.16: 50320195-50320196 on Build GRCh38
rs776525764 Chr.16: 50320302-50320302 on Build GRCh38
rs765295228 Chr.16: 50320307-50320307 on Build GRCh38
rs142511707 Chr.16: 50320318-50320318 on Build GRCh38
rs1030687417 Chr.16: 50320105-50320105 on Build GRCh38
rs398058226 Chr.16: 50320132-50320135 on Build GRCh38
rs765809918 Chr.16: 50319920-50319921 on Build GRCh38
rs375364426 Chr.16: 50320216-50320216 on Build GRCh38
rs758808958 Chr.16: 50320222-50320224 on Build GRCh38
rs79396113 Chr.16: 50320132-50320132 on Build GRCh38
rs148031885 Chr.16: 50320294-50320294 on Build GRCh38
rs767258863 Chr.16: 50319921-50319921 on Build GRCh38
rs370908080 Chr.16: 50319883-50319883 on Build GRCh38
rs772454259 Chr.16: 50319881-50319881 on Build GRCh38
rs751814174 Chr.16: 50320254-50320254 on Build GRCh38
rs760005909 Chr.16: 50319989-50319989 on Build GRCh38
rs771275978 Chr.16: 50319980-50319980 on Build GRCh38
rs760903415 Chr.16: 50320043-50320043 on Build GRCh38
rs199533431 Chr.16: 50320054-50320054 on Build GRCh38
rs115397158 Chr.16: 50319920-50319920 on Build GRCh38
rs766479762 Chr.16: 50319942-50319942 on Build GRCh38
rs149583258 Chr.16: 50319975-50319975 on Build GRCh38
rs756569822 Chr.16: 50320146-50320146 on Build GRCh38
rs375265731 Chr.16: 50319971-50319971 on Build GRCh38
rs778310174 Chr.16: 50319882-50319882 on Build GRCh38
rs765348237 Chr.16: 50320222-50320222 on Build GRCh38
rs755068621 Chr.16: 50319964-50319964 on Build GRCh38
rs184420261 Chr.16: 50320192-50320192 on Build GRCh38
rs748460528 Chr.16: 50319870-50319870 on Build GRCh38
rs747546538 Chr.16: 50319977-50319977 on Build GRCh38
rs1007854979 Chr.16: 50320311-50320311 on Build GRCh38
rs58387033 Chr.16: 50320183-50320183 on Build GRCh38
rs772936465 Chr.16: 50320295-50320295 on Build GRCh38
rs762558020 Chr.16: 50319909-50319909 on Build GRCh38
rs888693831 Chr.16: 50320021-50320021 on Build GRCh38
rs747854393 Chr.16: 50320034-50320034 on Build GRCh38
rs767793622 Chr.16: 50320262-50320262 on Build GRCh38
rs764381456 Chr.16: 50320253-50320253 on Build GRCh38
rs758738544 Chr.16: 50319972-50319972 on Build GRCh38
rs781513186 Chr.16: 50320272-50320272 on Build GRCh38
rs199608590 Chr.16: 50319933-50319933 on Build GRCh38
rs771891447 Chr.16: 50320035-50320035 on Build GRCh38
rs748760115 Chr.16: 50319990-50319990 on Build GRCh38
rs55820113 Chr.16: 50320129-50320132 on Build GRCh38
rs758685444 Chr.16: 50320235-50320235 on Build GRCh38
rs377516789 Chr.16: 50320132-50320133 on Build GRCh38
rs746738131 Chr.16: 50320299-50320299 on Build GRCh38
rs969469631 Chr.16: 50320169-50320169 on Build GRCh38
rs115302634 Chr.16: 50319994-50319994 on Build GRCh38
rs779118439 Chr.16: 50319868-50319868 on Build GRCh38
rs768366315 Chr.16: 50319914-50319914 on Build GRCh38
rs200438702 Chr.16: 50319916-50319916 on Build GRCh38
rs989541571 Chr.16: 50320137-50320137 on Build GRCh38
rs766174194 Chr.16: 50320199-50320199 on Build GRCh38
rs778627527 Chr.16: 50320285-50320285 on Build GRCh38
rs776578792 Chr.16: 50320215-50320215 on Build GRCh38
rs747802850 Chr.16: 50320293-50320293 on Build GRCh38
rs115491038 Chr.16: 50320312-50320312 on Build GRCh38
rs773999493 Chr.16: 50320041-50320044 on Build GRCh38
rs373486476 Chr.16: 50320279-50320279 on Build GRCh38
rs753048587 Chr.16: 50320065-50320065 on Build GRCh38
rs774928698 Chr.16: 50319893-50319893 on Build GRCh38
rs1007056217 Chr.16: 50320045-50320045 on Build GRCh38
rs998327952 Chr.16: 50320102-50320102 on Build GRCh38
rs956509845 Chr.16: 50320106-50320106 on Build GRCh38
rs1056991255 Chr.16: 50320290-50320290 on Build GRCh38
rs545410320 Chr.16: 50320144-50320144 on Build GRCh38
rs1048425459 Chr.16: 50320017-50320017 on Build GRCh38
rs769333393 Chr.16: 50319891-50319891 on Build GRCh38
rs560089245 Chr.16: 50320134-50320136 on Build GRCh38

View Full Product Details