Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 50320554-50321088 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRD7
Entrez Gene ID
29117
Gene Name
bromodomain containing 7
Gene Aliases
BP75, CELTIX1, NAG4
UniGene
Hs.437894
Gene Chromosome Location
Chr.16: 50317563-50368969 on Build GRCh38
Transcripts
NM_001173984.2 NM_013263.4 XM_011523046.2 XM_011523047.2 XM_011523048.2 XM_011523049.2 XM_011523050.2 XM_017023179.1 XM_017023180.1 XM_017023181.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4061079 c.1526G>A p.R509H Chr.16: 50320749-50320749 on Build GRCh38
6612742 c.1525C>T p.R509C Chr.16: 50320750-50320750 on Build GRCh38
4666120 c.1531G>T p.D511Y Chr.16: 50320744-50320744 on Build GRCh38
4061078 c.1563G>T p.L521L Chr.16: 50320712-50320712 on Build GRCh38
5363657 c.1539T>C p.S513S Chr.16: 50320736-50320736 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs960647064 Chr.16: 50320598-50320598 on Build GRCh38
rs766963212 Chr.16: 50320782-50320782 on Build GRCh38
rs753720662 Chr.16: 50320797-50320797 on Build GRCh38
rs773594295 Chr.16: 50320762-50320762 on Build GRCh38
rs574972855 Chr.16: 50320707-50320707 on Build GRCh38
rs1011196072 Chr.16: 50321020-50321020 on Build GRCh38
rs755693660 Chr.16: 50320793-50320793 on Build GRCh38
rs115288040 Chr.16: 50320668-50320668 on Build GRCh38
rs1049214142 Chr.16: 50320911-50320911 on Build GRCh38
rs143187051 Chr.16: 50320749-50320749 on Build GRCh38
rs773541186 Chr.16: 50320680-50320680 on Build GRCh38
rs756971066 Chr.16: 50320613-50320613 on Build GRCh38
rs139462413 Chr.16: 50320959-50320959 on Build GRCh38
rs750088076 Chr.16: 50320791-50320791 on Build GRCh38
rs980459076 Chr.16: 50320587-50320587 on Build GRCh38
rs754317764 Chr.16: 50320699-50320699 on Build GRCh38
rs546188056 Chr.16: 50320807-50320807 on Build GRCh38
rs201216846 Chr.16: 50320765-50320765 on Build GRCh38
rs376609159 Chr.16: 50320732-50320732 on Build GRCh38
rs772243482 Chr.16: 50320759-50320759 on Build GRCh38
rs759806638 Chr.16: 50320771-50320771 on Build GRCh38
rs749198621 Chr.16: 50320645-50320645 on Build GRCh38
rs538647843 Chr.16: 50320585-50320585 on Build GRCh38
rs372678031 Chr.16: 50320808-50320808 on Build GRCh38
rs868312458 Chr.16: 50320875-50320875 on Build GRCh38
rs751140715 Chr.16: 50320715-50320715 on Build GRCh38
rs765822070 Chr.16: 50320772-50320772 on Build GRCh38
rs779839531 Chr.16: 50320747-50320747 on Build GRCh38
rs189547709 Chr.16: 50320967-50320967 on Build GRCh38
rs867860491 Chr.16: 50320644-50320644 on Build GRCh38
rs980945831 Chr.16: 50320753-50320753 on Build GRCh38
rs930479821 Chr.16: 50320622-50320622 on Build GRCh38
rs745711983 Chr.16: 50320621-50320621 on Build GRCh38
rs754702314 Chr.16: 50320800-50320800 on Build GRCh38
rs765658994 Chr.16: 50320714-50320714 on Build GRCh38
rs185810190 Chr.16: 50320780-50320780 on Build GRCh38
rs971888887 Chr.16: 50320603-50320603 on Build GRCh38
rs555245080 Chr.16: 50320690-50320690 on Build GRCh38
rs761156191 Chr.16: 50320681-50320681 on Build GRCh38
rs111591872 Chr.16: 50320601-50320601 on Build GRCh38
rs199917789 Chr.16: 50320667-50320667 on Build GRCh38
rs1031308641 Chr.16: 50321015-50321015 on Build GRCh38
rs751216272 Chr.16: 50320673-50320675 on Build GRCh38
rs1034723811 Chr.16: 50320952-50320952 on Build GRCh38
rs929282894 Chr.16: 50320821-50320821 on Build GRCh38
rs572631943 Chr.16: 50320865-50320865 on Build GRCh38
rs1021354702 Chr.16: 50321024-50321024 on Build GRCh38
rs1038834699 Chr.16: 50320953-50320953 on Build GRCh38
rs775828554 Chr.16: 50320776-50320776 on Build GRCh38
rs550351931 Chr.16: 50321008-50321008 on Build GRCh38
rs35855329 Chr.16: 50320685-50320686 on Build GRCh38
rs558948948 Chr.16: 50320605-50320605 on Build GRCh38
rs778698359 Chr.16: 50320750-50320750 on Build GRCh38
rs778945743 Chr.16: 50320804-50320804 on Build GRCh38
rs181024331 Chr.16: 50320751-50320751 on Build GRCh38
rs755924791 Chr.16: 50320737-50320737 on Build GRCh38
rs769545756 Chr.16: 50320624-50320624 on Build GRCh38
rs780431590 Chr.16: 50320944-50320944 on Build GRCh38
rs765940451 Chr.16: 50320795-50320795 on Build GRCh38
rs772477059 Chr.16: 50320679-50320679 on Build GRCh38
rs373246781 Chr.16: 50320654-50320654 on Build GRCh38
rs1034922627 Chr.16: 50320590-50320590 on Build GRCh38
rs1001862690 Chr.16: 50320915-50320915 on Build GRCh38
rs1026661685 Chr.16: 50321045-50321045 on Build GRCh38
rs745981389 Chr.16: 50320691-50320691 on Build GRCh38
rs756738060 Chr.16: 50320720-50320720 on Build GRCh38
rs747173962 Chr.16: 50320763-50320763 on Build GRCh38
rs780863150 Chr.16: 50320721-50320721 on Build GRCh38
rs777122700 Chr.16: 50320769-50320769 on Build GRCh38
rs997406172 Chr.16: 50321002-50321002 on Build GRCh38
rs112415991 Chr.16: 50320616-50320616 on Build GRCh38
rs779996220 Chr.16: 50320637-50320637 on Build GRCh38

View Full Product Details