Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 53156692-53157208 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHD9
Entrez Gene ID
80205
Gene Name
chromodomain helicase DNA binding protein 9
Gene Aliases
AD013, CHD-9, CReMM, KISH2, PRIC320
UniGene
Hs.59159
Gene Chromosome Location
Chr.16: 53054991-53327502 on Build GRCh38
Transcripts
NM_001308319.1 NM_025134.4 XM_005256168.3 XM_005256169.2 XM_005256172.3 XM_006721281.2 XM_006721283.3 XM_011523347.2 XM_017023718.1 XM_017023719.1 XM_017023720.1 XM_017023721.1 XM_017023722.1 XM_017023723.1 XM_017023724.1 XM_017023725.1 XM_017023728.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3957643 c.839A>G p.H280R Chr.16: 53156928-53156928 on Build GRCh38
1201022 c.1115G>A p.G372D Chr.16: 53157204-53157204 on Build GRCh38
971409 c.781T>C p.S261P Chr.16: 53156870-53156870 on Build GRCh38
5820305 c.959A>T p.E320V Chr.16: 53157048-53157048 on Build GRCh38
971410 c.1080C>A p.N360K Chr.16: 53157169-53157169 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768904662 Chr.16: 53157126-53157126 on Build GRCh38
rs767088858 Chr.16: 53156839-53156839 on Build GRCh38
rs764837487 Chr.16: 53157082-53157082 on Build GRCh38
rs755727985 Chr.16: 53156745-53156745 on Build GRCh38
rs570493698 Chr.16: 53156840-53156840 on Build GRCh38
rs764545311 Chr.16: 53156961-53156961 on Build GRCh38
rs766084964 Chr.16: 53156865-53156865 on Build GRCh38
rs780512837 Chr.16: 53156743-53156743 on Build GRCh38
rs746127217 Chr.16: 53157036-53157036 on Build GRCh38
rs368501690 Chr.16: 53157055-53157055 on Build GRCh38
rs374223041 Chr.16: 53156791-53156791 on Build GRCh38
rs765888157 Chr.16: 53156729-53156729 on Build GRCh38
rs372368284 Chr.16: 53157054-53157054 on Build GRCh38
rs770457802 Chr.16: 53157070-53157070 on Build GRCh38
rs751550507 Chr.16: 53157095-53157095 on Build GRCh38
rs759957818 Chr.16: 53156716-53156716 on Build GRCh38
rs753472211 Chr.16: 53156868-53156868 on Build GRCh38
rs763306566 Chr.16: 53156833-53156833 on Build GRCh38
rs778877345 Chr.16: 53156761-53156761 on Build GRCh38
rs758182260 Chr.16: 53156984-53156984 on Build GRCh38
rs890921265 Chr.16: 53157023-53157023 on Build GRCh38
rs200005447 Chr.16: 53157080-53157080 on Build GRCh38
rs371392783 Chr.16: 53156755-53156755 on Build GRCh38
rs755585262 Chr.16: 53156853-53156853 on Build GRCh38
rs753141572 Chr.16: 53156731-53156731 on Build GRCh38
rs952153760 Chr.16: 53157122-53157122 on Build GRCh38
rs765686560 Chr.16: 53156815-53156815 on Build GRCh38
rs760045707 Chr.16: 53156814-53156814 on Build GRCh38
rs772615275 Chr.16: 53157153-53157153 on Build GRCh38
rs908125044 Chr.16: 53157171-53157171 on Build GRCh38
rs748675330 Chr.16: 53157138-53157138 on Build GRCh38
rs1026250197 Chr.16: 53157144-53157144 on Build GRCh38
rs770289729 Chr.16: 53156946-53156946 on Build GRCh38
rs776964626 Chr.16: 53156803-53156803 on Build GRCh38
rs138285755 Chr.16: 53156727-53156727 on Build GRCh38
rs748147627 Chr.16: 53156890-53156890 on Build GRCh38
rs746319819 Chr.16: 53156962-53156962 on Build GRCh38
rs368802058 Chr.16: 53156736-53156736 on Build GRCh38
rs376197651 Chr.16: 53157083-53157083 on Build GRCh38
rs762238500 Chr.16: 53157069-53157069 on Build GRCh38
rs370484239 Chr.16: 53156818-53156818 on Build GRCh38
rs777638518 Chr.16: 53157018-53157018 on Build GRCh38
rs746032556 Chr.16: 53156938-53156938 on Build GRCh38
rs759035782 Chr.16: 53157172-53157172 on Build GRCh38
rs753490752 Chr.16: 53156970-53156970 on Build GRCh38
rs750099482 Chr.16: 53156744-53156744 on Build GRCh38
rs754779388 Chr.16: 53156869-53156869 on Build GRCh38
rs760232739 Chr.16: 53156966-53156966 on Build GRCh38
rs1049265423 Chr.16: 53156894-53156894 on Build GRCh38
rs749117823 Chr.16: 53156751-53156751 on Build GRCh38
rs910328222 Chr.16: 53156901-53156901 on Build GRCh38
rs762601868 Chr.16: 53157180-53157180 on Build GRCh38
rs374801460 Chr.16: 53156960-53156960 on Build GRCh38
rs771328225 Chr.16: 53156801-53156801 on Build GRCh38
rs571704208 Chr.16: 53156850-53156850 on Build GRCh38
rs765814836 Chr.16: 53156969-53156969 on Build GRCh38
rs773677621 Chr.16: 53157164-53157164 on Build GRCh38
rs752483362 Chr.16: 53156981-53156981 on Build GRCh38
rs777052737 Chr.16: 53156930-53156930 on Build GRCh38
rs747976696 Chr.16: 53156771-53156771 on Build GRCh38
rs61759475 Chr.16: 53157043-53157043 on Build GRCh38
rs776070328 Chr.16: 53157075-53157075 on Build GRCh38
rs781448144 Chr.16: 53157109-53157109 on Build GRCh38
rs930341238 Chr.16: 53156893-53156893 on Build GRCh38
rs377416139 Chr.16: 53156795-53156795 on Build GRCh38
rs868708448 Chr.16: 53156813-53156813 on Build GRCh38
rs757223079 Chr.16: 53157107-53157107 on Build GRCh38
rs367557277 Chr.16: 53156972-53156972 on Build GRCh38
rs763893803 Chr.16: 53157093-53157093 on Build GRCh38
rs535501040 Chr.16: 53157123-53157123 on Build GRCh38
rs371390517 Chr.16: 53157031-53157031 on Build GRCh38
rs775681341 Chr.16: 53156947-53156947 on Build GRCh38
rs370289002 Chr.16: 53157129-53157129 on Build GRCh38
rs777745047 Chr.16: 53156902-53156902 on Build GRCh38
rs747099721 Chr.16: 53156914-53156914 on Build GRCh38
rs143930746 Chr.16: 53157040-53157040 on Build GRCh38
rs772196014 Chr.16: 53156790-53156790 on Build GRCh38
rs374355186 Chr.16: 53156746-53156746 on Build GRCh38
rs746932672 Chr.16: 53157027-53157027 on Build GRCh38
rs559672647 Chr.16: 53156807-53156807 on Build GRCh38
rs996239832 Chr.16: 53157060-53157060 on Build GRCh38
rs771131924 Chr.16: 53156919-53156919 on Build GRCh38
rs765101939 Chr.16: 53156980-53156980 on Build GRCh38
rs779207024 Chr.16: 53157136-53157136 on Build GRCh38
rs750583516 Chr.16: 53157114-53157114 on Build GRCh38
rs756210035 Chr.16: 53157118-53157118 on Build GRCh38
rs769049307 Chr.16: 53157048-53157048 on Build GRCh38
rs201124945 Chr.16: 53157035-53157035 on Build GRCh38
rs769335335 Chr.16: 53157179-53157179 on Build GRCh38
rs778503865 Chr.16: 53156874-53156874 on Build GRCh38
rs762716450 Chr.16: 53157092-53157092 on Build GRCh38
rs758218882 Chr.16: 53156898-53156898 on Build GRCh38
rs754782615 Chr.16: 53156810-53156810 on Build GRCh38

View Full Product Details