Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 53226174-53226695 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHD9
Entrez Gene ID
80205
Gene Name
chromodomain helicase DNA binding protein 9
Gene Aliases
AD013, CHD-9, CReMM, KISH2, PRIC320
UniGene
Hs.59159
Gene Chromosome Location
Chr.16: 53054991-53327502 on Build GRCh38
Transcripts
NM_001308319.1 NM_025134.4 XM_005256168.3 XM_005256169.2 XM_005256172.3 XM_005256176.3 XM_006721281.2 XM_006721283.3 XM_011523347.2 XM_017023718.1 XM_017023719.1 XM_017023720.1 XM_017023721.1 XM_017023722.1 XM_017023723.1 XM_017023724.1 XM_017023725.1 XM_017023726.1 XM_017023727.1 XM_017023728.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5851037 c.2015C>T p.P672L Chr.16: 53226484-53226484 on Build GRCh38
5783871 c.1934A>G p.Y645C Chr.16: 53226403-53226403 on Build GRCh38
299130 c.1954A>C p.K652Q Chr.16: 53226423-53226423 on Build GRCh38
6144849 c.1947A>G p.I649M Chr.16: 53226416-53226416 on Build GRCh38
260098 c.1982C>A p.S661Y Chr.16: 53226451-53226451 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761789559 Chr.16: 53226538-53226538 on Build GRCh38
rs769204023 Chr.16: 53226388-53226388 on Build GRCh38
rs567864367 Chr.16: 53226242-53226242 on Build GRCh38
rs984411688 Chr.16: 53226581-53226581 on Build GRCh38
rs750629025 Chr.16: 53226430-53226430 on Build GRCh38
rs773431903 Chr.16: 53226467-53226467 on Build GRCh38
rs763912634 Chr.16: 53226533-53226533 on Build GRCh38
rs370028859 Chr.16: 53226363-53226363 on Build GRCh38
rs762818172 Chr.16: 53226417-53226417 on Build GRCh38
rs540245665 Chr.16: 53226503-53226503 on Build GRCh38
rs758369655 Chr.16: 53226337-53226337 on Build GRCh38
rs769534090 Chr.16: 53226510-53226510 on Build GRCh38
rs919558752 Chr.16: 53226618-53226618 on Build GRCh38
rs751735383 Chr.16: 53226349-53226349 on Build GRCh38
rs1038475212 Chr.16: 53226238-53226238 on Build GRCh38
rs757414572 Chr.16: 53226426-53226426 on Build GRCh38
rs375075278 Chr.16: 53226404-53226404 on Build GRCh38
rs1046923262 Chr.16: 53226670-53226670 on Build GRCh38
rs888072205 Chr.16: 53226290-53226290 on Build GRCh38
rs749712411 Chr.16: 53226444-53226444 on Build GRCh38
rs752599793 Chr.16: 53226326-53226326 on Build GRCh38
rs760246838 Chr.16: 53226400-53226400 on Build GRCh38
rs924192949 Chr.16: 53226484-53226484 on Build GRCh38
rs900932035 Chr.16: 53226643-53226643 on Build GRCh38
rs929866241 Chr.16: 53226260-53226260 on Build GRCh38
rs746747482 Chr.16: 53226408-53226408 on Build GRCh38
rs185043204 Chr.16: 53226415-53226415 on Build GRCh38
rs748597833 Chr.16: 53226461-53226461 on Build GRCh38
rs182254882 Chr.16: 53226405-53226405 on Build GRCh38
rs199637615 Chr.16: 53226540-53226540 on Build GRCh38
rs756402358 Chr.16: 53226439-53226439 on Build GRCh38
rs779399078 Chr.16: 53226454-53226454 on Build GRCh38
rs761372435 Chr.16: 53226368-53226370 on Build GRCh38
rs539341095 Chr.16: 53226389-53226389 on Build GRCh38
rs373004025 Chr.16: 53226537-53226537 on Build GRCh38
rs577033390 Chr.16: 53226617-53226617 on Build GRCh38
rs751536236 Chr.16: 53226424-53226424 on Build GRCh38
rs765024417 Chr.16: 53226321-53226321 on Build GRCh38
rs938554916 Chr.16: 53226216-53226216 on Build GRCh38
rs372425397 Chr.16: 53226382-53226382 on Build GRCh38
rs567032914 Chr.16: 53226358-53226358 on Build GRCh38
rs929650158 Chr.16: 53226635-53226635 on Build GRCh38
rs757397985 Chr.16: 53226352-53226352 on Build GRCh38
rs1050794502 Chr.16: 53226224-53226224 on Build GRCh38
rs776351463 Chr.16: 53226346-53226347 on Build GRCh38
rs909797851 Chr.16: 53226590-53226590 on Build GRCh38
rs942373953 Chr.16: 53226237-53226237 on Build GRCh38
rs746033356 Chr.16: 53226360-53226360 on Build GRCh38
rs190551773 Chr.16: 53226267-53226267 on Build GRCh38
rs762907533 Chr.16: 53226529-53226529 on Build GRCh38
rs771317689 Chr.16: 53226577-53226577 on Build GRCh38
rs750568692 Chr.16: 53226556-53226556 on Build GRCh38
rs571012040 Chr.16: 53226341-53226341 on Build GRCh38
rs200018633 Chr.16: 53226453-53226453 on Build GRCh38
rs200382720 Chr.16: 53226428-53226428 on Build GRCh38
rs12599784 Chr.16: 53226247-53226247 on Build GRCh38
rs775266690 Chr.16: 53226526-53226526 on Build GRCh38
rs764184534 Chr.16: 53226421-53226421 on Build GRCh38
rs369540834 Chr.16: 53226412-53226412 on Build GRCh38
rs889573617 Chr.16: 53226231-53226231 on Build GRCh38
rs555097665 Chr.16: 53226440-53226440 on Build GRCh38
rs780413159 Chr.16: 53226381-53226381 on Build GRCh38
rs776167248 Chr.16: 53226496-53226496 on Build GRCh38
rs1002663444 Chr.16: 53226336-53226336 on Build GRCh38
rs942470223 Chr.16: 53226561-53226561 on Build GRCh38
rs777821340 Chr.16: 53226348-53226348 on Build GRCh38
rs1025531509 Chr.16: 53226280-53226280 on Build GRCh38

View Full Product Details