Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 56193604-56194112 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DKFZP434H168
Entrez Gene ID
26077
Gene Name
uncharacterized LOC26077
Gene Aliases
-
UniGene
Hs.721567
Gene Chromosome Location
Chr.16: 56192617-56194525 on Build GRCh38
Transcripts
NR_026889.1 NM_020988.2 NM_138736.2 XM_011523003.2

Gene Symbol
GNAO1
Entrez Gene ID
2775
Gene Name
G protein subunit alpha o1
Gene Aliases
EIEE17, G-ALPHA-o, GNAO, HLA-DQB1
UniGene
Hs.644524
Gene Chromosome Location
Chr.16: 56191339-56357444 on Build GRCh38
Transcripts
NR_026889.1 NM_020988.2 NM_138736.2 XM_011523003.2

SNP Detail

SNP ID Chromosome Location
rs569520872 Chr.16: 56193830-56193830 on Build GRCh38
rs1032595817 Chr.16: 56194016-56194016 on Build GRCh38
rs1006263497 Chr.16: 56193838-56193838 on Build GRCh38
rs976878772 Chr.16: 56194079-56194079 on Build GRCh38
rs1014065839 Chr.16: 56193921-56193921 on Build GRCh38
rs1025156040 Chr.16: 56193929-56193929 on Build GRCh38
rs781207601 Chr.16: 56194066-56194066 on Build GRCh38
rs529371734 Chr.16: 56193765-56193765 on Build GRCh38
rs987764234 Chr.16: 56193656-56193656 on Build GRCh38
rs969991143 Chr.16: 56193979-56193979 on Build GRCh38
rs940305351 Chr.16: 56193718-56193718 on Build GRCh38
rs577826125 Chr.16: 56193674-56193674 on Build GRCh38
rs1040885990 Chr.16: 56193697-56193697 on Build GRCh38
rs191699168 Chr.16: 56193858-56193858 on Build GRCh38
rs1037790354 Chr.16: 56193729-56193729 on Build GRCh38
rs898894661 Chr.16: 56193735-56193735 on Build GRCh38
rs1048369539 Chr.16: 56193768-56193768 on Build GRCh38
rs778894476 Chr.16: 56194047-56194047 on Build GRCh38
rs778116156 Chr.16: 56193993-56193993 on Build GRCh38
rs932598430 Chr.16: 56193759-56193759 on Build GRCh38
rs76026714 Chr.16: 56193997-56193997 on Build GRCh38
rs943750796 Chr.16: 56193686-56193686 on Build GRCh38
rs756325763 Chr.16: 56193906-56193906 on Build GRCh38
rs59779556 Chr.16: 56194053-56194053 on Build GRCh38
rs540249768 Chr.16: 56193663-56193663 on Build GRCh38
rs925525827 Chr.16: 56193640-56193640 on Build GRCh38
rs538914650 Chr.16: 56193826-56193826 on Build GRCh38
rs943819785 Chr.16: 56194063-56194063 on Build GRCh38
rs936365542 Chr.16: 56193641-56193641 on Build GRCh38
rs528528950 Chr.16: 56193642-56193642 on Build GRCh38
rs913530277 Chr.16: 56194062-56194062 on Build GRCh38
rs937115166 Chr.16: 56193753-56193753 on Build GRCh38
rs981403782 Chr.16: 56193633-56193633 on Build GRCh38
rs113938077 Chr.16: 56194021-56194021 on Build GRCh38

View Full Product Details