Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 56969676-56970202 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CETP
Entrez Gene ID
1071
Gene Name
cholesteryl ester transfer protein
Gene Aliases
BPIFF, HDLCQ10
UniGene
Hs.89538
Gene Chromosome Location
Chr.16: 56961923-56983844 on Build GRCh38
Transcripts
NM_000078.2 NM_001286085.1 XM_006721124.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2836116 c.523C>T p.R175* Chr.16: 56969997-56969997 on Build GRCh38
6627451 c.473C>G p.P158R Chr.16: 56969947-56969947 on Build GRCh38
5576531 c.492C>T p.F164F Chr.16: 56969966-56969966 on Build GRCh38
3818136 c.461G>A p.R154Q Chr.16: 56969935-56969935 on Build GRCh38
3818135 c.438G>C p.L146L Chr.16: 56969680-56969680 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs907604241 Chr.16: 56969744-56969744 on Build GRCh38
rs145690607 Chr.16: 56969998-56969998 on Build GRCh38
rs761326185 Chr.16: 56969941-56969941 on Build GRCh38
rs747572365 Chr.16: 56970011-56970011 on Build GRCh38
rs761700239 Chr.16: 56969899-56969899 on Build GRCh38
rs368753277 Chr.16: 56969960-56969960 on Build GRCh38
rs570472066 Chr.16: 56969802-56969802 on Build GRCh38
rs756262184 Chr.16: 56969722-56969722 on Build GRCh38
rs201660416 Chr.16: 56969900-56969900 on Build GRCh38
rs34716057 Chr.16: 56969934-56969934 on Build GRCh38
rs184615182 Chr.16: 56969935-56969935 on Build GRCh38
rs755245196 Chr.16: 56970037-56970037 on Build GRCh38
rs751494873 Chr.16: 56969707-56969707 on Build GRCh38
rs763564306 Chr.16: 56969895-56969895 on Build GRCh38
rs965914204 Chr.16: 56969704-56969704 on Build GRCh38
rs755969723 Chr.16: 56969907-56969907 on Build GRCh38
rs369184538 Chr.16: 56970039-56970039 on Build GRCh38
rs1018757045 Chr.16: 56969914-56969914 on Build GRCh38
rs753695890 Chr.16: 56969978-56969978 on Build GRCh38
rs758622494 Chr.16: 56970001-56970001 on Build GRCh38
rs369737295 Chr.16: 56969877-56969877 on Build GRCh38
rs781445411 Chr.16: 56969948-56969948 on Build GRCh38
rs570369308 Chr.16: 56969701-56969701 on Build GRCh38
rs781374059 Chr.16: 56970028-56970028 on Build GRCh38
rs553619710 Chr.16: 56969876-56969876 on Build GRCh38
rs891141 Chr.16: 56969811-56969811 on Build GRCh38
rs913317710 Chr.16: 56970108-56970108 on Build GRCh38
rs367562884 Chr.16: 56969708-56969708 on Build GRCh38
rs765506619 Chr.16: 56969996-56969996 on Build GRCh38
rs762432832 Chr.16: 56969696-56969696 on Build GRCh38
rs936016057 Chr.16: 56970090-56970090 on Build GRCh38
rs60896367 Chr.16: 56969726-56969726 on Build GRCh38
rs901701572 Chr.16: 56970150-56970150 on Build GRCh38
rs747376563 Chr.16: 56969931-56969931 on Build GRCh38
rs552291657 Chr.16: 56969761-56969761 on Build GRCh38
rs368552601 Chr.16: 56969869-56969869 on Build GRCh38
rs769984356 Chr.16: 56970052-56970053 on Build GRCh38
rs984971450 Chr.16: 56969736-56969736 on Build GRCh38
rs201778606 Chr.16: 56969940-56969940 on Build GRCh38
rs570087370 Chr.16: 56969734-56969734 on Build GRCh38
rs754054236 Chr.16: 56969911-56969911 on Build GRCh38
rs768657302 Chr.16: 56969731-56969731 on Build GRCh38
rs768891652 Chr.16: 56969936-56969939 on Build GRCh38
rs1040480633 Chr.16: 56970144-56970144 on Build GRCh38
rs554738455 Chr.16: 56969925-56969925 on Build GRCh38
rs772644949 Chr.16: 56969924-56969924 on Build GRCh38
rs563857383 Chr.16: 56970115-56970115 on Build GRCh38
rs946149173 Chr.16: 56970141-56970141 on Build GRCh38
rs757808322 Chr.16: 56970023-56970023 on Build GRCh38
rs751104316 Chr.16: 56969896-56969896 on Build GRCh38
rs878917072 Chr.16: 56969825-56969825 on Build GRCh38
rs150001454 Chr.16: 56969714-56969714 on Build GRCh38
rs775075583 Chr.16: 56969886-56969886 on Build GRCh38
rs891142 Chr.16: 56970065-56970065 on Build GRCh38
rs995998395 Chr.16: 56970153-56970153 on Build GRCh38
rs762497320 Chr.16: 56969894-56969894 on Build GRCh38
rs764465902 Chr.16: 56969897-56969897 on Build GRCh38
rs755085520 Chr.16: 56969985-56969985 on Build GRCh38
rs887678470 Chr.16: 56970177-56970177 on Build GRCh38
rs780918298 Chr.16: 56969709-56969709 on Build GRCh38
rs371368993 Chr.16: 56969774-56969774 on Build GRCh38
rs768136410 Chr.16: 56970036-56970036 on Build GRCh38
rs372251011 Chr.16: 56969906-56969906 on Build GRCh38
rs780290443 Chr.16: 56969909-56969909 on Build GRCh38
rs748342397 Chr.16: 56969954-56969957 on Build GRCh38
rs972835736 Chr.16: 56969764-56969764 on Build GRCh38
rs769561732 Chr.16: 56969882-56969882 on Build GRCh38
rs905881672 Chr.16: 56969831-56969831 on Build GRCh38
rs773174024 Chr.16: 56969955-56969955 on Build GRCh38
rs891143 Chr.16: 56970068-56970068 on Build GRCh38
rs778396826 Chr.16: 56970005-56970005 on Build GRCh38
rs766037201 Chr.16: 56969967-56969967 on Build GRCh38
rs991861534 Chr.16: 56969997-56969997 on Build GRCh38
rs1050347230 Chr.16: 56970157-56970157 on Build GRCh38
rs548545514 Chr.16: 56970095-56970095 on Build GRCh38
rs779450905 Chr.16: 56969864-56969864 on Build GRCh38
rs200767844 Chr.16: 56969993-56969993 on Build GRCh38
rs748614904 Chr.16: 56969865-56969865 on Build GRCh38
rs34847858 Chr.16: 56969816-56969816 on Build GRCh38
rs746250474 Chr.16: 56970030-56970030 on Build GRCh38
rs564885502 Chr.16: 56969939-56969939 on Build GRCh38
rs778814831 Chr.16: 56969917-56969917 on Build GRCh38
rs767437395 Chr.16: 56969946-56969946 on Build GRCh38
rs35752722 Chr.16: 56969749-56969749 on Build GRCh38

View Full Product Details