Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 57123323-57123724 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPNE2
Entrez Gene ID
221184
Gene Name
copine 2
Gene Aliases
COPN2, CPN2
UniGene
Hs.339809
Gene Chromosome Location
Chr.16: 57092543-57147966 on Build GRCh38
Transcripts
NM_152727.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1302050 c.881A>G p.Y294C Chr.16: 57123427-57123427 on Build GRCh38
5889304 c.884C>T p.S295F Chr.16: 57123430-57123430 on Build GRCh38
109123 c.905G>A p.G302E Chr.16: 57123451-57123451 on Build GRCh38
6634633 c.880T>C p.Y294H Chr.16: 57123426-57123426 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1008598279 Chr.16: 57123444-57123444 on Build GRCh38
rs765992722 Chr.16: 57123415-57123415 on Build GRCh38
rs182357941 Chr.16: 57123452-57123452 on Build GRCh38
rs748374486 Chr.16: 57123503-57123504 on Build GRCh38
rs765023981 Chr.16: 57123610-57123610 on Build GRCh38
rs111954210 Chr.16: 57123609-57123609 on Build GRCh38
rs529278663 Chr.16: 57123536-57123536 on Build GRCh38
rs775685097 Chr.16: 57123487-57123487 on Build GRCh38
rs746322572 Chr.16: 57123483-57123483 on Build GRCh38
rs536922622 Chr.16: 57123658-57123658 on Build GRCh38
rs774691813 Chr.16: 57123359-57123359 on Build GRCh38
rs142976078 Chr.16: 57123473-57123473 on Build GRCh38
rs759517860 Chr.16: 57123521-57123521 on Build GRCh38
rs1057081743 Chr.16: 57123578-57123578 on Build GRCh38
rs760290247 Chr.16: 57123408-57123408 on Build GRCh38
rs373916689 Chr.16: 57123420-57123420 on Build GRCh38
rs775851863 Chr.16: 57123391-57123391 on Build GRCh38
rs150440330 Chr.16: 57123488-57123488 on Build GRCh38
rs747051066 Chr.16: 57123464-57123464 on Build GRCh38
rs948650275 Chr.16: 57123604-57123604 on Build GRCh38
rs770926041 Chr.16: 57123466-57123466 on Build GRCh38
rs777184713 Chr.16: 57123372-57123372 on Build GRCh38
rs778811777 Chr.16: 57123425-57123425 on Build GRCh38
rs1043298602 Chr.16: 57123389-57123389 on Build GRCh38
rs141813722 Chr.16: 57123430-57123430 on Build GRCh38
rs761794678 Chr.16: 57123515-57123515 on Build GRCh38
rs138215255 Chr.16: 57123592-57123592 on Build GRCh38
rs138717944 Chr.16: 57123451-57123451 on Build GRCh38
rs954356076 Chr.16: 57123421-57123421 on Build GRCh38
rs904242081 Chr.16: 57123394-57123394 on Build GRCh38
rs140503310 Chr.16: 57123346-57123346 on Build GRCh38
rs894423462 Chr.16: 57123386-57123386 on Build GRCh38
rs778118275 Chr.16: 57123457-57123457 on Build GRCh38
rs938671303 Chr.16: 57123547-57123547 on Build GRCh38
rs374283081 Chr.16: 57123401-57123401 on Build GRCh38
rs142359304 Chr.16: 57123674-57123674 on Build GRCh38
rs370770978 Chr.16: 57123402-57123402 on Build GRCh38
rs1012683086 Chr.16: 57123388-57123388 on Build GRCh38
rs79310064 Chr.16: 57123524-57123524 on Build GRCh38
rs760258687 Chr.16: 57123504-57123504 on Build GRCh38
rs770314881 Chr.16: 57123485-57123485 on Build GRCh38
rs772855726 Chr.16: 57123496-57123496 on Build GRCh38
rs760036460 Chr.16: 57123373-57123373 on Build GRCh38
rs908658308 Chr.16: 57123533-57123533 on Build GRCh38
rs748992560 Chr.16: 57123367-57123371 on Build GRCh38
rs562163888 Chr.16: 57123529-57123529 on Build GRCh38
rs570311952 Chr.16: 57123687-57123687 on Build GRCh38
rs771609147 Chr.16: 57123490-57123490 on Build GRCh38
rs753771031 Chr.16: 57123418-57123418 on Build GRCh38
rs752580055 Chr.16: 57123426-57123426 on Build GRCh38
rs765524757 Chr.16: 57123376-57123376 on Build GRCh38
rs998734433 Chr.16: 57123631-57123631 on Build GRCh38

View Full Product Details