Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 57248353-57248853 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARL2BP
Entrez Gene ID
23568
Gene Name
ADP ribosylation factor like GTPase 2 binding protein
Gene Aliases
BART, BART1, RP66
UniGene
Hs.632873
Gene Chromosome Location
Chr.16: 57245126-57253635 on Build GRCh38
Transcripts
NM_012106.3 XM_011522977.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7356432 c.182T>C p.I61T Chr.16: 57248618-57248618 on Build GRCh38
704054 c.207C>A p.Y69* Chr.16: 57248643-57248643 on Build GRCh38
192250 c.207C>T p.Y69Y Chr.16: 57248643-57248643 on Build GRCh38
4061397 c.101-1G>A p.? Chr.16: 57248536-57248536 on Build GRCh38
5032156 c.207+1G>A p.? Chr.16: 57248644-57248644 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs35540615 Chr.16: 57248380-57248381 on Build GRCh38
rs879255568 Chr.16: 57248536-57248536 on Build GRCh38
rs764567016 Chr.16: 57248658-57248658 on Build GRCh38
rs558633783 Chr.16: 57248716-57248716 on Build GRCh38
rs75804875 Chr.16: 57248408-57248408 on Build GRCh38
rs1038814950 Chr.16: 57248399-57248399 on Build GRCh38
rs372547772 Chr.16: 57248404-57248404 on Build GRCh38
rs941301480 Chr.16: 57248398-57248398 on Build GRCh38
rs751906316 Chr.16: 57248662-57248662 on Build GRCh38
rs750949372 Chr.16: 57248517-57248517 on Build GRCh38
rs907021382 Chr.16: 57248465-57248465 on Build GRCh38
rs796103933 Chr.16: 57248695-57248696 on Build GRCh38
rs1001335538 Chr.16: 57248469-57248469 on Build GRCh38
rs762263722 Chr.16: 57248666-57248666 on Build GRCh38
rs780285650 Chr.16: 57248644-57248647 on Build GRCh38
rs760843664 Chr.16: 57248489-57248489 on Build GRCh38
rs1020405126 Chr.16: 57248670-57248670 on Build GRCh38
rs771882265 Chr.16: 57248574-57248574 on Build GRCh38
rs753124924 Chr.16: 57248521-57248521 on Build GRCh38
rs373350904 Chr.16: 57248513-57248513 on Build GRCh38
rs140789182 Chr.16: 57248592-57248592 on Build GRCh38
rs1055436259 Chr.16: 57248533-57248533 on Build GRCh38
rs955497438 Chr.16: 57248785-57248785 on Build GRCh38
rs779293413 Chr.16: 57248661-57248661 on Build GRCh38
rs776708097 Chr.16: 57248612-57248612 on Build GRCh38
rs186308074 Chr.16: 57248556-57248556 on Build GRCh38
rs147277394 Chr.16: 57248568-57248568 on Build GRCh38
rs189864695 Chr.16: 57248671-57248671 on Build GRCh38
rs779714473 Chr.16: 57248541-57248541 on Build GRCh38
rs759712636 Chr.16: 57248624-57248624 on Build GRCh38
rs771213865 Chr.16: 57248607-57248607 on Build GRCh38
rs772036880 Chr.16: 57248751-57248751 on Build GRCh38
rs398123053 Chr.16: 57248570-57248570 on Build GRCh38
rs534372884 Chr.16: 57248428-57248428 on Build GRCh38
rs199830550 Chr.16: 57248644-57248644 on Build GRCh38
rs747240176 Chr.16: 57248601-57248601 on Build GRCh38
rs779130053 Chr.16: 57248496-57248498 on Build GRCh38
rs1045550180 Chr.16: 57248436-57248436 on Build GRCh38
rs918500970 Chr.16: 57248400-57248400 on Build GRCh38
rs377362707 Chr.16: 57248534-57248534 on Build GRCh38
rs758880525 Chr.16: 57248634-57248639 on Build GRCh38
rs201489348 Chr.16: 57248643-57248643 on Build GRCh38
rs553095861 Chr.16: 57248497-57248497 on Build GRCh38
rs753029079 Chr.16: 57248574-57248575 on Build GRCh38
rs750123343 Chr.16: 57248537-57248537 on Build GRCh38
rs753452835 Chr.16: 57248542-57248542 on Build GRCh38
rs763416650 Chr.16: 57248531-57248531 on Build GRCh38
rs776630072 Chr.16: 57248500-57248500 on Build GRCh38
rs747351014 Chr.16: 57248695-57248696 on Build GRCh38
rs574634322 Chr.16: 57248517-57248517 on Build GRCh38
rs1000212508 Chr.16: 57248727-57248727 on Build GRCh38
rs1029940372 Chr.16: 57248742-57248742 on Build GRCh38
rs755770384 Chr.16: 57248538-57248538 on Build GRCh38
rs777490770 Chr.16: 57248580-57248580 on Build GRCh38

View Full Product Details