Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 57372387-57372881 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CX3CL1
Entrez Gene ID
6376
Gene Name
C-X3-C motif chemokine ligand 1
Gene Aliases
ABCD-3, C3Xkine, CXC3, CXC3C, NTN, NTT, SCYD1, fractalkine, neurotactin
UniGene
Hs.531668
Gene Chromosome Location
Chr.16: 57372461-57385048 on Build GRCh38
Transcripts
NM_001304392.1 NM_002996.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5813969 c.10A>G p.I4V Chr.16: 57372578-57372578 on Build GRCh38
1731684 c.68C>T p.A23V Chr.16: 57372636-57372636 on Build GRCh38
1302052 c.9G>A p.P3P Chr.16: 57372577-57372577 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs755823983 Chr.16: 57372690-57372690 on Build GRCh38
rs751917598 Chr.16: 57372685-57372685 on Build GRCh38
rs201608370 Chr.16: 57372607-57372607 on Build GRCh38
rs374004803 Chr.16: 57372513-57372513 on Build GRCh38
rs540846821 Chr.16: 57372855-57372855 on Build GRCh38
rs1013696679 Chr.16: 57372862-57372862 on Build GRCh38
rs777182844 Chr.16: 57372689-57372689 on Build GRCh38
rs558286943 Chr.16: 57372842-57372842 on Build GRCh38
rs372607275 Chr.16: 57372538-57372538 on Build GRCh38
rs765811669 Chr.16: 57372667-57372667 on Build GRCh38
rs76505297 Chr.16: 57372659-57372659 on Build GRCh38
rs542222411 Chr.16: 57372514-57372514 on Build GRCh38
rs375982954 Chr.16: 57372542-57372542 on Build GRCh38
rs758777234 Chr.16: 57372677-57372677 on Build GRCh38
rs758730102 Chr.16: 57372599-57372599 on Build GRCh38
rs141505173 Chr.16: 57372589-57372589 on Build GRCh38
rs772048321 Chr.16: 57372846-57372846 on Build GRCh38
rs868712649 Chr.16: 57372585-57372586 on Build GRCh38
rs777467251 Chr.16: 57372782-57372782 on Build GRCh38
rs748737416 Chr.16: 57372622-57372622 on Build GRCh38
rs1041027047 Chr.16: 57372458-57372458 on Build GRCh38
rs780735009 Chr.16: 57372532-57372532 on Build GRCh38
rs748216983 Chr.16: 57372816-57372816 on Build GRCh38
rs573435924 Chr.16: 57372845-57372845 on Build GRCh38
rs140854557 Chr.16: 57372576-57372576 on Build GRCh38
rs1015425699 Chr.16: 57372718-57372718 on Build GRCh38
rs73559216 Chr.16: 57372750-57372750 on Build GRCh38
rs1007867420 Chr.16: 57372457-57372457 on Build GRCh38
rs748595033 Chr.16: 57372597-57372597 on Build GRCh38
rs762681637 Chr.16: 57372521-57372521 on Build GRCh38
rs774128035 Chr.16: 57372628-57372628 on Build GRCh38
rs367908467 Chr.16: 57372783-57372783 on Build GRCh38
rs374258775 Chr.16: 57372629-57372629 on Build GRCh38
rs550527744 Chr.16: 57372678-57372678 on Build GRCh38
rs562214859 Chr.16: 57372744-57372744 on Build GRCh38
rs774177382 Chr.16: 57372785-57372785 on Build GRCh38
rs568793895 Chr.16: 57372725-57372725 on Build GRCh38
rs73559219 Chr.16: 57372780-57372780 on Build GRCh38
rs146188225 Chr.16: 57372600-57372600 on Build GRCh38
rs199506606 Chr.16: 57372528-57372528 on Build GRCh38
rs759512489 Chr.16: 57372662-57372662 on Build GRCh38
rs755847246 Chr.16: 57372464-57372464 on Build GRCh38
rs201377096 Chr.16: 57372553-57372553 on Build GRCh38
rs980683086 Chr.16: 57372573-57372573 on Build GRCh38
rs760908389 Chr.16: 57372644-57372644 on Build GRCh38
rs749752388 Chr.16: 57372533-57372533 on Build GRCh38
rs2301772 Chr.16: 57372487-57372487 on Build GRCh38
rs555243239 Chr.16: 57372491-57372491 on Build GRCh38
rs943839695 Chr.16: 57372778-57372778 on Build GRCh38
rs780860613 Chr.16: 57372418-57372418 on Build GRCh38
rs756281236 Chr.16: 57372585-57372585 on Build GRCh38
rs369287265 Chr.16: 57372546-57372546 on Build GRCh38
rs761734507 Chr.16: 57372632-57372632 on Build GRCh38
rs192552070 Chr.16: 57372751-57372751 on Build GRCh38
rs190569013 Chr.16: 57372543-57372543 on Build GRCh38
rs766501728 Chr.16: 57372647-57372647 on Build GRCh38
rs377237713 Chr.16: 57372635-57372635 on Build GRCh38
rs574449791 Chr.16: 57372512-57372512 on Build GRCh38
rs957857965 Chr.16: 57372760-57372760 on Build GRCh38
rs199688430 Chr.16: 57372588-57372588 on Build GRCh38
rs138544995 Chr.16: 57372579-57372579 on Build GRCh38
rs773579770 Chr.16: 57372636-57372636 on Build GRCh38
rs573899501 Chr.16: 57372772-57372772 on Build GRCh38
rs539510072 Chr.16: 57372734-57372734 on Build GRCh38
rs138031688 Chr.16: 57372428-57372428 on Build GRCh38
rs763934024 Chr.16: 57372564-57372564 on Build GRCh38
rs1052175252 Chr.16: 57372429-57372429 on Build GRCh38
rs779267755 Chr.16: 57372540-57372540 on Build GRCh38
rs932291222 Chr.16: 57372798-57372798 on Build GRCh38
rs779273242 Chr.16: 57372596-57372596 on Build GRCh38
rs1050622979 Chr.16: 57372616-57372616 on Build GRCh38
rs892156840 Chr.16: 57372442-57372442 on Build GRCh38
rs150111366 Chr.16: 57372577-57372577 on Build GRCh38
rs537781239 Chr.16: 57372779-57372779 on Build GRCh38
rs996375881 Chr.16: 57372484-57372484 on Build GRCh38
rs776243031 Chr.16: 57372545-57372545 on Build GRCh38
rs774264096 Chr.16: 57372575-57372575 on Build GRCh38
rs745525963 Chr.16: 57372440-57372440 on Build GRCh38
rs200444495 Chr.16: 57372527-57372527 on Build GRCh38
rs200604856 Chr.16: 57372595-57372595 on Build GRCh38
rs750583203 Chr.16: 57372531-57372531 on Build GRCh38
rs372653051 Chr.16: 57372556-57372556 on Build GRCh38
rs529365298 Chr.16: 57372766-57372766 on Build GRCh38
rs902067648 Chr.16: 57372460-57372460 on Build GRCh38
rs753160948 Chr.16: 57372673-57372673 on Build GRCh38
rs768691831 Chr.16: 57372624-57372624 on Build GRCh38
rs1050655060 Chr.16: 57372803-57372803 on Build GRCh38
rs897877132 Chr.16: 57372724-57372724 on Build GRCh38
rs546317124 Chr.16: 57372817-57372817 on Build GRCh38
rs564408894 Chr.16: 57372417-57372417 on Build GRCh38

View Full Product Details