Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 57383872-57384364 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CX3CL1
Entrez Gene ID
6376
Gene Name
C-X3-C motif chemokine ligand 1
Gene Aliases
ABCD-3, C3Xkine, CXC3, CXC3C, NTN, NTT, SCYD1, fractalkine, neurotactin
UniGene
Hs.531668
Gene Chromosome Location
Chr.16: 57372461-57385048 on Build GRCh38
Transcripts
NM_001304392.1 NM_002996.4

SNP Detail

SNP ID Chromosome Location
rs986229970 Chr.16: 57384190-57384190 on Build GRCh38
rs113692868 Chr.16: 57384032-57384032 on Build GRCh38
rs968017417 Chr.16: 57383931-57383931 on Build GRCh38
rs150426266 Chr.16: 57384186-57384186 on Build GRCh38
rs543866192 Chr.16: 57384168-57384168 on Build GRCh38
rs1020564236 Chr.16: 57383919-57383919 on Build GRCh38
rs772768730 Chr.16: 57384240-57384240 on Build GRCh38
rs35197397 Chr.16: 57383944-57383945 on Build GRCh38
rs926476910 Chr.16: 57384290-57384290 on Build GRCh38
rs140521964 Chr.16: 57383947-57383947 on Build GRCh38
rs2239352 Chr.16: 57384276-57384276 on Build GRCh38
rs761246617 Chr.16: 57384157-57384157 on Build GRCh38
rs765064302 Chr.16: 57384333-57384334 on Build GRCh38
rs376302702 Chr.16: 57384060-57384061 on Build GRCh38
rs1053531066 Chr.16: 57384318-57384318 on Build GRCh38
rs377382852 Chr.16: 57384129-57384129 on Build GRCh38
rs573445070 Chr.16: 57384027-57384027 on Build GRCh38
rs561347135 Chr.16: 57384154-57384154 on Build GRCh38
rs1046248340 Chr.16: 57384047-57384047 on Build GRCh38
rs549263909 Chr.16: 57384316-57384316 on Build GRCh38
rs558360410 Chr.16: 57384000-57384000 on Build GRCh38
rs781054593 Chr.16: 57384281-57384281 on Build GRCh38
rs566079435 Chr.16: 57384035-57384035 on Build GRCh38
rs184315910 Chr.16: 57383999-57383999 on Build GRCh38
rs2239351 Chr.16: 57383967-57383967 on Build GRCh38
rs919245118 Chr.16: 57384267-57384267 on Build GRCh38
rs575874363 Chr.16: 57383982-57383982 on Build GRCh38
rs554494485 Chr.16: 57384307-57384307 on Build GRCh38
rs957689305 Chr.16: 57383916-57383916 on Build GRCh38
rs866387691 Chr.16: 57384302-57384302 on Build GRCh38
rs986177540 Chr.16: 57383972-57383972 on Build GRCh38
rs893595403 Chr.16: 57384324-57384324 on Build GRCh38
rs41533845 Chr.16: 57383918-57383918 on Build GRCh38
rs573309038 Chr.16: 57384160-57384160 on Build GRCh38
rs1009258245 Chr.16: 57384138-57384138 on Build GRCh38
rs1053439770 Chr.16: 57384082-57384082 on Build GRCh38
rs2239350 Chr.16: 57383956-57383956 on Build GRCh38
rs540964149 Chr.16: 57384039-57384039 on Build GRCh38
rs201549742 Chr.16: 57384055-57384056 on Build GRCh38
rs1001912673 Chr.16: 57384071-57384071 on Build GRCh38
rs953499756 Chr.16: 57384187-57384187 on Build GRCh38
rs67090371 Chr.16: 57384211-57384211 on Build GRCh38
rs138151220 Chr.16: 57384264-57384264 on Build GRCh38
rs1021036551 Chr.16: 57384153-57384153 on Build GRCh38
rs546667389 Chr.16: 57383942-57383942 on Build GRCh38
rs981955216 Chr.16: 57384289-57384289 on Build GRCh38
rs770155652 Chr.16: 57384026-57384026 on Build GRCh38
rs551269928 Chr.16: 57384223-57384223 on Build GRCh38
rs905001585 Chr.16: 57384048-57384048 on Build GRCh38
rs568425375 Chr.16: 57384340-57384340 on Build GRCh38
rs567616417 Chr.16: 57384052-57384053 on Build GRCh38

View Full Product Details