Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 57384255-57384756 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CX3CL1
Entrez Gene ID
6376
Gene Name
C-X3-C motif chemokine ligand 1
Gene Aliases
ABCD-3, C3Xkine, CXC3, CXC3C, NTN, NTT, SCYD1, fractalkine, neurotactin
UniGene
Hs.531668
Gene Chromosome Location
Chr.16: 57372461-57385048 on Build GRCh38
Transcripts
NM_001304392.1 NM_002996.4

SNP Detail

SNP ID Chromosome Location
rs577654657 Chr.16: 57384576-57384576 on Build GRCh38
rs982064807 Chr.16: 57384458-57384458 on Build GRCh38
rs551068416 Chr.16: 57384354-57384354 on Build GRCh38
rs900846842 Chr.16: 57384537-57384537 on Build GRCh38
rs569853314 Chr.16: 57384379-57384379 on Build GRCh38
rs78604892 Chr.16: 57384697-57384697 on Build GRCh38
rs989361769 Chr.16: 57384487-57384487 on Build GRCh38
rs1030557015 Chr.16: 57384361-57384361 on Build GRCh38
rs555772837 Chr.16: 57384536-57384536 on Build GRCh38
rs1037882559 Chr.16: 57384658-57384658 on Build GRCh38
rs926476910 Chr.16: 57384290-57384290 on Build GRCh38
rs1016843645 Chr.16: 57384389-57384389 on Build GRCh38
rs2239352 Chr.16: 57384276-57384276 on Build GRCh38
rs926565316 Chr.16: 57384464-57384464 on Build GRCh38
rs894355228 Chr.16: 57384486-57384486 on Build GRCh38
rs567104444 Chr.16: 57384385-57384385 on Build GRCh38
rs1049008480 Chr.16: 57384480-57384480 on Build GRCh38
rs915023136 Chr.16: 57384518-57384518 on Build GRCh38
rs970767758 Chr.16: 57384724-57384724 on Build GRCh38
rs765064302 Chr.16: 57384333-57384334 on Build GRCh38
rs573272211 Chr.16: 57384475-57384475 on Build GRCh38
rs1053531066 Chr.16: 57384318-57384318 on Build GRCh38
rs550939372 Chr.16: 57384453-57384453 on Build GRCh38
rs34047449 Chr.16: 57384544-57384544 on Build GRCh38
rs560043145 Chr.16: 57384693-57384693 on Build GRCh38
rs115095473 Chr.16: 57384641-57384641 on Build GRCh38
rs997783431 Chr.16: 57384360-57384360 on Build GRCh38
rs1026787758 Chr.16: 57384704-57384704 on Build GRCh38
rs549263909 Chr.16: 57384316-57384316 on Build GRCh38
rs569354426 Chr.16: 57384357-57384357 on Build GRCh38
rs781054593 Chr.16: 57384281-57384281 on Build GRCh38
rs933675909 Chr.16: 57384548-57384548 on Build GRCh38
rs747128853 Chr.16: 57384669-57384669 on Build GRCh38
rs1026317366 Chr.16: 57384440-57384440 on Build GRCh38
rs149125746 Chr.16: 57384524-57384524 on Build GRCh38
rs2239353 Chr.16: 57384365-57384365 on Build GRCh38
rs963529398 Chr.16: 57384393-57384393 on Build GRCh38
rs554494485 Chr.16: 57384307-57384307 on Build GRCh38
rs371139054 Chr.16: 57384353-57384353 on Build GRCh38
rs937927338 Chr.16: 57384465-57384465 on Build GRCh38
rs756954379 Chr.16: 57384598-57384598 on Build GRCh38
rs542523601 Chr.16: 57384731-57384731 on Build GRCh38
rs866387691 Chr.16: 57384302-57384302 on Build GRCh38
rs1049407701 Chr.16: 57384549-57384549 on Build GRCh38
rs893595403 Chr.16: 57384324-57384324 on Build GRCh38
rs1042504822 Chr.16: 57384533-57384533 on Build GRCh38
rs1050812 Chr.16: 57384644-57384644 on Build GRCh38
rs981955216 Chr.16: 57384289-57384289 on Build GRCh38
rs889412346 Chr.16: 57384626-57384626 on Build GRCh38
rs866774072 Chr.16: 57384646-57384646 on Build GRCh38
rs534183428 Chr.16: 57384411-57384411 on Build GRCh38
rs568425375 Chr.16: 57384340-57384340 on Build GRCh38

View Full Product Details