Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 57528775-57529277 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC102A
Entrez Gene ID
92922
Gene Name
coiled-coil domain containing 102A
Gene Aliases
-
UniGene
Hs.644611
Gene Chromosome Location
Chr.16: 57512178-57538305 on Build GRCh38
Transcripts
NM_033212.3 XM_011523469.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6412945 c.235G>T p.E79* Chr.16: 57528943-57528943 on Build GRCh38
6229742 c.392C>T p.A131V Chr.16: 57528786-57528786 on Build GRCh38
4619723 c.314G>T p.R105L Chr.16: 57528864-57528864 on Build GRCh38
6404434 c.302C>T p.T101I Chr.16: 57528876-57528876 on Build GRCh38
3988544 c.240G>T p.L80L Chr.16: 57528938-57528938 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752967457 Chr.16: 57528835-57528835 on Build GRCh38
rs202154704 Chr.16: 57528927-57528927 on Build GRCh38
rs767457149 Chr.16: 57528963-57528963 on Build GRCh38
rs751755398 Chr.16: 57528852-57528852 on Build GRCh38
rs750946071 Chr.16: 57528966-57528966 on Build GRCh38
rs12935069 Chr.16: 57528892-57528892 on Build GRCh38
rs775536376 Chr.16: 57529140-57529140 on Build GRCh38
rs750848405 Chr.16: 57528877-57528877 on Build GRCh38
rs770971705 Chr.16: 57528806-57528806 on Build GRCh38
rs754271790 Chr.16: 57528986-57528986 on Build GRCh38
rs772655958 Chr.16: 57528804-57528804 on Build GRCh38
rs543367134 Chr.16: 57528795-57528795 on Build GRCh38
rs774691635 Chr.16: 57528944-57528944 on Build GRCh38
rs760918275 Chr.16: 57529186-57529214 on Build GRCh38
rs17845157 Chr.16: 57528893-57528893 on Build GRCh38
rs745370134 Chr.16: 57529122-57529122 on Build GRCh38
rs755307720 Chr.16: 57528994-57528994 on Build GRCh38
rs762836643 Chr.16: 57528928-57528928 on Build GRCh38
rs764397301 Chr.16: 57528837-57528837 on Build GRCh38
rs760169112 Chr.16: 57528805-57528805 on Build GRCh38
rs746571621 Chr.16: 57528915-57528915 on Build GRCh38
rs777401641 Chr.16: 57528907-57528907 on Build GRCh38
rs547141099 Chr.16: 57529218-57529218 on Build GRCh38
rs745722625 Chr.16: 57528924-57528924 on Build GRCh38
rs867053153 Chr.16: 57529101-57529101 on Build GRCh38
rs757250629 Chr.16: 57528863-57528863 on Build GRCh38
rs764898448 Chr.16: 57528823-57528823 on Build GRCh38
rs762849453 Chr.16: 57528814-57528829 on Build GRCh38
rs777453898 Chr.16: 57528995-57528995 on Build GRCh38
rs746599169 Chr.16: 57529002-57529002 on Build GRCh38
rs776657960 Chr.16: 57528920-57528920 on Build GRCh38
rs766862771 Chr.16: 57528984-57528984 on Build GRCh38
rs181771323 Chr.16: 57529233-57529233 on Build GRCh38
rs769918726 Chr.16: 57529126-57529126 on Build GRCh38
rs758107574 Chr.16: 57528896-57528896 on Build GRCh38
rs4784815 Chr.16: 57529199-57529199 on Build GRCh38
rs761976761 Chr.16: 57528959-57528959 on Build GRCh38
rs549604567 Chr.16: 57529005-57529005 on Build GRCh38
rs769724809 Chr.16: 57528798-57528802 on Build GRCh38
rs773003489 Chr.16: 57528809-57528810 on Build GRCh38
rs764450282 Chr.16: 57528943-57528943 on Build GRCh38
rs766086236 Chr.16: 57528832-57528843 on Build GRCh38
rs770294863 Chr.16: 57528919-57528919 on Build GRCh38
rs776404217 Chr.16: 57528807-57528807 on Build GRCh38
rs79928291 Chr.16: 57529061-57529061 on Build GRCh38
rs775916723 Chr.16: 57529010-57529011 on Build GRCh38
rs756521206 Chr.16: 57528980-57528980 on Build GRCh38
rs781748241 Chr.16: 57528870-57528870 on Build GRCh38
rs756533105 Chr.16: 57528884-57528884 on Build GRCh38
rs759395585 Chr.16: 57528820-57528820 on Build GRCh38
rs867974143 Chr.16: 57528901-57528901 on Build GRCh38
rs747816345 Chr.16: 57528895-57528895 on Build GRCh38

View Full Product Details