Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 57897377-57897912 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNGB1
Entrez Gene ID
1258
Gene Name
cyclic nucleotide gated channel beta 1
Gene Aliases
CNCG2, CNCG3L, CNCG4, CNG4, CNGB1B, GAR1, GARP, GARP2, RCNC2, RCNCb, RCNCbeta, RP45
UniGene
Hs.147062
Gene Chromosome Location
Chr.16: 57882340-57971116 on Build GRCh38
Transcripts
NM_001286130.1 NM_001297.4 XM_011522870.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3510568 c.3086G>A p.G1029E Chr.16: 57897805-57897805 on Build GRCh38
3510569 c.3045G>A p.G1015G Chr.16: 57897846-57897846 on Build GRCh38
3510570 c.3028T>C p.L1010L Chr.16: 57897863-57897863 on Build GRCh38
704026 c.3030G>A p.L1010L Chr.16: 57897861-57897861 on Build GRCh38
1378607 c.2989C>T p.R997C Chr.16: 57897902-57897902 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs954251923 Chr.16: 57897876-57897876 on Build GRCh38
rs374172975 Chr.16: 57897409-57897409 on Build GRCh38
rs1013586071 Chr.16: 57897811-57897811 on Build GRCh38
rs765061130 Chr.16: 57897872-57897872 on Build GRCh38
rs779871357 Chr.16: 57897761-57897761 on Build GRCh38
rs767619701 Chr.16: 57897529-57897529 on Build GRCh38
rs774960446 Chr.16: 57897426-57897426 on Build GRCh38
rs367764281 Chr.16: 57897459-57897459 on Build GRCh38
rs748180946 Chr.16: 57897510-57897510 on Build GRCh38
rs773593999 Chr.16: 57897454-57897454 on Build GRCh38
rs200287627 Chr.16: 57897505-57897505 on Build GRCh38
rs750434115 Chr.16: 57897752-57897752 on Build GRCh38
rs774133244 Chr.16: 57897592-57897592 on Build GRCh38
rs374437422 Chr.16: 57897590-57897590 on Build GRCh38
rs772877765 Chr.16: 57897858-57897858 on Build GRCh38
rs780864617 Chr.16: 57897540-57897540 on Build GRCh38
rs748288939 Chr.16: 57897569-57897569 on Build GRCh38
rs377767022 Chr.16: 57897788-57897788 on Build GRCh38
rs756012260 Chr.16: 57897535-57897535 on Build GRCh38
rs369503135 Chr.16: 57897507-57897507 on Build GRCh38
rs767959077 Chr.16: 57897447-57897447 on Build GRCh38
rs765306729 Chr.16: 57897554-57897557 on Build GRCh38
rs750087887 Chr.16: 57897573-57897573 on Build GRCh38
rs199704464 Chr.16: 57897819-57897819 on Build GRCh38
rs780388472 Chr.16: 57897503-57897503 on Build GRCh38
rs370740471 Chr.16: 57897877-57897877 on Build GRCh38
rs761481030 Chr.16: 57897593-57897593 on Build GRCh38
rs770189177 Chr.16: 57897824-57897824 on Build GRCh38
rs545721406 Chr.16: 57897417-57897417 on Build GRCh38
rs1048752301 Chr.16: 57897710-57897710 on Build GRCh38
rs928961626 Chr.16: 57897642-57897642 on Build GRCh38
rs192178968 Chr.16: 57897517-57897517 on Build GRCh38
rs937948897 Chr.16: 57897739-57897739 on Build GRCh38
rs773698242 Chr.16: 57897514-57897514 on Build GRCh38
rs902632425 Chr.16: 57897828-57897828 on Build GRCh38
rs763490882 Chr.16: 57897478-57897478 on Build GRCh38
rs779688908 Chr.16: 57897541-57897541 on Build GRCh38
rs757969882 Chr.16: 57897476-57897476 on Build GRCh38
rs539304668 Chr.16: 57897504-57897504 on Build GRCh38
rs775265608 Chr.16: 57897518-57897518 on Build GRCh38
rs578138182 Chr.16: 57897491-57897491 on Build GRCh38
rs148999583 Chr.16: 57897524-57897524 on Build GRCh38
rs151078189 Chr.16: 57897591-57897591 on Build GRCh38
rs760708332 Chr.16: 57897755-57897755 on Build GRCh38
rs753575496 Chr.16: 57897770-57897770 on Build GRCh38
rs767102691 Chr.16: 57897594-57897594 on Build GRCh38
rs1030574179 Chr.16: 57897657-57897657 on Build GRCh38
rs554514295 Chr.16: 57897740-57897740 on Build GRCh38
rs747778655 Chr.16: 57897833-57897833 on Build GRCh38
rs758524873 Chr.16: 57897795-57897795 on Build GRCh38
rs200487336 Chr.16: 57897859-57897859 on Build GRCh38
rs755959274 Chr.16: 57897496-57897496 on Build GRCh38
rs777856371 Chr.16: 57897800-57897800 on Build GRCh38
rs752290959 Chr.16: 57897464-57897464 on Build GRCh38
rs918890117 Chr.16: 57897635-57897635 on Build GRCh38
rs897497044 Chr.16: 57897462-57897462 on Build GRCh38
rs200581517 Chr.16: 57897492-57897492 on Build GRCh38
rs779121632 Chr.16: 57897565-57897565 on Build GRCh38
rs1021456731 Chr.16: 57897820-57897820 on Build GRCh38
rs200202348 Chr.16: 57897511-57897511 on Build GRCh38
rs771798164 Chr.16: 57897834-57897834 on Build GRCh38
rs142930143 Chr.16: 57897525-57897525 on Build GRCh38
rs28483578 Chr.16: 57897605-57897605 on Build GRCh38
rs201023939 Chr.16: 57897410-57897410 on Build GRCh38
rs1001259747 Chr.16: 57897857-57897857 on Build GRCh38
rs752889241 Chr.16: 57897789-57897789 on Build GRCh38
rs562124500 Chr.16: 57897713-57897713 on Build GRCh38
rs751034406 Chr.16: 57897485-57897485 on Build GRCh38
rs753353134 Chr.16: 57897489-57897489 on Build GRCh38
rs377324991 Chr.16: 57897585-57897585 on Build GRCh38
rs754741749 Chr.16: 57897777-57897777 on Build GRCh38
rs886479125 Chr.16: 57897670-57897670 on Build GRCh38
rs944407581 Chr.16: 57897612-57897612 on Build GRCh38
rs531122937 Chr.16: 57897557-57897557 on Build GRCh38
rs774186674 Chr.16: 57897829-57897829 on Build GRCh38
rs975836900 Chr.16: 57897617-57897617 on Build GRCh38
rs761350175 Chr.16: 57897528-57897528 on Build GRCh38
rs750379218 Chr.16: 57897533-57897533 on Build GRCh38
rs776629298 Chr.16: 57897866-57897866 on Build GRCh38
rs760975936 Chr.16: 57897456-57897456 on Build GRCh38
rs781682721 Chr.16: 57897814-57897814 on Build GRCh38
rs896334152 Chr.16: 57897767-57897767 on Build GRCh38
rs763732525 Chr.16: 57897421-57897421 on Build GRCh38
rs747053697 Chr.16: 57897812-57897812 on Build GRCh38
rs756806434 Chr.16: 57897496-57897497 on Build GRCh38
rs758652454 Chr.16: 57897883-57897883 on Build GRCh38
rs759293941 Chr.16: 57897867-57897867 on Build GRCh38
rs766297740 Chr.16: 57897766-57897766 on Build GRCh38
rs375338200 Chr.16: 57897890-57897890 on Build GRCh38
rs753617403 Chr.16: 57897547-57897547 on Build GRCh38
rs762202033 Chr.16: 57897436-57897436 on Build GRCh38
rs368830470 Chr.16: 57897586-57897586 on Build GRCh38
rs778294932 Chr.16: 57897579-57897579 on Build GRCh38

View Full Product Details