Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 58392255-58392748 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GINS3
Entrez Gene ID
64785
Gene Name
GINS complex subunit 3
Gene Aliases
PSF3
UniGene
Hs.47125
Gene Chromosome Location
Chr.16: 58392394-58406144 on Build GRCh38
Transcripts
NM_001126129.1 NM_001126130.1 NM_022770.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6555772 c.26A>T p.E9V Chr.16: 58392627-58392627 on Build GRCh38
971976 c.71A>G p.D24G Chr.16: 58392672-58392672 on Build GRCh38
6555771 c.26A>T p.E9V Chr.16: 58392627-58392627 on Build GRCh38
971977 c.100G>A p.V34M Chr.16: 58392701-58392701 on Build GRCh38
7098180 c.82T>C p.S28P Chr.16: 58392683-58392683 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760496922 Chr.16: 58392647-58392649 on Build GRCh38
rs3214480 Chr.16: 58392368-58392369 on Build GRCh38
rs775831664 Chr.16: 58392727-58392727 on Build GRCh38
rs962485238 Chr.16: 58392299-58392299 on Build GRCh38
rs936492421 Chr.16: 58392288-58392288 on Build GRCh38
rs576808262 Chr.16: 58392461-58392461 on Build GRCh38
rs62042599 Chr.16: 58392320-58392320 on Build GRCh38
rs775847847 Chr.16: 58392658-58392658 on Build GRCh38
rs765651451 Chr.16: 58392726-58392726 on Build GRCh38
rs971019267 Chr.16: 58392523-58392523 on Build GRCh38
rs748447484 Chr.16: 58392697-58392697 on Build GRCh38
rs760739147 Chr.16: 58392621-58392621 on Build GRCh38
rs773567768 Chr.16: 58392655-58392655 on Build GRCh38
rs868693014 Chr.16: 58392707-58392707 on Build GRCh38
rs1034556233 Chr.16: 58392403-58392403 on Build GRCh38
rs766786393 Chr.16: 58392519-58392519 on Build GRCh38
rs537549248 Chr.16: 58392431-58392431 on Build GRCh38
rs1040741132 Chr.16: 58392321-58392321 on Build GRCh38
rs942610927 Chr.16: 58392604-58392604 on Build GRCh38
rs546901449 Chr.16: 58392308-58392308 on Build GRCh38
rs1010305656 Chr.16: 58392389-58392389 on Build GRCh38
rs555054408 Chr.16: 58392361-58392361 on Build GRCh38
rs200341369 Chr.16: 58392579-58392579 on Build GRCh38
rs749114767 Chr.16: 58392588-58392588 on Build GRCh38
rs914391884 Chr.16: 58392596-58392596 on Build GRCh38
rs967792027 Chr.16: 58392513-58392513 on Build GRCh38
rs972487509 Chr.16: 58392512-58392512 on Build GRCh38
rs779885077 Chr.16: 58392670-58392670 on Build GRCh38
rs754865985 Chr.16: 58392688-58392688 on Build GRCh38
rs374613772 Chr.16: 58392724-58392724 on Build GRCh38
rs1006839436 Chr.16: 58392474-58392474 on Build GRCh38
rs772185636 Chr.16: 58392702-58392702 on Build GRCh38
rs1024281711 Chr.16: 58392509-58392509 on Build GRCh38
rs1046794221 Chr.16: 58392644-58392644 on Build GRCh38
rs541351258 Chr.16: 58392528-58392528 on Build GRCh38
rs193241920 Chr.16: 58392719-58392719 on Build GRCh38
rs1005714920 Chr.16: 58392394-58392394 on Build GRCh38
rs977923652 Chr.16: 58392281-58392281 on Build GRCh38
rs897965692 Chr.16: 58392671-58392671 on Build GRCh38
rs759925901 Chr.16: 58392624-58392624 on Build GRCh38
rs201487958 Chr.16: 58392704-58392704 on Build GRCh38
rs201862849 Chr.16: 58392713-58392713 on Build GRCh38
rs528437020 Chr.16: 58392284-58392284 on Build GRCh38
rs773423789 Chr.16: 58392716-58392716 on Build GRCh38
rs397839962 Chr.16: 58392370-58392371 on Build GRCh38
rs765555442 Chr.16: 58392631-58392631 on Build GRCh38
rs1053701319 Chr.16: 58392304-58392304 on Build GRCh38
rs533255262 Chr.16: 58392303-58392303 on Build GRCh38
rs558566590 Chr.16: 58392441-58392441 on Build GRCh38
rs142471085 Chr.16: 58392538-58392538 on Build GRCh38
rs115821869 Chr.16: 58392311-58392311 on Build GRCh38
rs778022038 Chr.16: 58392414-58392414 on Build GRCh38
rs755598024 Chr.16: 58392652-58392652 on Build GRCh38
rs923736448 Chr.16: 58392282-58392282 on Build GRCh38
rs769952973 Chr.16: 58392655-58392658 on Build GRCh38
rs900800911 Chr.16: 58392640-58392640 on Build GRCh38
rs768529144 Chr.16: 58392593-58392593 on Build GRCh38
rs375975025 Chr.16: 58392571-58392571 on Build GRCh38
rs3743565 Chr.16: 58392278-58392278 on Build GRCh38
rs777044261 Chr.16: 58392723-58392723 on Build GRCh38
rs888573667 Chr.16: 58392371-58392371 on Build GRCh38
rs918239732 Chr.16: 58392307-58392307 on Build GRCh38
rs187767416 Chr.16: 58392366-58392366 on Build GRCh38
rs937264072 Chr.16: 58392541-58392541 on Build GRCh38
rs137882871 Chr.16: 58392615-58392615 on Build GRCh38
rs763185349 Chr.16: 58392709-58392709 on Build GRCh38
rs149042036 Chr.16: 58392623-58392623 on Build GRCh38
rs3833045 Chr.16: 58392367-58392368 on Build GRCh38
rs756801247 Chr.16: 58392552-58392552 on Build GRCh38
rs893241859 Chr.16: 58392375-58392375 on Build GRCh38
rs771004609 Chr.16: 58392721-58392721 on Build GRCh38
rs766810427 Chr.16: 58392622-58392622 on Build GRCh38
rs145143759 Chr.16: 58392641-58392641 on Build GRCh38
rs749202312 Chr.16: 58392674-58392674 on Build GRCh38
rs745880484 Chr.16: 58392555-58392555 on Build GRCh38
rs977465020 Chr.16: 58392516-58392516 on Build GRCh38
rs200442342 Chr.16: 58392595-58392595 on Build GRCh38
rs751265242 Chr.16: 58392632-58392632 on Build GRCh38
rs946139982 Chr.16: 58392620-58392620 on Build GRCh38
rs1005662563 Chr.16: 58392661-58392661 on Build GRCh38
rs1042132919 Chr.16: 58392401-58392401 on Build GRCh38
rs780549349 Chr.16: 58392554-58392554 on Build GRCh38
rs779978571 Chr.16: 58392580-58392580 on Build GRCh38
rs368074568 Chr.16: 58392526-58392526 on Build GRCh38
rs1016420936 Chr.16: 58392507-58392507 on Build GRCh38
rs749908545 Chr.16: 58392649-58392649 on Build GRCh38
rs1029165810 Chr.16: 58392448-58392448 on Build GRCh38
rs953548983 Chr.16: 58392460-58392460 on Build GRCh38
rs900851986 Chr.16: 58392350-58392350 on Build GRCh38
rs778836316 Chr.16: 58392693-58392693 on Build GRCh38
rs767123662 Chr.16: 58392647-58392647 on Build GRCh38
rs989344311 Chr.16: 58392546-58392546 on Build GRCh38

View Full Product Details