Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 66613397-66613896 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CMTM3
Entrez Gene ID
123920
Gene Name
CKLF like MARVEL transmembrane domain containing 3
Gene Aliases
BNAS2, CKLFSF3
UniGene
Hs.298198
Gene Chromosome Location
Chr.16: 66604032-66613892 on Build GRCh38
Transcripts
NM_144601.3 NM_181553.2 NR_037613.1 NR_037619.1 XM_006721131.1 XM_011522861.1 XM_017022954.1

Gene Symbol
CMTM4
Entrez Gene ID
146223
Gene Name
CKLF like MARVEL transmembrane domain containing 4
Gene Aliases
CKLFSF4
UniGene
Hs.643961
Gene Chromosome Location
Chr.16: 66598178-66696707 on Build GRCh38
Transcripts
NM_144601.3 NM_181553.2 NR_037613.1 NR_037619.1 XM_006721131.1 XM_011522861.1 XM_017022954.1

SNP Detail

SNP ID Chromosome Location
rs572465844 Chr.16: 66613663-66613663 on Build GRCh38
rs906201596 Chr.16: 66613555-66613555 on Build GRCh38
rs894628438 Chr.16: 66613586-66613586 on Build GRCh38
rs545570622 Chr.16: 66613594-66613594 on Build GRCh38
rs887941912 Chr.16: 66613421-66613421 on Build GRCh38
rs931004783 Chr.16: 66613645-66613645 on Build GRCh38
rs1032193519 Chr.16: 66613858-66613858 on Build GRCh38
rs897876783 Chr.16: 66613652-66613652 on Build GRCh38
rs913740419 Chr.16: 66613765-66613765 on Build GRCh38
rs774784137 Chr.16: 66613772-66613772 on Build GRCh38
rs1056092687 Chr.16: 66613576-66613576 on Build GRCh38
rs888033304 Chr.16: 66613712-66613712 on Build GRCh38
rs981896521 Chr.16: 66613826-66613826 on Build GRCh38
rs943765319 Chr.16: 66613604-66613604 on Build GRCh38
rs979316245 Chr.16: 66613545-66613548 on Build GRCh38
rs370967832 Chr.16: 66613538-66613538 on Build GRCh38
rs966644694 Chr.16: 66613819-66613819 on Build GRCh38
rs532279805 Chr.16: 66613870-66613870 on Build GRCh38
rs945157594 Chr.16: 66613532-66613532 on Build GRCh38
rs992280262 Chr.16: 66613517-66613517 on Build GRCh38
rs187602847 Chr.16: 66613780-66613780 on Build GRCh38
rs1016452126 Chr.16: 66613598-66613598 on Build GRCh38
rs989688640 Chr.16: 66613757-66613757 on Build GRCh38
rs769646189 Chr.16: 66613540-66613547 on Build GRCh38
rs556936770 Chr.16: 66613472-66613472 on Build GRCh38
rs11552590 Chr.16: 66613524-66613524 on Build GRCh38
rs1014080416 Chr.16: 66613792-66613792 on Build GRCh38
rs1002785603 Chr.16: 66613438-66613438 on Build GRCh38
rs986004574 Chr.16: 66613724-66613724 on Build GRCh38
rs541198170 Chr.16: 66613693-66613693 on Build GRCh38
rs11552589 Chr.16: 66613688-66613688 on Build GRCh38
rs1033735891 Chr.16: 66613497-66613497 on Build GRCh38
rs896815510 Chr.16: 66613626-66613626 on Build GRCh38
rs1036809798 Chr.16: 66613605-66613605 on Build GRCh38
rs961088503 Chr.16: 66613514-66613514 on Build GRCh38
rs530085313 Chr.16: 66613721-66613721 on Build GRCh38
rs927605867 Chr.16: 66613868-66613868 on Build GRCh38
rs932469411 Chr.16: 66613558-66613558 on Build GRCh38
rs563204531 Chr.16: 66613838-66613838 on Build GRCh38
rs767927140 Chr.16: 66613735-66613735 on Build GRCh38
rs925207459 Chr.16: 66613549-66613549 on Build GRCh38
rs185244564 Chr.16: 66613715-66613715 on Build GRCh38
rs576816487 Chr.16: 66613482-66613482 on Build GRCh38
rs945081088 Chr.16: 66613812-66613812 on Build GRCh38
rs759901051 Chr.16: 66613726-66613726 on Build GRCh38
rs565357688 Chr.16: 66613636-66613636 on Build GRCh38
rs1000830569 Chr.16: 66613833-66613833 on Build GRCh38
rs565393018 Chr.16: 66613574-66613574 on Build GRCh38

View Full Product Details