Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 66720815-66721318 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DYNC1LI2
Entrez Gene ID
1783
Gene Name
dynein cytoplasmic 1 light intermediate chain 2
Gene Aliases
DNCLI2, LIC2
UniGene
Hs.369068
Gene Chromosome Location
Chr.16: 66720893-66751828 on Build GRCh38
Transcripts
NM_001286157.1 NM_001323955.1 NM_006141.2 NR_136658.1

SNP Detail

SNP ID Chromosome Location
rs769002788 Chr.16: 66721071-66721071 on Build GRCh38
rs142105386 Chr.16: 66721119-66721119 on Build GRCh38
rs909246714 Chr.16: 66721116-66721116 on Build GRCh38
rs186866232 Chr.16: 66720871-66720871 on Build GRCh38
rs1017786333 Chr.16: 66721040-66721040 on Build GRCh38
rs1042240225 Chr.16: 66721174-66721174 on Build GRCh38
rs1007845709 Chr.16: 66721031-66721031 on Build GRCh38
rs535388698 Chr.16: 66720983-66720983 on Build GRCh38
rs772769223 Chr.16: 66720964-66720964 on Build GRCh38
rs966572761 Chr.16: 66721047-66721047 on Build GRCh38
rs568248032 Chr.16: 66720984-66720984 on Build GRCh38
rs896363450 Chr.16: 66720912-66720913 on Build GRCh38
rs762645518 Chr.16: 66720975-66720975 on Build GRCh38
rs750814775 Chr.16: 66720889-66720889 on Build GRCh38
rs1032033373 Chr.16: 66721003-66721003 on Build GRCh38
rs1026789264 Chr.16: 66721087-66721087 on Build GRCh38
rs1053438891 Chr.16: 66721283-66721283 on Build GRCh38
rs151179697 Chr.16: 66721125-66721125 on Build GRCh38
rs984738413 Chr.16: 66721106-66721106 on Build GRCh38
rs766882420 Chr.16: 66720935-66720935 on Build GRCh38
rs917840857 Chr.16: 66721126-66721126 on Build GRCh38
rs564249643 Chr.16: 66721243-66721243 on Build GRCh38
rs573728206 Chr.16: 66720888-66720888 on Build GRCh38
rs949212292 Chr.16: 66721137-66721137 on Build GRCh38
rs923666372 Chr.16: 66721234-66721234 on Build GRCh38
rs950849997 Chr.16: 66721096-66721096 on Build GRCh38
rs572861218 Chr.16: 66720929-66720929 on Build GRCh38
rs148671531 Chr.16: 66720911-66720911 on Build GRCh38
rs370389034 Chr.16: 66720995-66720995 on Build GRCh38
rs544427551 Chr.16: 66721195-66721195 on Build GRCh38
rs893377875 Chr.16: 66721008-66721008 on Build GRCh38
rs984823085 Chr.16: 66720861-66720861 on Build GRCh38
rs570641476 Chr.16: 66720847-66720847 on Build GRCh38
rs902272713 Chr.16: 66720990-66720990 on Build GRCh38
rs11553682 Chr.16: 66720950-66720950 on Build GRCh38
rs185055465 Chr.16: 66721286-66721286 on Build GRCh38
rs937922560 Chr.16: 66721117-66721117 on Build GRCh38
rs976286391 Chr.16: 66721054-66721054 on Build GRCh38
rs936461400 Chr.16: 66721254-66721254 on Build GRCh38
rs192153510 Chr.16: 66721124-66721124 on Build GRCh38
rs774072142 Chr.16: 66721178-66721178 on Build GRCh38

View Full Product Details