Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 66722375-66722888 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DYNC1LI2
Entrez Gene ID
1783
Gene Name
dynein cytoplasmic 1 light intermediate chain 2
Gene Aliases
DNCLI2, LIC2
UniGene
Hs.369068
Gene Chromosome Location
Chr.16: 66720893-66751828 on Build GRCh38
Transcripts
NM_001286157.1 NM_001323955.1 NM_006141.2 NR_136658.1

SNP Detail

SNP ID Chromosome Location
rs1013843695 Chr.16: 66722731-66722731 on Build GRCh38
rs1040946853 Chr.16: 66722833-66722833 on Build GRCh38
rs948220720 Chr.16: 66722809-66722809 on Build GRCh38
rs942314141 Chr.16: 66722787-66722787 on Build GRCh38
rs960590279 Chr.16: 66722730-66722730 on Build GRCh38
rs1023427407 Chr.16: 66722734-66722734 on Build GRCh38
rs989296941 Chr.16: 66722554-66722554 on Build GRCh38
rs752616575 Chr.16: 66722532-66722532 on Build GRCh38
rs560335414 Chr.16: 66722851-66722851 on Build GRCh38
rs1047994168 Chr.16: 66722429-66722429 on Build GRCh38
rs982387820 Chr.16: 66722796-66722796 on Build GRCh38
rs903889709 Chr.16: 66722627-66722627 on Build GRCh38
rs922164701 Chr.16: 66722848-66722848 on Build GRCh38
rs996238551 Chr.16: 66722557-66722557 on Build GRCh38
rs565603735 Chr.16: 66722801-66722801 on Build GRCh38
rs908115639 Chr.16: 66722752-66722752 on Build GRCh38
rs748896149 Chr.16: 66722820-66722820 on Build GRCh38
rs919661228 Chr.16: 66722763-66722763 on Build GRCh38
rs906625944 Chr.16: 66722860-66722860 on Build GRCh38
rs769292684 Chr.16: 66722750-66722750 on Build GRCh38
rs951072666 Chr.16: 66722773-66722773 on Build GRCh38
rs557996216 Chr.16: 66722440-66722440 on Build GRCh38
rs774871003 Chr.16: 66722762-66722762 on Build GRCh38
rs1044199241 Chr.16: 66722623-66722623 on Build GRCh38
rs929629123 Chr.16: 66722711-66722711 on Build GRCh38
rs376649689 Chr.16: 66722417-66722417 on Build GRCh38
rs900593623 Chr.16: 66722553-66722553 on Build GRCh38
rs957961811 Chr.16: 66722536-66722536 on Build GRCh38
rs540333718 Chr.16: 66722712-66722712 on Build GRCh38
rs963782634 Chr.16: 66722609-66722609 on Build GRCh38
rs922347422 Chr.16: 66722700-66722701 on Build GRCh38
rs193134950 Chr.16: 66722458-66722458 on Build GRCh38
rs1018150118 Chr.16: 66722572-66722572 on Build GRCh38
rs908145453 Chr.16: 66722819-66722819 on Build GRCh38
rs371571850 Chr.16: 66722809-66722812 on Build GRCh38
rs185638863 Chr.16: 66722519-66722519 on Build GRCh38
rs999514571 Chr.16: 66722648-66722648 on Build GRCh38
rs895355628 Chr.16: 66722797-66722797 on Build GRCh38
rs1040519435 Chr.16: 66722487-66722487 on Build GRCh38

View Full Product Details