Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 66729733-66730259 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DYNC1LI2
Entrez Gene ID
1783
Gene Name
dynein cytoplasmic 1 light intermediate chain 2
Gene Aliases
DNCLI2, LIC2
UniGene
Hs.369068
Gene Chromosome Location
Chr.16: 66720893-66751828 on Build GRCh38
Transcripts
NM_001286157.1 NM_001323955.1 NM_006141.2 NR_136658.1 XM_017023008.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6650634 c.980T>G p.F327C Chr.16: 66730173-66730173 on Build GRCh38
4061686 c.942G>T p.W314C Chr.16: 66730211-66730211 on Build GRCh38
139015 c.943G>A p.D315N Chr.16: 66730210-66730210 on Build GRCh38
5851182 c.1028C>T p.P343L Chr.16: 66730125-66730125 on Build GRCh38
139016 c.942G>A p.W314* Chr.16: 66730211-66730211 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750133866 Chr.16: 66730146-66730146 on Build GRCh38
rs917811846 Chr.16: 66730016-66730016 on Build GRCh38
rs1003859847 Chr.16: 66730108-66730108 on Build GRCh38
rs754383868 Chr.16: 66730161-66730161 on Build GRCh38
rs956670611 Chr.16: 66730050-66730050 on Build GRCh38
rs111420970 Chr.16: 66729839-66729839 on Build GRCh38
rs145562779 Chr.16: 66730121-66730121 on Build GRCh38
rs747068258 Chr.16: 66730066-66730066 on Build GRCh38
rs995406072 Chr.16: 66729971-66729971 on Build GRCh38
rs367817929 Chr.16: 66730070-66730070 on Build GRCh38
rs755378264 Chr.16: 66729805-66729805 on Build GRCh38
rs935327788 Chr.16: 66729794-66729797 on Build GRCh38
rs562078163 Chr.16: 66729954-66729954 on Build GRCh38
rs1023845924 Chr.16: 66730009-66730009 on Build GRCh38
rs200872939 Chr.16: 66730174-66730174 on Build GRCh38
rs925312003 Chr.16: 66729926-66729926 on Build GRCh38
rs776820398 Chr.16: 66730132-66730132 on Build GRCh38
rs373012856 Chr.16: 66730160-66730160 on Build GRCh38
rs1975632 Chr.16: 66729843-66729843 on Build GRCh38
rs1052755319 Chr.16: 66729794-66729794 on Build GRCh38
rs1003852029 Chr.16: 66729869-66729869 on Build GRCh38
rs542599158 Chr.16: 66729935-66729935 on Build GRCh38
rs573509717 Chr.16: 66729908-66729908 on Build GRCh38
rs1050807345 Chr.16: 66729848-66729848 on Build GRCh38
rs199834045 Chr.16: 66730077-66730077 on Build GRCh38
rs778918758 Chr.16: 66730076-66730076 on Build GRCh38
rs34706526 Chr.16: 66730166-66730166 on Build GRCh38
rs183511734 Chr.16: 66730021-66730021 on Build GRCh38
rs755028987 Chr.16: 66730072-66730072 on Build GRCh38
rs557959542 Chr.16: 66729836-66729836 on Build GRCh38
rs780941102 Chr.16: 66730098-66730098 on Build GRCh38
rs979956479 Chr.16: 66730229-66730229 on Build GRCh38
rs533478282 Chr.16: 66730090-66730090 on Build GRCh38
rs946525470 Chr.16: 66729833-66729833 on Build GRCh38
rs566955186 Chr.16: 66730149-66730149 on Build GRCh38
rs777665024 Chr.16: 66730034-66730034 on Build GRCh38
rs201855076 Chr.16: 66730206-66730206 on Build GRCh38
rs775336788 Chr.16: 66730118-66730118 on Build GRCh38
rs1044836021 Chr.16: 66730028-66730028 on Build GRCh38
rs927910357 Chr.16: 66729789-66729794 on Build GRCh38
rs988479905 Chr.16: 66729945-66729945 on Build GRCh38
rs538071538 Chr.16: 66729829-66729829 on Build GRCh38
rs767031978 Chr.16: 66730136-66730136 on Build GRCh38
rs755502936 Chr.16: 66730165-66730165 on Build GRCh38
rs976220254 Chr.16: 66729842-66729842 on Build GRCh38
rs1015572853 Chr.16: 66729887-66729887 on Build GRCh38
rs886744709 Chr.16: 66729866-66729866 on Build GRCh38
rs767987040 Chr.16: 66729786-66729790 on Build GRCh38
rs375885888 Chr.16: 66730120-66730120 on Build GRCh38
rs372962688 Chr.16: 66730228-66730228 on Build GRCh38
rs893744457 Chr.16: 66729815-66729815 on Build GRCh38
rs979540984 Chr.16: 66729874-66729874 on Build GRCh38
rs770890136 Chr.16: 66730070-66730071 on Build GRCh38
rs758526335 Chr.16: 66730168-66730168 on Build GRCh38
rs997688881 Chr.16: 66730157-66730157 on Build GRCh38
rs371020424 Chr.16: 66729850-66729850 on Build GRCh38
rs368729372 Chr.16: 66730158-66730158 on Build GRCh38
rs988185310 Chr.16: 66730054-66730054 on Build GRCh38
rs141617175 Chr.16: 66730106-66730106 on Build GRCh38
rs886882250 Chr.16: 66730128-66730128 on Build GRCh38

View Full Product Details