Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 66772580-66773021 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TERB1
Entrez Gene ID
283847
Gene Name
telomere repeat binding bouquet formation protein 1
Gene Aliases
CCDC79
UniGene
Hs.376505
Gene Chromosome Location
Chr.16: 66754410-66802801 on Build GRCh38
Transcripts
NM_001136505.1 XM_011523004.2 XM_011523005.2 XM_011523006.2 XM_011523007.2 XM_011523008.2 XM_011523009.2 XM_011523010.2 XM_011523011.2 XM_011523012.2 XM_011523014.2 XM_011523015.2 XM_011523016.2 XM_011523017.1 XM_011523018.1 XM_011523020.1 XM_017023157.1 XM_017023158.1 XM_017023159.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6318941 c.1212C>A p.H404Q Chr.16: 66772649-66772649 on Build GRCh38
1378793 c.1176G>T p.K392N Chr.16: 66772685-66772685 on Build GRCh38
4615880 c.1145delC p.P382fs*10 Chr.16: 66772716-66772716 on Build GRCh38
6326196 c.1272+5G>T p.? Chr.16: 66772584-66772584 on Build GRCh38
5385130 c.1227G>C p.K409N Chr.16: 66772634-66772634 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs918808154 Chr.16: 66772690-66772690 on Build GRCh38
rs966101948 Chr.16: 66772941-66772941 on Build GRCh38
rs778607097 Chr.16: 66772634-66772634 on Build GRCh38
rs755361111 Chr.16: 66772701-66772701 on Build GRCh38
rs903410012 Chr.16: 66772804-66772804 on Build GRCh38
rs191266990 Chr.16: 66772903-66772903 on Build GRCh38
rs946061877 Chr.16: 66772803-66772803 on Build GRCh38
rs986904088 Chr.16: 66772737-66772737 on Build GRCh38
rs1016006161 Chr.16: 66772933-66772933 on Build GRCh38
rs962019738 Chr.16: 66772938-66772938 on Build GRCh38
rs887528189 Chr.16: 66772608-66772608 on Build GRCh38
rs896195544 Chr.16: 66772769-66772769 on Build GRCh38
rs537078053 Chr.16: 66772795-66772795 on Build GRCh38
rs1017686354 Chr.16: 66772936-66772936 on Build GRCh38
rs754531131 Chr.16: 66772631-66772631 on Build GRCh38
rs1002719962 Chr.16: 66772895-66772895 on Build GRCh38
rs1054793768 Chr.16: 66772756-66772756 on Build GRCh38
rs971668380 Chr.16: 66772959-66772959 on Build GRCh38
rs919197244 Chr.16: 66772971-66772971 on Build GRCh38
rs550617621 Chr.16: 66772913-66772913 on Build GRCh38
rs531342569 Chr.16: 66772922-66772925 on Build GRCh38
rs777324184 Chr.16: 66772676-66772676 on Build GRCh38
rs747334460 Chr.16: 66772770-66772770 on Build GRCh38
rs771210509 Chr.16: 66772774-66772774 on Build GRCh38
rs1007688900 Chr.16: 66772919-66772919 on Build GRCh38
rs779051129 Chr.16: 66772817-66772817 on Build GRCh38
rs1033145848 Chr.16: 66772843-66772843 on Build GRCh38
rs1006419957 Chr.16: 66772926-66772926 on Build GRCh38
rs972995594 Chr.16: 66772665-66772665 on Build GRCh38
rs760099518 Chr.16: 66772794-66772794 on Build GRCh38
rs893166578 Chr.16: 66772801-66772801 on Build GRCh38
rs747784647 Chr.16: 66772645-66772645 on Build GRCh38
rs1034212150 Chr.16: 66772902-66772902 on Build GRCh38
rs541114227 Chr.16: 66772857-66772857 on Build GRCh38
rs771709802 Chr.16: 66772646-66772646 on Build GRCh38
rs976458789 Chr.16: 66772969-66772969 on Build GRCh38
rs755328732 Chr.16: 66772766-66772767 on Build GRCh38
rs934161080 Chr.16: 66772691-66772691 on Build GRCh38
rs957520814 Chr.16: 66772891-66772891 on Build GRCh38
rs937727849 Chr.16: 66772664-66772664 on Build GRCh38
rs911317999 Chr.16: 66772762-66772762 on Build GRCh38
rs777106740 Chr.16: 66772775-66772775 on Build GRCh38
rs377675573 Chr.16: 66772797-66772797 on Build GRCh38
rs908896220 Chr.16: 66772625-66772625 on Build GRCh38
rs765255598 Chr.16: 66772666-66772666 on Build GRCh38
rs907096134 Chr.16: 66772888-66772888 on Build GRCh38
rs755111236 Chr.16: 66772798-66772798 on Build GRCh38
rs999036948 Chr.16: 66772822-66772822 on Build GRCh38
rs1025873390 Chr.16: 66772970-66772970 on Build GRCh38
rs766947896 Chr.16: 66772643-66772643 on Build GRCh38

View Full Product Details