Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 66908030-66908530 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDH16
Entrez Gene ID
1014
Gene Name
cadherin 16
Gene Aliases
-
UniGene
Hs.513660
Gene Chromosome Location
Chr.16: 66908122-66918984 on Build GRCh38
Transcripts
NM_001204744.1 NM_001204745.1 NM_001204746.1 NM_004062.3 XM_005255770.2 XM_011522807.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7134714 c.2446C>T p.P816S Chr.16: 66908436-66908436 on Build GRCh38
260022 c.2442G>T p.K814N Chr.16: 66908440-66908440 on Build GRCh38
7131171 c.2461G>A p.D821N Chr.16: 66908421-66908421 on Build GRCh38
4670823 c.2467G>A p.V823M Chr.16: 66908415-66908415 on Build GRCh38
1629952 c.2395A>G p.I799V Chr.16: 66908487-66908487 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1028897975 Chr.16: 66908399-66908399 on Build GRCh38
rs1048618663 Chr.16: 66908156-66908156 on Build GRCh38
rs867791958 Chr.16: 66908306-66908306 on Build GRCh38
rs775289811 Chr.16: 66908360-66908360 on Build GRCh38
rs369123671 Chr.16: 66908348-66908348 on Build GRCh38
rs940400414 Chr.16: 66908094-66908094 on Build GRCh38
rs764061124 Chr.16: 66908388-66908388 on Build GRCh38
rs574892955 Chr.16: 66908225-66908225 on Build GRCh38
rs764007727 Chr.16: 66908497-66908497 on Build GRCh38
rs555850837 Chr.16: 66908073-66908073 on Build GRCh38
rs750026002 Chr.16: 66908415-66908415 on Build GRCh38
rs751030839 Chr.16: 66908401-66908401 on Build GRCh38
rs774288779 Chr.16: 66908508-66908508 on Build GRCh38
rs772128634 Chr.16: 66908455-66908455 on Build GRCh38
rs1031357062 Chr.16: 66908139-66908139 on Build GRCh38
rs557776890 Chr.16: 66908341-66908341 on Build GRCh38
rs762804685 Chr.16: 66908492-66908492 on Build GRCh38
rs184789166 Chr.16: 66908063-66908063 on Build GRCh38
rs932523166 Chr.16: 66908443-66908443 on Build GRCh38
rs971697756 Chr.16: 66908103-66908103 on Build GRCh38
rs904485416 Chr.16: 66908346-66908346 on Build GRCh38
rs141566500 Chr.16: 66908435-66908435 on Build GRCh38
rs904278404 Chr.16: 66908108-66908108 on Build GRCh38
rs971336818 Chr.16: 66908256-66908256 on Build GRCh38
rs952805238 Chr.16: 66908331-66908331 on Build GRCh38
rs766727414 Chr.16: 66908346-66908349 on Build GRCh38
rs931562829 Chr.16: 66908137-66908137 on Build GRCh38
rs971751573 Chr.16: 66908440-66908440 on Build GRCh38
rs771195731 Chr.16: 66908484-66908484 on Build GRCh38
rs1010255757 Chr.16: 66908203-66908203 on Build GRCh38
rs748160777 Chr.16: 66908437-66908437 on Build GRCh38
rs138685191 Chr.16: 66908427-66908427 on Build GRCh38
rs763942488 Chr.16: 66908222-66908222 on Build GRCh38
rs577822568 Chr.16: 66908498-66908498 on Build GRCh38
rs572059845 Chr.16: 66908297-66908297 on Build GRCh38
rs775236344 Chr.16: 66908490-66908490 on Build GRCh38
rs557000775 Chr.16: 66908193-66908193 on Build GRCh38
rs747209643 Chr.16: 66908481-66908481 on Build GRCh38
rs1025209985 Chr.16: 66908251-66908251 on Build GRCh38
rs756850918 Chr.16: 66908402-66908402 on Build GRCh38
rs150908551 Chr.16: 66908465-66908465 on Build GRCh38
rs77463171 Chr.16: 66908303-66908303 on Build GRCh38
rs369634064 Chr.16: 66908416-66908416 on Build GRCh38
rs908567724 Chr.16: 66908379-66908379 on Build GRCh38
rs150191844 Chr.16: 66908361-66908361 on Build GRCh38
rs996685252 Chr.16: 66908055-66908055 on Build GRCh38
rs764942295 Chr.16: 66908345-66908345 on Build GRCh38
rs751689698 Chr.16: 66908444-66908444 on Build GRCh38
rs920281031 Chr.16: 66908447-66908447 on Build GRCh38
rs760817475 Chr.16: 66908344-66908344 on Build GRCh38
rs980958786 Chr.16: 66908269-66908269 on Build GRCh38
rs370771037 Chr.16: 66908434-66908434 on Build GRCh38
rs999914920 Chr.16: 66908133-66908133 on Build GRCh38
rs1041395276 Chr.16: 66908411-66908411 on Build GRCh38
rs1057468204 Chr.16: 66908264-66908264 on Build GRCh38
rs904329420 Chr.16: 66908499-66908499 on Build GRCh38
rs758718784 Chr.16: 66908095-66908095 on Build GRCh38
rs766952149 Chr.16: 66908409-66908409 on Build GRCh38
rs920230316 Chr.16: 66908127-66908127 on Build GRCh38

View Full Product Details