Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 67029008-67029495 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CBFB
Entrez Gene ID
865
Gene Name
core-binding factor, beta subunit
Gene Aliases
PEBP2B
UniGene
Hs.460988
Gene Chromosome Location
Chr.16: 67029147-67101058 on Build GRCh38
Transcripts
NM_001755.2 NM_022845.2 XM_005256212.3 XM_006721321.2 XM_017023812.1 XM_017023813.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6955768 c.10G>T p.V4F Chr.16: 67029417-67029417 on Build GRCh38
1478961 c.61C>G p.L21V Chr.16: 67029468-67029468 on Build GRCh38
6950206 c.21C>G p.D7E Chr.16: 67029428-67029428 on Build GRCh38
6951428 c.2T>A p.M1K Chr.16: 67029409-67029409 on Build GRCh38
5800424 c.36C>A p.F12L Chr.16: 67029443-67029443 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs537006114 Chr.16: 67029367-67029367 on Build GRCh38
rs557593884 Chr.16: 67029189-67029194 on Build GRCh38
rs888319888 Chr.16: 67029097-67029097 on Build GRCh38
rs753837988 Chr.16: 67029377-67029377 on Build GRCh38
rs918361865 Chr.16: 67029056-67029056 on Build GRCh38
rs752659053 Chr.16: 67029362-67029362 on Build GRCh38
rs754814736 Chr.16: 67029378-67029378 on Build GRCh38
rs775966318 Chr.16: 67029425-67029425 on Build GRCh38
rs912169686 Chr.16: 67029329-67029329 on Build GRCh38
rs568086733 Chr.16: 67029207-67029207 on Build GRCh38
rs981083106 Chr.16: 67029086-67029086 on Build GRCh38
rs1035085974 Chr.16: 67029089-67029089 on Build GRCh38
rs867508880 Chr.16: 67029246-67029246 on Build GRCh38
rs865892706 Chr.16: 67029445-67029445 on Build GRCh38
rs986417062 Chr.16: 67029322-67029322 on Build GRCh38
rs751762472 Chr.16: 67029369-67029369 on Build GRCh38
rs1040751321 Chr.16: 67029035-67029036 on Build GRCh38
rs895805145 Chr.16: 67029172-67029172 on Build GRCh38
rs763020415 Chr.16: 67029364-67029364 on Build GRCh38
rs565826364 Chr.16: 67029154-67029154 on Build GRCh38
rs751599981 Chr.16: 67029374-67029386 on Build GRCh38
rs960386629 Chr.16: 67029095-67029095 on Build GRCh38
rs776808645 Chr.16: 67029404-67029404 on Build GRCh38
rs201510751 Chr.16: 67029433-67029433 on Build GRCh38
rs970044799 Chr.16: 67029080-67029080 on Build GRCh38
rs760100158 Chr.16: 67029416-67029416 on Build GRCh38
rs909736069 Chr.16: 67029394-67029394 on Build GRCh38
rs929788426 Chr.16: 67029060-67029060 on Build GRCh38
rs778771923 Chr.16: 67029465-67029465 on Build GRCh38
rs1017106102 Chr.16: 67029075-67029075 on Build GRCh38
rs754795321 Chr.16: 67029464-67029464 on Build GRCh38
rs751811349 Chr.16: 67029434-67029434 on Build GRCh38
rs943766609 Chr.16: 67029034-67029034 on Build GRCh38
rs984334537 Chr.16: 67029386-67029386 on Build GRCh38
rs1055782327 Chr.16: 67029163-67029163 on Build GRCh38
rs771353968 Chr.16: 67029399-67029399 on Build GRCh38
rs545327734 Chr.16: 67029476-67029476 on Build GRCh38
rs1000733816 Chr.16: 67029226-67029226 on Build GRCh38
rs778488249 Chr.16: 67029381-67029381 on Build GRCh38
rs1001473893 Chr.16: 67029156-67029156 on Build GRCh38
rs761975945 Chr.16: 67029440-67029440 on Build GRCh38
rs937197456 Chr.16: 67029398-67029398 on Build GRCh38
rs534683373 Chr.16: 67029174-67029174 on Build GRCh38
rs918332105 Chr.16: 67029372-67029372 on Build GRCh38
rs554672034 Chr.16: 67029198-67029198 on Build GRCh38
rs987778686 Chr.16: 67029100-67029100 on Build GRCh38
rs1020373639 Chr.16: 67029188-67029189 on Build GRCh38
rs964999771 Chr.16: 67029360-67029360 on Build GRCh38
rs764360121 Chr.16: 67029368-67029368 on Build GRCh38
rs769625293 Chr.16: 67029472-67029472 on Build GRCh38
rs562591005 Chr.16: 67029419-67029419 on Build GRCh38
rs371495589 Chr.16: 67029458-67029458 on Build GRCh38
rs370549438 Chr.16: 67029393-67029393 on Build GRCh38
rs557299355 Chr.16: 67029117-67029118 on Build GRCh38
rs1032204129 Chr.16: 67029086-67029119 on Build GRCh38
rs763073693 Chr.16: 67029426-67029426 on Build GRCh38
rs552338480 Chr.16: 67029151-67029151 on Build GRCh38
rs755947368 Chr.16: 67029370-67029370 on Build GRCh38
rs377649851 Chr.16: 67029391-67029391 on Build GRCh38
rs375495193 Chr.16: 67029385-67029385 on Build GRCh38
rs766211689 Chr.16: 67029446-67029446 on Build GRCh38
rs779674724 Chr.16: 67029371-67029371 on Build GRCh38
rs953728613 Chr.16: 67029263-67029263 on Build GRCh38

View Full Product Details