Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 67098875-67099404 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CBFB
Entrez Gene ID
865
Gene Name
core-binding factor, beta subunit
Gene Aliases
PEBP2B
UniGene
Hs.460988
Gene Chromosome Location
Chr.16: 67029147-67101058 on Build GRCh38
Transcripts
NM_001755.2 NM_022845.2 XM_005256212.3 XM_006721321.2 XM_017023812.1 XM_017023813.1

SNP Detail

SNP ID Chromosome Location
rs1017217123 Chr.16: 67099113-67099117 on Build GRCh38
rs577324926 Chr.16: 67099333-67099333 on Build GRCh38
rs1006578641 Chr.16: 67098913-67098913 on Build GRCh38
rs559399757 Chr.16: 67099283-67099283 on Build GRCh38
rs949323451 Chr.16: 67099248-67099248 on Build GRCh38
rs139771367 Chr.16: 67099364-67099364 on Build GRCh38
rs576620719 Chr.16: 67099215-67099215 on Build GRCh38
rs556786610 Chr.16: 67099213-67099213 on Build GRCh38
rs774155755 Chr.16: 67099315-67099315 on Build GRCh38
rs76848151 Chr.16: 67098923-67098923 on Build GRCh38
rs755961224 Chr.16: 67099272-67099273 on Build GRCh38
rs999672975 Chr.16: 67099043-67099043 on Build GRCh38
rs1016094830 Chr.16: 67099138-67099138 on Build GRCh38
rs902586527 Chr.16: 67099266-67099266 on Build GRCh38
rs1032954235 Chr.16: 67099075-67099075 on Build GRCh38
rs971879301 Chr.16: 67099163-67099163 on Build GRCh38
rs557399079 Chr.16: 67099312-67099312 on Build GRCh38
rs990013419 Chr.16: 67099260-67099260 on Build GRCh38
rs963216543 Chr.16: 67099139-67099139 on Build GRCh38
rs1035775704 Chr.16: 67099000-67099000 on Build GRCh38
rs1026148924 Chr.16: 67099164-67099164 on Build GRCh38
rs1013666877 Chr.16: 67098943-67098948 on Build GRCh38
rs574889843 Chr.16: 67098916-67098916 on Build GRCh38
rs910573604 Chr.16: 67099195-67099195 on Build GRCh38
rs964313684 Chr.16: 67099132-67099132 on Build GRCh38
rs983291210 Chr.16: 67099166-67099166 on Build GRCh38
rs374475448 Chr.16: 67098959-67098959 on Build GRCh38
rs1013103562 Chr.16: 67099126-67099126 on Build GRCh38
rs531721557 Chr.16: 67099230-67099234 on Build GRCh38
rs902600106 Chr.16: 67098963-67098963 on Build GRCh38
rs117985432 Chr.16: 67099187-67099187 on Build GRCh38
rs559084802 Chr.16: 67098900-67098900 on Build GRCh38
rs915406635 Chr.16: 67099269-67099269 on Build GRCh38
rs954646831 Chr.16: 67099166-67099166 on Build GRCh38
rs986157486 Chr.16: 67099085-67099085 on Build GRCh38
rs895070333 Chr.16: 67099235-67099236 on Build GRCh38
rs574712752 Chr.16: 67098990-67098990 on Build GRCh38
rs16957169 Chr.16: 67099273-67099273 on Build GRCh38
rs894210908 Chr.16: 67099016-67099016 on Build GRCh38
rs565993310 Chr.16: 67098918-67098918 on Build GRCh38
rs11641437 Chr.16: 67099286-67099286 on Build GRCh38
rs1042594669 Chr.16: 67099302-67099302 on Build GRCh38
rs764738239 Chr.16: 67099083-67099087 on Build GRCh38
rs923000619 Chr.16: 67099165-67099166 on Build GRCh38

View Full Product Details