Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 67148932-67149434 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
B3GNT9
Entrez Gene ID
84752
Gene Name
UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 9
Gene Aliases
-
UniGene
Hs.744158
Gene Chromosome Location
Chr.16: 67148102-67150999 on Build GRCh38
Transcripts
NM_033309.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5851194 c.1180G>A p.A394T Chr.16: 67149306-67149306 on Build GRCh38
4664910 c.1151G>A p.G384E Chr.16: 67149335-67149335 on Build GRCh38
6342407 c.1188C>A p.G396G Chr.16: 67149298-67149298 on Build GRCh38
5572193 c.1126T>C p.W376R Chr.16: 67149360-67149360 on Build GRCh38
3712060 c.1146G>T p.P382P Chr.16: 67149340-67149340 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778795860 Chr.16: 67149246-67149246 on Build GRCh38
rs767594917 Chr.16: 67149084-67149084 on Build GRCh38
rs932450880 Chr.16: 67149392-67149392 on Build GRCh38
rs529872490 Chr.16: 67149182-67149182 on Build GRCh38
rs375461694 Chr.16: 67149239-67149239 on Build GRCh38
rs552206907 Chr.16: 67149345-67149345 on Build GRCh38
rs904951915 Chr.16: 67149381-67149381 on Build GRCh38
rs527644990 Chr.16: 67149100-67149100 on Build GRCh38
rs774303308 Chr.16: 67149053-67149053 on Build GRCh38
rs759292285 Chr.16: 67149331-67149331 on Build GRCh38
rs921034211 Chr.16: 67149375-67149375 on Build GRCh38
rs777309795 Chr.16: 67149270-67149270 on Build GRCh38
rs926109935 Chr.16: 67149212-67149212 on Build GRCh38
rs746274790 Chr.16: 67149376-67149376 on Build GRCh38
rs574168003 Chr.16: 67149281-67149281 on Build GRCh38
rs770244868 Chr.16: 67149379-67149379 on Build GRCh38
rs770473464 Chr.16: 67149285-67149285 on Build GRCh38
rs762653444 Chr.16: 67149021-67149021 on Build GRCh38
rs368296957 Chr.16: 67149017-67149017 on Build GRCh38
rs962760128 Chr.16: 67149092-67149092 on Build GRCh38
rs921373369 Chr.16: 67149122-67149122 on Build GRCh38
rs761777202 Chr.16: 67149078-67149078 on Build GRCh38
rs780539408 Chr.16: 67149313-67149334 on Build GRCh38
rs540950512 Chr.16: 67149170-67149170 on Build GRCh38
rs565145374 Chr.16: 67149058-67149058 on Build GRCh38
rs188607326 Chr.16: 67149010-67149010 on Build GRCh38
rs753590870 Chr.16: 67149247-67149247 on Build GRCh38
rs753678246 Chr.16: 67149327-67149327 on Build GRCh38
rs757863139 Chr.16: 67149340-67149340 on Build GRCh38
rs747647685 Chr.16: 67149251-67149251 on Build GRCh38
rs777251168 Chr.16: 67149314-67149314 on Build GRCh38
rs749735978 Chr.16: 67149385-67149385 on Build GRCh38
rs769037531 Chr.16: 67149390-67149390 on Build GRCh38
rs765188639 Chr.16: 67149334-67149334 on Build GRCh38
rs773379094 Chr.16: 67149405-67149405 on Build GRCh38
rs1002462735 Chr.16: 67148980-67148980 on Build GRCh38
rs534531397 Chr.16: 67149022-67149022 on Build GRCh38
rs753625831 Chr.16: 67149238-67149238 on Build GRCh38
rs774547745 Chr.16: 67149401-67149401 on Build GRCh38
rs751031186 Chr.16: 67149344-67149344 on Build GRCh38
rs375361015 Chr.16: 67149404-67149404 on Build GRCh38
rs976711089 Chr.16: 67149118-67149118 on Build GRCh38
rs565692012 Chr.16: 67149348-67149348 on Build GRCh38
rs932729349 Chr.16: 67149161-67149161 on Build GRCh38
rs946486824 Chr.16: 67149316-67149316 on Build GRCh38
rs762412976 Chr.16: 67149059-67149060 on Build GRCh38
rs757882253 Chr.16: 67149269-67149269 on Build GRCh38
rs746221761 Chr.16: 67149282-67149282 on Build GRCh38
rs891058824 Chr.16: 67149009-67149009 on Build GRCh38
rs764039776 Chr.16: 67149036-67149036 on Build GRCh38
rs780970155 Chr.16: 67149363-67149363 on Build GRCh38
rs34729812 Chr.16: 67149172-67149173 on Build GRCh38
rs768722404 Chr.16: 67149291-67149291 on Build GRCh38
rs749852807 Chr.16: 67149131-67149131 on Build GRCh38
rs765914160 Chr.16: 67149242-67149242 on Build GRCh38
rs766093458 Chr.16: 67149231-67149231 on Build GRCh38
rs934821769 Chr.16: 67149276-67149276 on Build GRCh38
rs755698077 Chr.16: 67149146-67149146 on Build GRCh38
rs777206179 Chr.16: 67149278-67149278 on Build GRCh38
rs746618352 Chr.16: 67149271-67149271 on Build GRCh38
rs200117165 Chr.16: 67149303-67149303 on Build GRCh38
rs569777979 Chr.16: 67149290-67149290 on Build GRCh38
rs549700395 Chr.16: 67149277-67149277 on Build GRCh38
rs752628911 Chr.16: 67149336-67149336 on Build GRCh38
rs758746608 Chr.16: 67149249-67149249 on Build GRCh38
rs780449270 Chr.16: 67149383-67149383 on Build GRCh38
rs956300968 Chr.16: 67149280-67149280 on Build GRCh38
rs146150540 Chr.16: 67148984-67148984 on Build GRCh38
rs928849118 Chr.16: 67149210-67149210 on Build GRCh38
rs767713025 Chr.16: 67149323-67149323 on Build GRCh38
rs376815785 Chr.16: 67149099-67149099 on Build GRCh38
rs560829065 Chr.16: 67149175-67149175 on Build GRCh38
rs1004151346 Chr.16: 67149043-67149043 on Build GRCh38
rs774439018 Chr.16: 67149302-67149302 on Build GRCh38
rs776111111 Chr.16: 67149286-67149286 on Build GRCh38
rs1027234731 Chr.16: 67149403-67149403 on Build GRCh38
rs775823959 Chr.16: 67148999-67148999 on Build GRCh38

View Full Product Details