Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 72008639-72009139 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DHODH
Entrez Gene ID
1723
Gene Name
dihydroorotate dehydrogenase (quinone)
Gene Aliases
DHOdehase, POADS, URA1
UniGene
Hs.654427
Gene Chromosome Location
Chr.16: 72008744-72027659 on Build GRCh38
Transcripts
NM_001361.4 XM_005255827.3 XM_017022990.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1379664 c.19A>C p.K7Q Chr.16: 72008783-72008783 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs778232899 Chr.16: 72008765-72008765 on Build GRCh38
rs889914979 Chr.16: 72009105-72009105 on Build GRCh38
rs913635582 Chr.16: 72008795-72008795 on Build GRCh38
rs535429409 Chr.16: 72008939-72008939 on Build GRCh38
rs1002382191 Chr.16: 72008681-72008681 on Build GRCh38
rs941837750 Chr.16: 72008675-72008675 on Build GRCh38
rs375133590 Chr.16: 72008680-72008680 on Build GRCh38
rs562695864 Chr.16: 72008900-72008900 on Build GRCh38
rs756559311 Chr.16: 72008716-72008716 on Build GRCh38
rs766660091 Chr.16: 72008829-72008829 on Build GRCh38
rs913711701 Chr.16: 72008965-72008965 on Build GRCh38
rs767479870 Chr.16: 72008805-72008805 on Build GRCh38
rs919216940 Chr.16: 72008851-72008851 on Build GRCh38
rs534756196 Chr.16: 72008946-72008946 on Build GRCh38
rs757094207 Chr.16: 72008797-72008797 on Build GRCh38
rs376006031 Chr.16: 72008708-72008708 on Build GRCh38
rs918155849 Chr.16: 72009032-72009032 on Build GRCh38
rs900015268 Chr.16: 72008738-72008739 on Build GRCh38
rs3213423 Chr.16: 72008926-72008926 on Build GRCh38
rs549243362 Chr.16: 72008917-72008917 on Build GRCh38
rs113616219 Chr.16: 72009061-72009061 on Build GRCh38
rs775336922 Chr.16: 72008793-72008793 on Build GRCh38
rs750095043 Chr.16: 72008962-72008962 on Build GRCh38
rs929999412 Chr.16: 72008705-72008705 on Build GRCh38
rs771428943 Chr.16: 72008770-72008770 on Build GRCh38
rs966382050 Chr.16: 72008959-72008959 on Build GRCh38
rs550748249 Chr.16: 72008895-72008895 on Build GRCh38
rs531001116 Chr.16: 72008908-72008908 on Build GRCh38
rs949695236 Chr.16: 72009034-72009034 on Build GRCh38
rs1055349939 Chr.16: 72008826-72008826 on Build GRCh38
rs921311352 Chr.16: 72008923-72008923 on Build GRCh38
rs1043318811 Chr.16: 72008746-72008746 on Build GRCh38
rs982828854 Chr.16: 72008884-72008884 on Build GRCh38
rs532854343 Chr.16: 72008756-72008756 on Build GRCh38
rs149512475 Chr.16: 72008885-72008885 on Build GRCh38
rs979111838 Chr.16: 72008982-72008982 on Build GRCh38
rs369013567 Chr.16: 72008709-72008709 on Build GRCh38
rs776384667 Chr.16: 72008788-72008788 on Build GRCh38
rs755565137 Chr.16: 72008754-72008754 on Build GRCh38
rs777542834 Chr.16: 72008777-72008777 on Build GRCh38
rs895502582 Chr.16: 72008996-72008996 on Build GRCh38
rs912556638 Chr.16: 72008914-72008914 on Build GRCh38
rs1055235785 Chr.16: 72009027-72009027 on Build GRCh38
rs760776971 Chr.16: 72008822-72008822 on Build GRCh38
rs1007309050 Chr.16: 72008941-72008941 on Build GRCh38
rs1012135918 Chr.16: 72008739-72008739 on Build GRCh38
rs779550807 Chr.16: 72009017-72009017 on Build GRCh38
rs1022644102 Chr.16: 72008821-72008821 on Build GRCh38
rs944548885 Chr.16: 72008779-72008779 on Build GRCh38
rs571566080 Chr.16: 72008983-72008983 on Build GRCh38
rs746640238 Chr.16: 72008782-72008782 on Build GRCh38
rs540520522 Chr.16: 72008792-72008792 on Build GRCh38
rs560924899 Chr.16: 72008768-72008768 on Build GRCh38
rs191475180 Chr.16: 72009015-72009015 on Build GRCh38
rs925043209 Chr.16: 72009014-72009014 on Build GRCh38
rs937682107 Chr.16: 72008818-72008818 on Build GRCh38
rs758054723 Chr.16: 72008812-72008812 on Build GRCh38
rs754287528 Chr.16: 72008835-72008835 on Build GRCh38
rs369533480 Chr.16: 72008682-72008682 on Build GRCh38
rs3213422 Chr.16: 72008783-72008783 on Build GRCh38
rs371063686 Chr.16: 72008857-72008857 on Build GRCh38
rs113000464 Chr.16: 72009096-72009096 on Build GRCh38
rs896614647 Chr.16: 72008683-72008683 on Build GRCh38
rs367906932 Chr.16: 72008735-72008735 on Build GRCh38
rs1043469068 Chr.16: 72008809-72008809 on Build GRCh38
rs138009379 Chr.16: 72008713-72008713 on Build GRCh38
rs1037629018 Chr.16: 72008871-72008871 on Build GRCh38
rs945160096 Chr.16: 72008801-72008801 on Build GRCh38
rs995676486 Chr.16: 72009070-72009070 on Build GRCh38
rs147209921 Chr.16: 72009069-72009069 on Build GRCh38
rs558067019 Chr.16: 72009097-72009097 on Build GRCh38
rs761964393 Chr.16: 72008794-72008794 on Build GRCh38
rs376699089 Chr.16: 72008836-72008836 on Build GRCh38
rs966534304 Chr.16: 72008974-72008974 on Build GRCh38
rs750723546 Chr.16: 72008813-72008813 on Build GRCh38
rs878961369 Chr.16: 72008661-72008661 on Build GRCh38
rs542376596 Chr.16: 72008756-72008756 on Build GRCh38
rs546880141 Chr.16: 72008967-72008967 on Build GRCh38
rs569571301 Chr.16: 72009113-72009113 on Build GRCh38
rs34270657 Chr.16: 72008736-72008736 on Build GRCh38
rs903400168 Chr.16: 72008747-72008747 on Build GRCh38
rs995560290 Chr.16: 72008920-72008920 on Build GRCh38

View Full Product Details