Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 72014569-72015023 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DHODH
Entrez Gene ID
1723
Gene Name
dihydroorotate dehydrogenase (quinone)
Gene Aliases
DHOdehase, POADS, URA1
UniGene
Hs.654427
Gene Chromosome Location
Chr.16: 72008744-72027659 on Build GRCh38
Transcripts
NM_001361.4 XM_005255827.3 XM_005255829.3 XM_017022990.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4167771 c.380G>T p.G127V Chr.16: 72014618-72014618 on Build GRCh38
6396633 c.392C>A p.P131H Chr.16: 72014630-72014630 on Build GRCh38
1379666 c.404G>A p.R135H Chr.16: 72014642-72014642 on Build GRCh38
4262208 c.348G>T p.E116D Chr.16: 72014586-72014586 on Build GRCh38
6043003 c.371C>A p.P124H Chr.16: 72014609-72014609 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752149320 Chr.16: 72014896-72014896 on Build GRCh38
rs959032130 Chr.16: 72014736-72014736 on Build GRCh38
rs972950508 Chr.16: 72014778-72014778 on Build GRCh38
rs755151957 Chr.16: 72014707-72014707 on Build GRCh38
rs187230970 Chr.16: 72014735-72014735 on Build GRCh38
rs112904410 Chr.16: 72014697-72014697 on Build GRCh38
rs757198772 Chr.16: 72014616-72014616 on Build GRCh38
rs931288273 Chr.16: 72014595-72014595 on Build GRCh38
rs534111439 Chr.16: 72014925-72014925 on Build GRCh38
rs1047711689 Chr.16: 72014786-72014786 on Build GRCh38
rs758638400 Chr.16: 72014716-72014716 on Build GRCh38
rs1038935029 Chr.16: 72014683-72014683 on Build GRCh38
rs901790915 Chr.16: 72014882-72014882 on Build GRCh38
rs748362867 Chr.16: 72014634-72014634 on Build GRCh38
rs918654786 Chr.16: 72014781-72014781 on Build GRCh38
rs535006080 Chr.16: 72014754-72014754 on Build GRCh38
rs200251747 Chr.16: 72014682-72014682 on Build GRCh38
rs570031927 Chr.16: 72014700-72014700 on Build GRCh38
rs573654196 Chr.16: 72014834-72014834 on Build GRCh38
rs749711209 Chr.16: 72014828-72014828 on Build GRCh38
rs530854110 Chr.16: 72014729-72014729 on Build GRCh38
rs765268364 Chr.16: 72014710-72014710 on Build GRCh38
rs747401750 Chr.16: 72014724-72014724 on Build GRCh38
rs553114904 Chr.16: 72014761-72014761 on Build GRCh38
rs190857490 Chr.16: 72014856-72014856 on Build GRCh38
rs139757496 Chr.16: 72014732-72014732 on Build GRCh38
rs1000584082 Chr.16: 72014913-72014913 on Build GRCh38
rs973409307 Chr.16: 72014955-72014955 on Build GRCh38
rs571539092 Chr.16: 72014763-72014763 on Build GRCh38
rs911190759 Chr.16: 72014823-72014823 on Build GRCh38
rs745400739 Chr.16: 72014687-72014687 on Build GRCh38
rs767309772 Chr.16: 72014617-72014617 on Build GRCh38
rs750314421 Chr.16: 72014701-72014701 on Build GRCh38
rs1007391643 Chr.16: 72014665-72014665 on Build GRCh38
rs894806988 Chr.16: 72014918-72014918 on Build GRCh38
rs745880870 Chr.16: 72014691-72014691 on Build GRCh38
rs886354614 Chr.16: 72014807-72014807 on Build GRCh38
rs180738282 Chr.16: 72014696-72014696 on Build GRCh38
rs1021103679 Chr.16: 72014725-72014725 on Build GRCh38
rs970997966 Chr.16: 72014942-72014942 on Build GRCh38
rs779164186 Chr.16: 72014627-72014627 on Build GRCh38
rs201230446 Chr.16: 72014641-72014641 on Build GRCh38
rs1011960739 Chr.16: 72014924-72014924 on Build GRCh38
rs948510977 Chr.16: 72014740-72014740 on Build GRCh38
rs753983639 Chr.16: 72014619-72014619 on Build GRCh38
rs771495304 Chr.16: 72014681-72014681 on Build GRCh38
rs747285736 Chr.16: 72014671-72014671 on Build GRCh38
rs1004647127 Chr.16: 72014820-72014821 on Build GRCh38
rs754954525 Chr.16: 72014625-72014625 on Build GRCh38
rs931385878 Chr.16: 72014815-72014815 on Build GRCh38
rs573855645 Chr.16: 72014914-72014914 on Build GRCh38
rs902360113 Chr.16: 72014906-72014906 on Build GRCh38
rs762666197 Chr.16: 72014693-72014693 on Build GRCh38
rs916939676 Chr.16: 72014974-72014975 on Build GRCh38
rs996962377 Chr.16: 72014846-72014846 on Build GRCh38
rs777928749 Chr.16: 72014723-72014723 on Build GRCh38
rs889797882 Chr.16: 72014642-72014642 on Build GRCh38
rs752964576 Chr.16: 72014713-72014713 on Build GRCh38
rs542876018 Chr.16: 72014928-72014928 on Build GRCh38
rs183365066 Chr.16: 72014864-72014864 on Build GRCh38
rs1025787598 Chr.16: 72014979-72014979 on Build GRCh38
rs763084831 Chr.16: 72014723-72014724 on Build GRCh38
rs8062895 Chr.16: 72014733-72014733 on Build GRCh38
rs769420197 Chr.16: 72014689-72014689 on Build GRCh38
rs778086368 Chr.16: 72014652-72014652 on Build GRCh38
rs993459653 Chr.16: 72014966-72014966 on Build GRCh38
rs984213848 Chr.16: 72014996-72014996 on Build GRCh38
rs953000976 Chr.16: 72014995-72014995 on Build GRCh38
rs1042192520 Chr.16: 72014901-72014901 on Build GRCh38
rs994639313 Chr.16: 72014783-72014783 on Build GRCh38
rs538704510 Chr.16: 72014821-72014821 on Build GRCh38
rs948349338 Chr.16: 72014989-72014989 on Build GRCh38
rs997290309 Chr.16: 72014886-72014886 on Build GRCh38
rs376315110 Chr.16: 72014748-72014748 on Build GRCh38

View Full Product Details