Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 72096732-72097230 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DHX38
Entrez Gene ID
9785
Gene Name
DEAH-box helicase 38
Gene Aliases
DDX38, PRP16, PRPF16
UniGene
Hs.151218
Gene Chromosome Location
Chr.16: 72093665-72112912 on Build GRCh38
Transcripts
NM_014003.3 XM_005256269.1 XM_011523484.1 XM_011523485.1 XM_017023913.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6643645 c.376G>A p.E126K Chr.16: 72096874-72096874 on Build GRCh38
6145079 c.420G>T p.R140R Chr.16: 72096918-72096918 on Build GRCh38
7170403 c.331C>T p.R111W Chr.16: 72096829-72096829 on Build GRCh38
6643643 c.362C>T p.P121L Chr.16: 72096860-72096860 on Build GRCh38
3276486 c.441G>A p.S147S Chr.16: 72096939-72096939 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375181581 Chr.16: 72096839-72096839 on Build GRCh38
rs1000862160 Chr.16: 72097098-72097098 on Build GRCh38
rs768452875 Chr.16: 72096793-72096793 on Build GRCh38
rs771239607 Chr.16: 72096808-72096808 on Build GRCh38
rs565114277 Chr.16: 72096785-72096785 on Build GRCh38
rs866854517 Chr.16: 72096993-72096993 on Build GRCh38
rs749000951 Chr.16: 72096860-72096860 on Build GRCh38
rs146872571 Chr.16: 72096963-72096963 on Build GRCh38
rs143851929 Chr.16: 72096927-72096927 on Build GRCh38
rs192983504 Chr.16: 72096836-72096836 on Build GRCh38
rs1024388037 Chr.16: 72097097-72097097 on Build GRCh38
rs142444234 Chr.16: 72096948-72096948 on Build GRCh38
rs969795761 Chr.16: 72097186-72097186 on Build GRCh38
rs779959677 Chr.16: 72096788-72096788 on Build GRCh38
rs370388859 Chr.16: 72096838-72096838 on Build GRCh38
rs184663344 Chr.16: 72097157-72097157 on Build GRCh38
rs760440239 Chr.16: 72096908-72096908 on Build GRCh38
rs551943754 Chr.16: 72096867-72096867 on Build GRCh38
rs749498330 Chr.16: 72097025-72097025 on Build GRCh38
rs542179450 Chr.16: 72097084-72097084 on Build GRCh38
rs751140795 Chr.16: 72096837-72096837 on Build GRCh38
rs147413193 Chr.16: 72096861-72096861 on Build GRCh38
rs200501313 Chr.16: 72096907-72096907 on Build GRCh38
rs775810927 Chr.16: 72096835-72096835 on Build GRCh38
rs772671521 Chr.16: 72096975-72096975 on Build GRCh38
rs773376795 Chr.16: 72096794-72096794 on Build GRCh38
rs768904320 Chr.16: 72097033-72097033 on Build GRCh38
rs148616280 Chr.16: 72096939-72096939 on Build GRCh38
rs745462803 Chr.16: 72096786-72096786 on Build GRCh38
rs751442718 Chr.16: 72097012-72097012 on Build GRCh38
rs201640902 Chr.16: 72096985-72096985 on Build GRCh38
rs759687102 Chr.16: 72096821-72096821 on Build GRCh38
rs774728184 Chr.16: 72096895-72096895 on Build GRCh38
rs563363299 Chr.16: 72097190-72097190 on Build GRCh38
rs749274207 Chr.16: 72096792-72096792 on Build GRCh38
rs962383734 Chr.16: 72097114-72097114 on Build GRCh38
rs76109028 Chr.16: 72096920-72096920 on Build GRCh38
rs746110198 Chr.16: 72096876-72096876 on Build GRCh38
rs748059500 Chr.16: 72096865-72096865 on Build GRCh38
rs749463201 Chr.16: 72096957-72096957 on Build GRCh38
rs772921146 Chr.16: 72096976-72096976 on Build GRCh38
rs139547116 Chr.16: 72096873-72096873 on Build GRCh38
rs775991107 Chr.16: 72096879-72096879 on Build GRCh38
rs772759523 Chr.16: 72097050-72097050 on Build GRCh38
rs145090795 Chr.16: 72096921-72096921 on Build GRCh38
rs758983126 Chr.16: 72097156-72097156 on Build GRCh38
rs769008594 Chr.16: 72096890-72096890 on Build GRCh38
rs1024050282 Chr.16: 72096952-72096952 on Build GRCh38
rs368122255 Chr.16: 72096849-72096849 on Build GRCh38
rs780039631 Chr.16: 72096859-72096859 on Build GRCh38
rs371468268 Chr.16: 72097091-72097091 on Build GRCh38
rs776376075 Chr.16: 72096981-72096981 on Build GRCh38
rs546446584 Chr.16: 72096769-72096769 on Build GRCh38
rs528398566 Chr.16: 72096767-72096767 on Build GRCh38
rs778702496 Chr.16: 72096917-72096917 on Build GRCh38
rs972042535 Chr.16: 72097116-72097116 on Build GRCh38
rs752514487 Chr.16: 72096994-72096994 on Build GRCh38
rs189547316 Chr.16: 72097158-72097158 on Build GRCh38
rs368707127 Chr.16: 72096974-72096974 on Build GRCh38
rs560957594 Chr.16: 72097130-72097130 on Build GRCh38
rs776849536 Chr.16: 72096818-72096818 on Build GRCh38
rs549408121 Chr.16: 72096913-72096913 on Build GRCh38
rs763215842 Chr.16: 72096889-72096889 on Build GRCh38
rs750135709 Chr.16: 72096841-72096841 on Build GRCh38
rs992526274 Chr.16: 72096951-72096951 on Build GRCh38
rs746171802 Chr.16: 72096897-72096897 on Build GRCh38
rs765927258 Chr.16: 72096982-72096982 on Build GRCh38
rs114680015 Chr.16: 72097021-72097021 on Build GRCh38
rs972681537 Chr.16: 72096938-72096938 on Build GRCh38
rs780155892 Chr.16: 72096949-72096949 on Build GRCh38
rs911007147 Chr.16: 72097191-72097191 on Build GRCh38
rs769973036 Chr.16: 72096877-72096877 on Build GRCh38
rs367750088 Chr.16: 72096787-72096787 on Build GRCh38
rs777674693 Chr.16: 72096996-72096996 on Build GRCh38
rs778699985 Chr.16: 72096864-72096864 on Build GRCh38
rs762581336 Chr.16: 72097094-72097094 on Build GRCh38
rs375554088 Chr.16: 72096969-72096969 on Build GRCh38
rs372428319 Chr.16: 72096858-72096858 on Build GRCh38
rs753722533 Chr.16: 72096914-72096914 on Build GRCh38
rs147170871 Chr.16: 72097020-72097020 on Build GRCh38
rs763916361 Chr.16: 72096813-72096814 on Build GRCh38
rs766168832 Chr.16: 72097096-72097096 on Build GRCh38
rs779189314 Chr.16: 72097042-72097042 on Build GRCh38
rs760434453 Chr.16: 72096790-72096790 on Build GRCh38
rs765620584 Chr.16: 72096833-72096833 on Build GRCh38
rs762344139 Chr.16: 72096896-72096896 on Build GRCh38
rs757003948 Chr.16: 72096774-72096774 on Build GRCh38
rs1020522001 Chr.16: 72096995-72096995 on Build GRCh38
rs887168085 Chr.16: 72096758-72096758 on Build GRCh38
rs747052931 Chr.16: 72096800-72096800 on Build GRCh38
rs1050362 Chr.16: 72096916-72096916 on Build GRCh38
rs764963782 Chr.16: 72096989-72096989 on Build GRCh38
rs748514865 Chr.16: 72097049-72097049 on Build GRCh38
rs1023997313 Chr.16: 72097163-72097163 on Build GRCh38

View Full Product Details