Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 74450403-74450914 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLG1
Entrez Gene ID
2734
Gene Name
golgi glycoprotein 1
Gene Aliases
CFR-1, ESL-1, MG-160, MG160
UniGene
Hs.109731
Gene Chromosome Location
Chr.16: 74447427-74607144 on Build GRCh38
Transcripts
NM_001145666.1 NM_001145667.1 NM_012201.5 NR_027264.1 NR_027265.1

SNP Detail

SNP ID Chromosome Location
rs568988599 Chr.16: 74450473-74450474 on Build GRCh38
rs559724697 Chr.16: 74450593-74450593 on Build GRCh38
rs368525069 Chr.16: 74450844-74450844 on Build GRCh38
rs1030611058 Chr.16: 74450721-74450721 on Build GRCh38
rs534290582 Chr.16: 74450841-74450841 on Build GRCh38
rs1045607321 Chr.16: 74450827-74450827 on Build GRCh38
rs567466187 Chr.16: 74450839-74450839 on Build GRCh38
rs945387196 Chr.16: 74450509-74450509 on Build GRCh38
rs958363519 Chr.16: 74450803-74450803 on Build GRCh38
rs181327611 Chr.16: 74450667-74450667 on Build GRCh38
rs963462254 Chr.16: 74450651-74450651 on Build GRCh38
rs958592460 Chr.16: 74450634-74450634 on Build GRCh38
rs776276305 Chr.16: 74450709-74450709 on Build GRCh38
rs542939482 Chr.16: 74450575-74450575 on Build GRCh38
rs577767479 Chr.16: 74450883-74450883 on Build GRCh38
rs536597995 Chr.16: 74450778-74450778 on Build GRCh38
rs928576077 Chr.16: 74450856-74450856 on Build GRCh38
rs1054364117 Chr.16: 74450785-74450785 on Build GRCh38
rs1002808849 Chr.16: 74450596-74450596 on Build GRCh38
rs1051990969 Chr.16: 74450695-74450695 on Build GRCh38
rs529245510 Chr.16: 74450508-74450508 on Build GRCh38
rs1009667729 Chr.16: 74450788-74450788 on Build GRCh38
rs559083092 Chr.16: 74450499-74450499 on Build GRCh38
rs997753846 Chr.16: 74450718-74450718 on Build GRCh38
rs894351700 Chr.16: 74450640-74450640 on Build GRCh38
rs557857933 Chr.16: 74450869-74450869 on Build GRCh38
rs547596951 Chr.16: 74450847-74450848 on Build GRCh38
rs564781748 Chr.16: 74450666-74450666 on Build GRCh38
rs571240924 Chr.16: 74450848-74450851 on Build GRCh38
rs189731819 Chr.16: 74450457-74450457 on Build GRCh38
rs1009615132 Chr.16: 74450549-74450549 on Build GRCh38
rs549203194 Chr.16: 74450620-74450620 on Build GRCh38
rs185921763 Chr.16: 74450804-74450804 on Build GRCh38
rs375555132 Chr.16: 74450761-74450761 on Build GRCh38
rs1012799204 Chr.16: 74450829-74450829 on Build GRCh38
rs915720374 Chr.16: 74450805-74450805 on Build GRCh38
rs551867096 Chr.16: 74450623-74450623 on Build GRCh38
rs935817594 Chr.16: 74450622-74450622 on Build GRCh38
rs1017826473 Chr.16: 74450432-74450432 on Build GRCh38
rs893939444 Chr.16: 74450786-74450786 on Build GRCh38
rs761874592 Chr.16: 74450769-74450769 on Build GRCh38
rs1035640666 Chr.16: 74450809-74450809 on Build GRCh38
rs894378463 Chr.16: 74450822-74450822 on Build GRCh38
rs908999709 Chr.16: 74450641-74450641 on Build GRCh38
rs948635806 Chr.16: 74450646-74450646 on Build GRCh38
rs965340157 Chr.16: 74450434-74450434 on Build GRCh38
rs948585358 Chr.16: 74450808-74450808 on Build GRCh38
rs1003529177 Chr.16: 74450807-74450807 on Build GRCh38
rs1020974031 Chr.16: 74450560-74450560 on Build GRCh38
rs964916099 Chr.16: 74450713-74450713 on Build GRCh38
rs371221555 Chr.16: 74450459-74450459 on Build GRCh38
rs145549473 Chr.16: 74450686-74450686 on Build GRCh38
rs887537396 Chr.16: 74450676-74450676 on Build GRCh38
rs112328819 Chr.16: 74450864-74450864 on Build GRCh38
rs1053905509 Chr.16: 74450507-74450507 on Build GRCh38
rs1026042830 Chr.16: 74450668-74450668 on Build GRCh38
rs1051938263 Chr.16: 74450458-74450458 on Build GRCh38
rs563577456 Chr.16: 74450460-74450460 on Build GRCh38
rs925355138 Chr.16: 74450798-74450798 on Build GRCh38
rs925377356 Chr.16: 74450621-74450621 on Build GRCh38
rs578212774 Chr.16: 74450708-74450708 on Build GRCh38
rs978100492 Chr.16: 74450789-74450789 on Build GRCh38
rs1034660645 Chr.16: 74450625-74450625 on Build GRCh38
rs1026002558 Chr.16: 74450862-74450862 on Build GRCh38
rs943585416 Chr.16: 74450770-74450770 on Build GRCh38
rs976042875 Chr.16: 74450467-74450467 on Build GRCh38
rs910782938 Chr.16: 74450748-74450748 on Build GRCh38
rs867622854 Chr.16: 74450843-74450843 on Build GRCh38
rs571619563 Chr.16: 74450654-74450654 on Build GRCh38
rs746554155 Chr.16: 74450881-74450881 on Build GRCh38
rs1018193259 Chr.16: 74450710-74450710 on Build GRCh38

View Full Product Details