Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 74718887-74719380 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FA2H
Entrez Gene ID
79152
Gene Name
fatty acid 2-hydroxylase
Gene Aliases
FAAH, FAH1, FAXDC1, SCS7, SPG35
UniGene
Hs.461329
Gene Chromosome Location
Chr.16: 74712955-74774831 on Build GRCh38
Transcripts
NM_024306.4 XM_011523317.2 XM_011523319.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5522623 c.665G>T p.G222V Chr.16: 74719109-74719109 on Build GRCh38
2690964 c.650G>A p.G217E Chr.16: 74719124-74719124 on Build GRCh38
5456567 c.772G>A p.G258S Chr.16: 74719002-74719002 on Build GRCh38
973664 c.640A>G p.M214V Chr.16: 74719134-74719134 on Build GRCh38
3512190 c.727C>T p.P243S Chr.16: 74719047-74719047 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771722151 Chr.16: 74719192-74719192 on Build GRCh38
rs386792123 Chr.16: 74718923-74718924 on Build GRCh38
rs200118098 Chr.16: 74718944-74718944 on Build GRCh38
rs377171975 Chr.16: 74719167-74719167 on Build GRCh38
rs771564922 Chr.16: 74719101-74719101 on Build GRCh38
rs1026526407 Chr.16: 74719152-74719152 on Build GRCh38
rs201968011 Chr.16: 74718949-74718949 on Build GRCh38
rs935889912 Chr.16: 74719317-74719317 on Build GRCh38
rs200203230 Chr.16: 74718967-74718967 on Build GRCh38
rs368285542 Chr.16: 74719347-74719347 on Build GRCh38
rs183814366 Chr.16: 74719171-74719171 on Build GRCh38
rs368669121 Chr.16: 74718981-74718981 on Build GRCh38
rs777744385 Chr.16: 74719338-74719338 on Build GRCh38
rs757119964 Chr.16: 74719065-74719065 on Build GRCh38
rs760000001 Chr.16: 74719034-74719034 on Build GRCh38
rs989170228 Chr.16: 74719198-74719198 on Build GRCh38
rs550285066 Chr.16: 74718972-74718972 on Build GRCh38
rs781156520 Chr.16: 74719174-74719174 on Build GRCh38
rs779856022 Chr.16: 74719153-74719153 on Build GRCh38
rs371293493 Chr.16: 74719003-74719003 on Build GRCh38
rs761217557 Chr.16: 74719020-74719020 on Build GRCh38
rs780906296 Chr.16: 74718990-74718990 on Build GRCh38
rs369504312 Chr.16: 74719105-74719105 on Build GRCh38
rs563816059 Chr.16: 74719199-74719199 on Build GRCh38
rs939232748 Chr.16: 74719292-74719293 on Build GRCh38
rs372699585 Chr.16: 74719341-74719341 on Build GRCh38
rs746782405 Chr.16: 74718939-74718939 on Build GRCh38
rs111306468 Chr.16: 74718923-74718923 on Build GRCh38
rs906411491 Chr.16: 74719075-74719075 on Build GRCh38
rs1054114951 Chr.16: 74719029-74719029 on Build GRCh38
rs774032544 Chr.16: 74719195-74719195 on Build GRCh38
rs776399428 Chr.16: 74718971-74718971 on Build GRCh38
rs770751836 Chr.16: 74718940-74718940 on Build GRCh38
rs797045569 Chr.16: 74719012-74719012 on Build GRCh38
rs761645282 Chr.16: 74718992-74718992 on Build GRCh38
rs751056475 Chr.16: 74719140-74719140 on Build GRCh38
rs767002644 Chr.16: 74719206-74719206 on Build GRCh38
rs201126702 Chr.16: 74719041-74719041 on Build GRCh38
rs371280297 Chr.16: 74718977-74718977 on Build GRCh38
rs771187125 Chr.16: 74719173-74719173 on Build GRCh38
rs878855081 Chr.16: 74719083-74719083 on Build GRCh38
rs770149919 Chr.16: 74719120-74719120 on Build GRCh38
rs774228483 Chr.16: 74718947-74718947 on Build GRCh38
rs772321218 Chr.16: 74719006-74719006 on Build GRCh38
rs377559323 Chr.16: 74718925-74718925 on Build GRCh38
rs952055477 Chr.16: 74718930-74718930 on Build GRCh38
rs778973971 Chr.16: 74719072-74719072 on Build GRCh38
rs955192546 Chr.16: 74719233-74719233 on Build GRCh38
rs2074629 Chr.16: 74719181-74719181 on Build GRCh38
rs768757432 Chr.16: 74719001-74719001 on Build GRCh38
rs749929064 Chr.16: 74719209-74719209 on Build GRCh38
rs756025962 Chr.16: 74719150-74719150 on Build GRCh38
rs767899097 Chr.16: 74718955-74718955 on Build GRCh38
rs760590903 Chr.16: 74719210-74719210 on Build GRCh38
rs371803211 Chr.16: 74719099-74719099 on Build GRCh38
rs146684287 Chr.16: 74719024-74719024 on Build GRCh38
rs114682163 Chr.16: 74719217-74719217 on Build GRCh38
rs761379296 Chr.16: 74719141-74719141 on Build GRCh38
rs775231176 Chr.16: 74718922-74718922 on Build GRCh38
rs387907039 Chr.16: 74719071-74719071 on Build GRCh38
rs770208780 Chr.16: 74719182-74719182 on Build GRCh38
rs6564161 Chr.16: 74718943-74718943 on Build GRCh38
rs1035789947 Chr.16: 74718917-74718917 on Build GRCh38
rs566281289 Chr.16: 74719249-74719249 on Build GRCh38
rs748364155 Chr.16: 74719087-74719087 on Build GRCh38
rs894465216 Chr.16: 74719334-74719334 on Build GRCh38
rs1057169685 Chr.16: 74719309-74719309 on Build GRCh38
rs376180814 Chr.16: 74719102-74719102 on Build GRCh38
rs1054150208 Chr.16: 74719323-74719323 on Build GRCh38
rs775750642 Chr.16: 74719125-74719125 on Build GRCh38
rs752131780 Chr.16: 74718979-74718979 on Build GRCh38
rs372445274 Chr.16: 74719154-74719154 on Build GRCh38
rs746907255 Chr.16: 74718982-74718982 on Build GRCh38
rs200307057 Chr.16: 74718976-74718976 on Build GRCh38
rs776851677 Chr.16: 74719033-74719033 on Build GRCh38
rs768936756 Chr.16: 74718946-74718946 on Build GRCh38
rs387907172 Chr.16: 74719067-74719067 on Build GRCh38
rs767410582 Chr.16: 74719144-74719144 on Build GRCh38
rs993592575 Chr.16: 74719143-74719143 on Build GRCh38
rs76575876 Chr.16: 74719224-74719224 on Build GRCh38
rs201468381 Chr.16: 74718963-74718963 on Build GRCh38
rs773459488 Chr.16: 74719011-74719011 on Build GRCh38
rs772449065 Chr.16: 74719094-74719094 on Build GRCh38
rs759577964 Chr.16: 74719139-74719139 on Build GRCh38
rs758016356 Chr.16: 74719049-74719049 on Build GRCh38
rs555874983 Chr.16: 74718989-74718989 on Build GRCh38
rs924557156 Chr.16: 74719250-74719250 on Build GRCh38
rs191369932 Chr.16: 74718924-74718924 on Build GRCh38
rs367933620 Chr.16: 74719056-74719056 on Build GRCh38
rs768292367 Chr.16: 74719194-74719194 on Build GRCh38
rs781172621 Chr.16: 74719066-74719066 on Build GRCh38
rs541338991 Chr.16: 74719146-74719146 on Build GRCh38
rs572112273 Chr.16: 74719070-74719070 on Build GRCh38
rs774693133 Chr.16: 74719002-74719002 on Build GRCh38
rs375122652 Chr.16: 74718964-74718964 on Build GRCh38
rs923346045 Chr.16: 74719228-74719228 on Build GRCh38
rs763367012 Chr.16: 74719193-74719193 on Build GRCh38
rs757972119 Chr.16: 74718980-74718980 on Build GRCh38
rs142632840 Chr.16: 74719042-74719042 on Build GRCh38
rs971696331 Chr.16: 74718906-74718906 on Build GRCh38
rs763544424 Chr.16: 74719050-74719050 on Build GRCh38

View Full Product Details