Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 75566746-75567237 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GABARAPL2
Entrez Gene ID
11345
Gene Name
GABA type A receptor associated protein like 2
Gene Aliases
ATG8, ATG8C, GATE-16, GATE16, GEF-2, GEF2
UniGene
Hs.461379
Gene Chromosome Location
Chr.16: 75566351-75577881 on Build GRCh38
Transcripts
NM_007285.6

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
311336 c.53C>A p.S18Y Chr.16: 75566870-75566870 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs750311316 Chr.16: 75566885-75566885 on Build GRCh38
rs375741946 Chr.16: 75566844-75566844 on Build GRCh38
rs763670284 Chr.16: 75566921-75566921 on Build GRCh38
rs1041965677 Chr.16: 75567130-75567130 on Build GRCh38
rs772023878 Chr.16: 75566831-75566831 on Build GRCh38
rs1023854478 Chr.16: 75566901-75566901 on Build GRCh38
rs531508958 Chr.16: 75567145-75567145 on Build GRCh38
rs781137565 Chr.16: 75566884-75566884 on Build GRCh38
rs778998506 Chr.16: 75566826-75566826 on Build GRCh38
rs888094292 Chr.16: 75566796-75566796 on Build GRCh38
rs113358738 Chr.16: 75567074-75567074 on Build GRCh38
rs369716750 Chr.16: 75566835-75566835 on Build GRCh38
rs909561950 Chr.16: 75567028-75567028 on Build GRCh38
rs938655371 Chr.16: 75566961-75566961 on Build GRCh38
rs550330104 Chr.16: 75567049-75567049 on Build GRCh38
rs999625754 Chr.16: 75567153-75567153 on Build GRCh38
rs901011346 Chr.16: 75566808-75566808 on Build GRCh38
rs201919472 Chr.16: 75566825-75566825 on Build GRCh38
rs370484660 Chr.16: 75567142-75567142 on Build GRCh38
rs757202296 Chr.16: 75566876-75566876 on Build GRCh38
rs199877777 Chr.16: 75566915-75566915 on Build GRCh38
rs758493167 Chr.16: 75566945-75566945 on Build GRCh38
rs11556294 Chr.16: 75566872-75566872 on Build GRCh38
rs201130277 Chr.16: 75566916-75566916 on Build GRCh38
rs894360505 Chr.16: 75566976-75566976 on Build GRCh38
rs987049789 Chr.16: 75566881-75566881 on Build GRCh38
rs750362776 Chr.16: 75566936-75566936 on Build GRCh38
rs767619879 Chr.16: 75566933-75566933 on Build GRCh38
rs1032392235 Chr.16: 75567163-75567163 on Build GRCh38
rs968870393 Chr.16: 75567149-75567149 on Build GRCh38
rs1041710881 Chr.16: 75566767-75566767 on Build GRCh38
rs929732179 Chr.16: 75566774-75566774 on Build GRCh38
rs987056439 Chr.16: 75567047-75567047 on Build GRCh38
rs757844968 Chr.16: 75566815-75566815 on Build GRCh38
rs770020138 Chr.16: 75566910-75566910 on Build GRCh38
rs763470207 Chr.16: 75566874-75566874 on Build GRCh38
rs200373287 Chr.16: 75566817-75566817 on Build GRCh38
rs773809839 Chr.16: 75566834-75566834 on Build GRCh38
rs754330496 Chr.16: 75566923-75566924 on Build GRCh38
rs975677748 Chr.16: 75567072-75567072 on Build GRCh38
rs370301495 Chr.16: 75566871-75566871 on Build GRCh38
rs752189385 Chr.16: 75566813-75566813 on Build GRCh38
rs780756685 Chr.16: 75567114-75567114 on Build GRCh38
rs898982776 Chr.16: 75567008-75567008 on Build GRCh38
rs533454209 Chr.16: 75567005-75567005 on Build GRCh38
rs748317076 Chr.16: 75566829-75566829 on Build GRCh38
rs771943322 Chr.16: 75566862-75566862 on Build GRCh38
rs771363267 Chr.16: 75566906-75566906 on Build GRCh38
rs748287599 Chr.16: 75566892-75566892 on Build GRCh38
rs765010205 Chr.16: 75566870-75566870 on Build GRCh38
rs991076349 Chr.16: 75566854-75566854 on Build GRCh38
rs373260428 Chr.16: 75566799-75566799 on Build GRCh38
rs933911362 Chr.16: 75567075-75567075 on Build GRCh38
rs144908510 Chr.16: 75566889-75566889 on Build GRCh38
rs550476239 Chr.16: 75566938-75566938 on Build GRCh38
rs751045838 Chr.16: 75566924-75566924 on Build GRCh38
rs766049190 Chr.16: 75566940-75566940 on Build GRCh38
rs8192517 Chr.16: 75566824-75566824 on Build GRCh38
rs8192518 Chr.16: 75566902-75566902 on Build GRCh38
rs780818378 Chr.16: 75566862-75566862 on Build GRCh38
rs569692896 Chr.16: 75566964-75566964 on Build GRCh38
rs1002813714 Chr.16: 75566798-75566798 on Build GRCh38
rs751535250 Chr.16: 75566875-75566875 on Build GRCh38
rs924652034 Chr.16: 75567158-75567158 on Build GRCh38
rs1048075364 Chr.16: 75566791-75566791 on Build GRCh38
rs964613075 Chr.16: 75567059-75567059 on Build GRCh38
rs765865013 Chr.16: 75566946-75566947 on Build GRCh38
rs918371060 Chr.16: 75566773-75566773 on Build GRCh38
rs538606117 Chr.16: 75567013-75567013 on Build GRCh38
rs903170703 Chr.16: 75566785-75566786 on Build GRCh38
rs761401850 Chr.16: 75566928-75566928 on Build GRCh38
rs1016540161 Chr.16: 75567001-75567001 on Build GRCh38
rs562210168 Chr.16: 75566943-75566943 on Build GRCh38
rs977536431 Chr.16: 75567151-75567151 on Build GRCh38
rs751417306 Chr.16: 75566820-75566820 on Build GRCh38
rs954384747 Chr.16: 75567036-75567036 on Build GRCh38
rs11556293 Chr.16: 75566895-75566895 on Build GRCh38
rs762461677 Chr.16: 75566920-75566920 on Build GRCh38
rs747530890 Chr.16: 75566905-75566905 on Build GRCh38
rs777916004 Chr.16: 75566946-75566946 on Build GRCh38
rs777123029 Chr.16: 75566908-75566908 on Build GRCh38
rs759196152 Chr.16: 75566848-75566848 on Build GRCh38
rs772226891 Chr.16: 75566894-75566894 on Build GRCh38
rs1026014465 Chr.16: 75567017-75567017 on Build GRCh38
rs747716058 Chr.16: 75566955-75566955 on Build GRCh38
rs545541607 Chr.16: 75566821-75566821 on Build GRCh38
rs533523125 Chr.16: 75566788-75566788 on Build GRCh38
rs764743547 Chr.16: 75566811-75566811 on Build GRCh38
rs754512842 Chr.16: 75567058-75567058 on Build GRCh38
rs562648594 Chr.16: 75567105-75567105 on Build GRCh38
rs150359361 Chr.16: 75566951-75566951 on Build GRCh38
rs756014362 Chr.16: 75566937-75566937 on Build GRCh38
rs544646907 Chr.16: 75567032-75567032 on Build GRCh38
rs377417014 Chr.16: 75567000-75567000 on Build GRCh38
rs374021918 Chr.16: 75566887-75566887 on Build GRCh38
rs182503917 Chr.16: 75566979-75566979 on Build GRCh38
rs754578979 Chr.16: 75566804-75566804 on Build GRCh38
rs751101148 Chr.16: 75567010-75567010 on Build GRCh38
rs982719745 Chr.16: 75566960-75566960 on Build GRCh38

View Full Product Details