Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 81365247-81365745 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GAN
Entrez Gene ID
8139
Gene Name
gigaxonin
Gene Aliases
GAN1, KLHL16
UniGene
Hs.112569
Gene Chromosome Location
Chr.16: 81314941-81380198 on Build GRCh38
Transcripts
NM_022041.3 XM_017023734.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4063103 c.1386G>A p.V462V Chr.16: 81365362-81365362 on Build GRCh38
4063104 c.1421G>T p.G474V Chr.16: 81365397-81365397 on Build GRCh38
3691197 c.1374-1G>T p.? Chr.16: 81365349-81365349 on Build GRCh38
6669211 c.1453A>G p.S485G Chr.16: 81365429-81365429 on Build GRCh38
4063105 c.1449G>A p.Q483Q Chr.16: 81365425-81365425 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs539966784 Chr.16: 81365564-81365564 on Build GRCh38
rs1004198770 Chr.16: 81365664-81365664 on Build GRCh38
rs375587395 Chr.16: 81365715-81365715 on Build GRCh38
rs1007418148 Chr.16: 81365313-81365313 on Build GRCh38
rs1005694749 Chr.16: 81365290-81365290 on Build GRCh38
rs1056703500 Chr.16: 81365571-81365571 on Build GRCh38
rs200342055 Chr.16: 81365364-81365364 on Build GRCh38
rs979135867 Chr.16: 81365709-81365709 on Build GRCh38
rs901720169 Chr.16: 81365598-81365598 on Build GRCh38
rs763752873 Chr.16: 81365434-81365434 on Build GRCh38
rs748724208 Chr.16: 81365482-81365482 on Build GRCh38
rs374707169 Chr.16: 81365305-81365305 on Build GRCh38
rs981306914 Chr.16: 81365687-81365687 on Build GRCh38
rs2615701 Chr.16: 81365700-81365700 on Build GRCh38
rs968154302 Chr.16: 81365638-81365638 on Build GRCh38
rs779309535 Chr.16: 81365529-81365529 on Build GRCh38
rs763621343 Chr.16: 81365277-81365277 on Build GRCh38
rs570149197 Chr.16: 81365676-81365676 on Build GRCh38
rs150843370 Chr.16: 81365691-81365691 on Build GRCh38
rs756485979 Chr.16: 81365374-81365374 on Build GRCh38
rs748209210 Chr.16: 81365406-81365406 on Build GRCh38
rs375408288 Chr.16: 81365378-81365378 on Build GRCh38
rs373999262 Chr.16: 81365518-81365518 on Build GRCh38
rs888624884 Chr.16: 81365289-81365289 on Build GRCh38
rs1049302531 Chr.16: 81365506-81365506 on Build GRCh38
rs774299994 Chr.16: 81365331-81365331 on Build GRCh38
rs369693274 Chr.16: 81365420-81365420 on Build GRCh38
rs746724274 Chr.16: 81365372-81365372 on Build GRCh38
rs777535272 Chr.16: 81365367-81365367 on Build GRCh38
rs755423556 Chr.16: 81365470-81365470 on Build GRCh38
rs760434877 Chr.16: 81365347-81365347 on Build GRCh38
rs1007778141 Chr.16: 81365580-81365580 on Build GRCh38
rs119485094 Chr.16: 81365405-81365405 on Build GRCh38
rs1033858849 Chr.16: 81365689-81365689 on Build GRCh38
rs750919773 Chr.16: 81365412-81365412 on Build GRCh38
rs951434096 Chr.16: 81365414-81365414 on Build GRCh38
rs763429596 Chr.16: 81365328-81365328 on Build GRCh38
rs372398691 Chr.16: 81365566-81365566 on Build GRCh38
rs376869954 Chr.16: 81365315-81365315 on Build GRCh38
rs763632331 Chr.16: 81365516-81365516 on Build GRCh38
rs769676930 Chr.16: 81365549-81365549 on Build GRCh38
rs780023453 Chr.16: 81365304-81365304 on Build GRCh38
rs147725941 Chr.16: 81365359-81365359 on Build GRCh38
rs376616910 Chr.16: 81365513-81365513 on Build GRCh38
rs997396817 Chr.16: 81365599-81365599 on Build GRCh38
rs771877883 Chr.16: 81365316-81365316 on Build GRCh38
rs564079455 Chr.16: 81365534-81365534 on Build GRCh38
rs756866064 Chr.16: 81365452-81365452 on Build GRCh38
rs938969930 Chr.16: 81365288-81365288 on Build GRCh38
rs1038886615 Chr.16: 81365597-81365597 on Build GRCh38
rs767056278 Chr.16: 81365519-81365519 on Build GRCh38
rs200677251 Chr.16: 81365435-81365435 on Build GRCh38
rs962844978 Chr.16: 81365681-81365681 on Build GRCh38
rs750134156 Chr.16: 81365521-81365521 on Build GRCh38
rs921827074 Chr.16: 81365682-81365682 on Build GRCh38
rs199515586 Chr.16: 81365475-81365475 on Build GRCh38
rs1043850694 Chr.16: 81365557-81365557 on Build GRCh38
rs375347652 Chr.16: 81365330-81365330 on Build GRCh38
rs753663291 Chr.16: 81365487-81365488 on Build GRCh38
rs183926788 Chr.16: 81365271-81365271 on Build GRCh38
rs369724483 Chr.16: 81365334-81365334 on Build GRCh38
rs1037194311 Chr.16: 81365538-81365538 on Build GRCh38
rs773282330 Chr.16: 81365487-81365487 on Build GRCh38
rs771077333 Chr.16: 81365497-81365497 on Build GRCh38
rs745759658 Chr.16: 81365537-81365537 on Build GRCh38
rs959319084 Chr.16: 81365341-81365341 on Build GRCh38
rs887513837 Chr.16: 81365508-81365508 on Build GRCh38
rs199870222 Chr.16: 81365360-81365360 on Build GRCh38
rs372011670 Chr.16: 81365322-81365322 on Build GRCh38
rs746211490 Chr.16: 81365323-81365323 on Build GRCh38
rs372112530 Chr.16: 81365340-81365340 on Build GRCh38
rs945013814 Chr.16: 81365701-81365701 on Build GRCh38
rs780427229 Chr.16: 81365453-81365453 on Build GRCh38
rs776058780 Chr.16: 81365326-81365326 on Build GRCh38
rs931636191 Chr.16: 81365499-81365499 on Build GRCh38
rs372771036 Chr.16: 81365302-81365302 on Build GRCh38
rs119485088 Chr.16: 81365432-81365432 on Build GRCh38
rs895157388 Chr.16: 81365572-81365572 on Build GRCh38
rs541676751 Chr.16: 81365284-81365284 on Build GRCh38
rs903684371 Chr.16: 81365306-81365306 on Build GRCh38
rs546830463 Chr.16: 81365607-81365607 on Build GRCh38
rs965818762 Chr.16: 81365671-81365671 on Build GRCh38
rs749638084 Chr.16: 81365455-81365455 on Build GRCh38
rs148340261 Chr.16: 81365625-81365625 on Build GRCh38
rs766155831 Chr.16: 81365357-81365357 on Build GRCh38
rs769697301 Chr.16: 81365511-81365511 on Build GRCh38
rs550833523 Chr.16: 81365647-81365647 on Build GRCh38
rs776957244 Chr.16: 81365502-81365502 on Build GRCh38
rs542067662 Chr.16: 81365426-81365426 on Build GRCh38
rs772129535 Chr.16: 81365485-81365485 on Build GRCh38
rs998119133 Chr.16: 81365392-81365392 on Build GRCh38
rs756812739 Chr.16: 81365440-81365440 on Build GRCh38
rs774365963 Chr.16: 81365404-81365404 on Build GRCh38
rs781255806 Chr.16: 81365505-81365505 on Build GRCh38
rs752274190 Chr.16: 81365363-81365363 on Build GRCh38
rs770320474 Chr.16: 81365325-81365325 on Build GRCh38
rs141523201 Chr.16: 81365640-81365640 on Build GRCh38
rs119485089 Chr.16: 81365423-81365423 on Build GRCh38
rs534711983 Chr.16: 81365717-81365717 on Build GRCh38
rs769076379 Chr.16: 81365388-81365388 on Build GRCh38
rs551737207 Chr.16: 81365524-81365524 on Build GRCh38
rs761982534 Chr.16: 81365332-81365332 on Build GRCh38
rs1056158914 Chr.16: 81365291-81365291 on Build GRCh38
rs753748994 Chr.16: 81365358-81365358 on Build GRCh38
rs897450269 Chr.16: 81365550-81365550 on Build GRCh38
rs143413333 Chr.16: 81365489-81365489 on Build GRCh38
rs762996927 Chr.16: 81365515-81365515 on Build GRCh38
rs766129455 Chr.16: 81365422-81365422 on Build GRCh38
rs188203725 Chr.16: 81365693-81365693 on Build GRCh38
rs35201963 Chr.16: 81365715-81365715 on Build GRCh38
rs75378898 Chr.16: 81365719-81365719 on Build GRCh38
rs146576740 Chr.16: 81365421-81365421 on Build GRCh38

View Full Product Details