Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 84566120-84566627 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COTL1
Entrez Gene ID
23406
Gene Name
coactosin like F-actin binding protein 1
Gene Aliases
CLP
UniGene
Hs.289092
Gene Chromosome Location
Chr.16: 84565596-84618096 on Build GRCh38
Transcripts
NM_021149.3

SNP Detail

SNP ID Chromosome Location
rs1017731392 Chr.16: 84566319-84566319 on Build GRCh38
rs58297692 Chr.16: 84566183-84566183 on Build GRCh38
rs921165377 Chr.16: 84566461-84566461 on Build GRCh38
rs1024643293 Chr.16: 84566568-84566568 on Build GRCh38
rs528828261 Chr.16: 84566348-84566348 on Build GRCh38
rs247862 Chr.16: 84566260-84566260 on Build GRCh38
rs532159444 Chr.16: 84566295-84566295 on Build GRCh38
rs973334708 Chr.16: 84566443-84566443 on Build GRCh38
rs188279251 Chr.16: 84566507-84566507 on Build GRCh38
rs987809245 Chr.16: 84566359-84566359 on Build GRCh38
rs1034540671 Chr.16: 84566558-84566558 on Build GRCh38
rs767394231 Chr.16: 84566219-84566219 on Build GRCh38
rs540879859 Chr.16: 84566241-84566241 on Build GRCh38
rs13521 Chr.16: 84566364-84566364 on Build GRCh38
rs1051769002 Chr.16: 84566453-84566453 on Build GRCh38
rs899303717 Chr.16: 84566546-84566546 on Build GRCh38
rs374169718 Chr.16: 84566240-84566240 on Build GRCh38
rs561174546 Chr.16: 84566239-84566239 on Build GRCh38
rs995065838 Chr.16: 84566347-84566347 on Build GRCh38
rs1015061790 Chr.16: 84566301-84566301 on Build GRCh38
rs943475735 Chr.16: 84566469-84566469 on Build GRCh38
rs764413495 Chr.16: 84566547-84566547 on Build GRCh38
rs147378148 Chr.16: 84566208-84566208 on Build GRCh38
rs568649416 Chr.16: 84566365-84566365 on Build GRCh38
rs1009683711 Chr.16: 84566555-84566555 on Build GRCh38
rs1039068868 Chr.16: 84566491-84566491 on Build GRCh38
rs563343686 Chr.16: 84566290-84566290 on Build GRCh38
rs1000303334 Chr.16: 84566500-84566500 on Build GRCh38
rs539423589 Chr.16: 84566205-84566205 on Build GRCh38
rs138017120 Chr.16: 84566499-84566499 on Build GRCh38
rs537188444 Chr.16: 84566170-84566170 on Build GRCh38
rs967146599 Chr.16: 84566382-84566382 on Build GRCh38
rs751647301 Chr.16: 84566590-84566590 on Build GRCh38
rs929332501 Chr.16: 84566449-84566449 on Build GRCh38
rs926564486 Chr.16: 84566249-84566249 on Build GRCh38
rs968940158 Chr.16: 84566207-84566207 on Build GRCh38
rs899336132 Chr.16: 84566337-84566337 on Build GRCh38
rs528514891 Chr.16: 84566185-84566185 on Build GRCh38
rs956599390 Chr.16: 84566353-84566353 on Build GRCh38
rs116476329 Chr.16: 84566184-84566184 on Build GRCh38
rs570578669 Chr.16: 84566446-84566446 on Build GRCh38
rs867758408 Chr.16: 84566363-84566363 on Build GRCh38
rs192385111 Chr.16: 84566230-84566230 on Build GRCh38
rs1036468318 Chr.16: 84566520-84566520 on Build GRCh38
rs573143169 Chr.16: 84566553-84566553 on Build GRCh38
rs190458776 Chr.16: 84566159-84566159 on Build GRCh38
rs780718634 Chr.16: 84566565-84566566 on Build GRCh38
rs958972290 Chr.16: 84566356-84566356 on Build GRCh38
rs933923149 Chr.16: 84566462-84566462 on Build GRCh38
rs968825857 Chr.16: 84566336-84566336 on Build GRCh38
rs1052311461 Chr.16: 84566294-84566294 on Build GRCh38
rs941928720 Chr.16: 84566256-84566256 on Build GRCh38
rs956372613 Chr.16: 84566228-84566228 on Build GRCh38
rs77609704 Chr.16: 84566419-84566419 on Build GRCh38
rs980475368 Chr.16: 84566401-84566401 on Build GRCh38
rs1031915467 Chr.16: 84566503-84566503 on Build GRCh38
rs1039597543 Chr.16: 84566254-84566254 on Build GRCh38
rs202078214 Chr.16: 84566227-84566227 on Build GRCh38
rs247861 Chr.16: 84566415-84566415 on Build GRCh38
rs1000420430 Chr.16: 84566341-84566341 on Build GRCh38
rs890413136 Chr.16: 84566467-84566467 on Build GRCh38
rs543543968 Chr.16: 84566270-84566270 on Build GRCh38
rs940747684 Chr.16: 84566512-84566512 on Build GRCh38
rs9934750 Chr.16: 84566299-84566299 on Build GRCh38
rs992960256 Chr.16: 84566198-84566198 on Build GRCh38
rs530508127 Chr.16: 84566544-84566544 on Build GRCh38
rs973661578 Chr.16: 84566148-84566148 on Build GRCh38
rs559064375 Chr.16: 84566314-84566314 on Build GRCh38

View Full Product Details