Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 84617740-84618234 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COTL1
Entrez Gene ID
23406
Gene Name
coactosin like F-actin binding protein 1
Gene Aliases
CLP
UniGene
Hs.289092
Gene Chromosome Location
Chr.16: 84565596-84618096 on Build GRCh38
Transcripts
NM_021149.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7286585 c.62G>A p.G21D Chr.16: 84617853-84617853 on Build GRCh38
5689117 c.39G>C p.A13A Chr.16: 84617876-84617876 on Build GRCh38
7297479 c.48G>A p.L16L Chr.16: 84617867-84617867 on Build GRCh38
4953128 c.54C>T p.R18R Chr.16: 84617861-84617861 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs959796031 Chr.16: 84618122-84618123 on Build GRCh38
rs886666858 Chr.16: 84618077-84618077 on Build GRCh38
rs542958429 Chr.16: 84617936-84617936 on Build GRCh38
rs925272625 Chr.16: 84618176-84618176 on Build GRCh38
rs565448013 Chr.16: 84617989-84617989 on Build GRCh38
rs569153901 Chr.16: 84618145-84618145 on Build GRCh38
rs931018920 Chr.16: 84617804-84617804 on Build GRCh38
rs1053006921 Chr.16: 84617867-84617867 on Build GRCh38
rs535423165 Chr.16: 84617960-84617960 on Build GRCh38
rs923320856 Chr.16: 84618213-84618213 on Build GRCh38
rs902043826 Chr.16: 84617948-84617948 on Build GRCh38
rs947326161 Chr.16: 84618041-84618041 on Build GRCh38
rs867811277 Chr.16: 84617862-84617862 on Build GRCh38
rs959891765 Chr.16: 84618026-84618026 on Build GRCh38
rs976339517 Chr.16: 84617803-84617803 on Build GRCh38
rs760920177 Chr.16: 84617821-84617821 on Build GRCh38
rs938267656 Chr.16: 84617762-84617762 on Build GRCh38
rs920796353 Chr.16: 84617786-84617786 on Build GRCh38
rs1035476597 Chr.16: 84618110-84618110 on Build GRCh38
rs778581449 Chr.16: 84617906-84617906 on Build GRCh38
rs1018155727 Chr.16: 84617944-84617944 on Build GRCh38
rs768888381 Chr.16: 84617879-84617879 on Build GRCh38
rs972935165 Chr.16: 84617995-84617995 on Build GRCh38
rs982739701 Chr.16: 84617758-84617758 on Build GRCh38
rs970170551 Chr.16: 84617998-84617998 on Build GRCh38
rs770616974 Chr.16: 84617870-84617870 on Build GRCh38
rs997713320 Chr.16: 84618066-84618066 on Build GRCh38
rs780877751 Chr.16: 84617943-84617943 on Build GRCh38
rs556688347 Chr.16: 84617929-84617929 on Build GRCh38
rs756752414 Chr.16: 84617942-84617942 on Build GRCh38
rs1016307804 Chr.16: 84618099-84618099 on Build GRCh38
rs567600273 Chr.16: 84618047-84618047 on Build GRCh38
rs1005860558 Chr.16: 84617980-84617980 on Build GRCh38
rs551536905 Chr.16: 84617971-84617971 on Build GRCh38
rs748523608 Chr.16: 84617958-84617958 on Build GRCh38
rs754101818 Chr.16: 84617900-84617900 on Build GRCh38
rs1015947605 Chr.16: 84617993-84617993 on Build GRCh38
rs901992887 Chr.16: 84618061-84618061 on Build GRCh38
rs527793750 Chr.16: 84618020-84618020 on Build GRCh38
rs769419641 Chr.16: 84617953-84617953 on Build GRCh38
rs768019411 Chr.16: 84617886-84617886 on Build GRCh38
rs568111787 Chr.16: 84617950-84617950 on Build GRCh38
rs762230766 Chr.16: 84617884-84617884 on Build GRCh38
rs980317753 Chr.16: 84617845-84617845 on Build GRCh38
rs913070234 Chr.16: 84618141-84618141 on Build GRCh38
rs776232968 Chr.16: 84617871-84617871 on Build GRCh38
rs557918370 Chr.16: 84617823-84617823 on Build GRCh38
rs71390484 Chr.16: 84618151-84618151 on Build GRCh38
rs978736443 Chr.16: 84618172-84618172 on Build GRCh38
rs987147357 Chr.16: 84618122-84618122 on Build GRCh38
rs979881705 Chr.16: 84618012-84618012 on Build GRCh38
rs752502057 Chr.16: 84617911-84617911 on Build GRCh38
rs753727953 Chr.16: 84617824-84617824 on Build GRCh38
rs71390483 Chr.16: 84618050-84618050 on Build GRCh38
rs1047937162 Chr.16: 84618069-84618069 on Build GRCh38
rs955843739 Chr.16: 84618093-84618093 on Build GRCh38
rs575551089 Chr.16: 84617776-84617776 on Build GRCh38
rs777740869 Chr.16: 84617922-84617922 on Build GRCh38
rs573530665 Chr.16: 84617929-84617929 on Build GRCh38
rs746559633 Chr.16: 84617933-84617933 on Build GRCh38
rs559900949 Chr.16: 84617877-84617877 on Build GRCh38
rs968573191 Chr.16: 84618144-84618144 on Build GRCh38
rs762009395 Chr.16: 84617811-84617811 on Build GRCh38
rs549334346 Chr.16: 84618129-84618129 on Build GRCh38
rs950349423 Chr.16: 84617968-84617968 on Build GRCh38
rs928185653 Chr.16: 84617760-84617760 on Build GRCh38
rs928458516 Chr.16: 84618015-84618015 on Build GRCh38
rs377560658 Chr.16: 84617812-84617812 on Build GRCh38
rs938596004 Chr.16: 84617854-84617854 on Build GRCh38
rs913124022 Chr.16: 84618030-84618030 on Build GRCh38
rs750581233 Chr.16: 84617895-84617895 on Build GRCh38
rs370872374 Chr.16: 84617815-84617815 on Build GRCh38
rs1006479794 Chr.16: 84618079-84618079 on Build GRCh38
rs547823859 Chr.16: 84618046-84618046 on Build GRCh38
rs764614568 Chr.16: 84617942-84617966 on Build GRCh38
rs764699182 Chr.16: 84617802-84617802 on Build GRCh38
rs943407026 Chr.16: 84618187-84618187 on Build GRCh38
rs746469890 Chr.16: 84617860-84617860 on Build GRCh38
rs538112365 Chr.16: 84618168-84618168 on Build GRCh38
rs763279643 Chr.16: 84617797-84617797 on Build GRCh38
rs17854247 Chr.16: 84617909-84617909 on Build GRCh38
rs755174079 Chr.16: 84617903-84617903 on Build GRCh38
rs758286218 Chr.16: 84617919-84617919 on Build GRCh38
rs752003141 Chr.16: 84617808-84617808 on Build GRCh38
rs868516191 Chr.16: 84617883-84617883 on Build GRCh38
rs1039515278 Chr.16: 84618196-84618196 on Build GRCh38
rs760807991 Chr.16: 84617899-84617899 on Build GRCh38
rs868581104 Chr.16: 84618109-84618109 on Build GRCh38
rs969782232 Chr.16: 84618105-84618105 on Build GRCh38
rs988678120 Chr.16: 84618028-84618028 on Build GRCh38
rs144159131 Chr.16: 84617846-84617846 on Build GRCh38
rs771981709 Chr.16: 84617849-84617849 on Build GRCh38
rs540291446 Chr.16: 84617857-84617857 on Build GRCh38
rs748052311 Chr.16: 84617847-84617847 on Build GRCh38

View Full Product Details