Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 85788869-85789350 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EMC8
Entrez Gene ID
10328
Gene Name
ER membrane protein complex subunit 8
Gene Aliases
C16orf2, C16orf4, COX4NB, FAM158B, NOC4
UniGene
Hs.173162
Gene Chromosome Location
Chr.16: 85778624-85799744 on Build GRCh38
Transcripts
NM_001142288.1 NM_006067.4 XM_011522813.2 XM_017022867.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5655802 c.305C>T p.A102V Chr.16: 85788977-85788977 on Build GRCh38
6653637 c.262G>A p.V88M Chr.16: 85789020-85789020 on Build GRCh38
7284711 c.308+5G>A p.? Chr.16: 85788969-85788969 on Build GRCh38
4674260 c.292C>T p.R98* Chr.16: 85788990-85788990 on Build GRCh38
974525 c.234T>C p.I78I Chr.16: 85789048-85789048 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs886921902 Chr.16: 85789253-85789253 on Build GRCh38
rs561102800 Chr.16: 85788906-85788906 on Build GRCh38
rs887895929 Chr.16: 85789025-85789025 on Build GRCh38
rs1029221847 Chr.16: 85789133-85789133 on Build GRCh38
rs938339114 Chr.16: 85789198-85789198 on Build GRCh38
rs574727747 Chr.16: 85789312-85789312 on Build GRCh38
rs894362372 Chr.16: 85789223-85789223 on Build GRCh38
rs969704102 Chr.16: 85789172-85789172 on Build GRCh38
rs746432525 Chr.16: 85789069-85789069 on Build GRCh38
rs976751895 Chr.16: 85789118-85789118 on Build GRCh38
rs572057331 Chr.16: 85789224-85789224 on Build GRCh38
rs878859883 Chr.16: 85789176-85789176 on Build GRCh38
rs755215472 Chr.16: 85789267-85789267 on Build GRCh38
rs547661215 Chr.16: 85789080-85789080 on Build GRCh38
rs761683906 Chr.16: 85788988-85788988 on Build GRCh38
rs990314632 Chr.16: 85789142-85789142 on Build GRCh38
rs529894288 Chr.16: 85788909-85788909 on Build GRCh38
rs766495153 Chr.16: 85789217-85789217 on Build GRCh38
rs757850605 Chr.16: 85788965-85788965 on Build GRCh38
rs781634782 Chr.16: 85788967-85788967 on Build GRCh38
rs370843220 Chr.16: 85788932-85788932 on Build GRCh38
rs928421515 Chr.16: 85789182-85789182 on Build GRCh38
rs112141090 Chr.16: 85789026-85789026 on Build GRCh38
rs2233452 Chr.16: 85789107-85789107 on Build GRCh38
rs767290019 Chr.16: 85788946-85788946 on Build GRCh38
rs911104004 Chr.16: 85789238-85789238 on Build GRCh38
rs748262512 Chr.16: 85789090-85789090 on Build GRCh38
rs757799105 Chr.16: 85789056-85789056 on Build GRCh38
rs756400390 Chr.16: 85789070-85789070 on Build GRCh38
rs770626707 Chr.16: 85788898-85788898 on Build GRCh38
rs768715754 Chr.16: 85788982-85788982 on Build GRCh38
rs767451358 Chr.16: 85788989-85788989 on Build GRCh38
rs897268020 Chr.16: 85789280-85789280 on Build GRCh38
rs370152957 Chr.16: 85788962-85788962 on Build GRCh38
rs747503145 Chr.16: 85788963-85788963 on Build GRCh38
rs371429464 Chr.16: 85788957-85788957 on Build GRCh38
rs558541079 Chr.16: 85789213-85789213 on Build GRCh38
rs947271695 Chr.16: 85788950-85788950 on Build GRCh38
rs189904972 Chr.16: 85789082-85789082 on Build GRCh38
rs752258220 Chr.16: 85789049-85789049 on Build GRCh38
rs952989232 Chr.16: 85789132-85789132 on Build GRCh38
rs932120740 Chr.16: 85788919-85788919 on Build GRCh38
rs953547265 Chr.16: 85789138-85789138 on Build GRCh38
rs773838372 Chr.16: 85789174-85789174 on Build GRCh38
rs944073360 Chr.16: 85789250-85789250 on Build GRCh38
rs549257737 Chr.16: 85788916-85788916 on Build GRCh38
rs768883105 Chr.16: 85789085-85789085 on Build GRCh38
rs1027748374 Chr.16: 85789020-85789020 on Build GRCh38
rs757826967 Chr.16: 85789327-85789327 on Build GRCh38
rs775595049 Chr.16: 85788931-85788931 on Build GRCh38
rs28414324 Chr.16: 85789096-85789096 on Build GRCh38
rs375488047 Chr.16: 85788945-85788945 on Build GRCh38
rs766246718 Chr.16: 85789017-85789017 on Build GRCh38
rs534254475 Chr.16: 85789057-85789057 on Build GRCh38
rs184684645 Chr.16: 85788969-85788969 on Build GRCh38
rs776262653 Chr.16: 85788915-85788915 on Build GRCh38
rs146555773 Chr.16: 85789021-85789021 on Build GRCh38
rs541367136 Chr.16: 85789281-85789281 on Build GRCh38
rs750235527 Chr.16: 85788948-85788948 on Build GRCh38
rs988689192 Chr.16: 85789222-85789222 on Build GRCh38
rs1049683752 Chr.16: 85789235-85789235 on Build GRCh38
rs2233453 Chr.16: 85788955-85788955 on Build GRCh38
rs1014917925 Chr.16: 85788897-85788897 on Build GRCh38
rs1004966046 Chr.16: 85789031-85789031 on Build GRCh38
rs753340379 Chr.16: 85788956-85788956 on Build GRCh38
rs536436687 Chr.16: 85789091-85789091 on Build GRCh38
rs180759307 Chr.16: 85789306-85789306 on Build GRCh38
rs1048297198 Chr.16: 85789232-85789232 on Build GRCh38
rs769913331 Chr.16: 85788929-85788929 on Build GRCh38
rs149622398 Chr.16: 85789183-85789183 on Build GRCh38
rs765440868 Chr.16: 85789313-85789313 on Build GRCh38
rs934513206 Chr.16: 85788908-85788908 on Build GRCh38
rs753884414 Chr.16: 85789228-85789228 on Build GRCh38
rs773020357 Chr.16: 85789005-85789005 on Build GRCh38
rs914742694 Chr.16: 85789150-85789150 on Build GRCh38
rs906851838 Chr.16: 85788959-85788959 on Build GRCh38
rs200274492 Chr.16: 85789012-85789012 on Build GRCh38
rs1022619441 Chr.16: 85789136-85789136 on Build GRCh38
rs751465400 Chr.16: 85788937-85788937 on Build GRCh38
rs774116960 Chr.16: 85789063-85789064 on Build GRCh38
rs754733342 Chr.16: 85789035-85789035 on Build GRCh38
rs560069282 Chr.16: 85788971-85788971 on Build GRCh38
rs538329513 Chr.16: 85789192-85789192 on Build GRCh38
rs774351547 Chr.16: 85789087-85789087 on Build GRCh38
rs774473908 Chr.16: 85788985-85788985 on Build GRCh38
rs1041416538 Chr.16: 85789261-85789261 on Build GRCh38

View Full Product Details