Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 86567536-86568028 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOXC2
Entrez Gene ID
2303
Gene Name
forkhead box C2
Gene Aliases
FKHL14, LD, MFH-1, MFH1
UniGene
Hs.436448
Gene Chromosome Location
Chr.16: 86567251-86568933 on Build GRCh38
Transcripts
NM_005251.2 NR_125795.1

Gene Symbol
FOXC2-AS1
Entrez Gene ID
103752587
Gene Name
FOXC2 antisense RNA 1
Gene Aliases
ODRUL
UniGene
Hs.609366
Gene Chromosome Location
Chr.16: 86565145-86567761 on Build GRCh38
Transcripts
NM_005251.2 NR_125795.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6230413 c.635T>C p.V212A Chr.16: 86567970-86567970 on Build GRCh38
1302319 c.563C>T p.P188L Chr.16: 86567898-86567898 on Build GRCh38
6145243 c.363_364CC>AA p.H122N Chr.16: 86567698-86567699 on Build GRCh38
365085 c.364C>A p.H122N Chr.16: 86567699-86567699 on Build GRCh38
1207404 c.409G>A p.D137N Chr.16: 86567744-86567744 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765735196 Chr.16: 86567888-86567888 on Build GRCh38
rs750515811 Chr.16: 86567942-86567942 on Build GRCh38
rs200751941 Chr.16: 86567918-86567918 on Build GRCh38
rs751774927 Chr.16: 86567898-86567898 on Build GRCh38
rs201895173 Chr.16: 86567969-86567969 on Build GRCh38
rs778706674 Chr.16: 86567958-86567958 on Build GRCh38
rs764469563 Chr.16: 86567937-86567937 on Build GRCh38
rs200301728 Chr.16: 86567994-86567994 on Build GRCh38
rs769046093 Chr.16: 86567996-86567996 on Build GRCh38
rs759417459 Chr.16: 86567617-86567617 on Build GRCh38
rs750865152 Chr.16: 86567586-86567586 on Build GRCh38
rs199924880 Chr.16: 86567865-86567865 on Build GRCh38
rs756307803 Chr.16: 86567943-86567943 on Build GRCh38
rs367861811 Chr.16: 86567869-86567869 on Build GRCh38
rs758480988 Chr.16: 86567823-86567823 on Build GRCh38
rs957867993 Chr.16: 86567559-86567559 on Build GRCh38
rs748990709 Chr.16: 86567910-86567910 on Build GRCh38
rs202085650 Chr.16: 86567882-86567882 on Build GRCh38
rs779124969 Chr.16: 86567603-86567603 on Build GRCh38
rs781462601 Chr.16: 86567856-86567856 on Build GRCh38
rs765569542 Chr.16: 86567803-86567803 on Build GRCh38
rs752429537 Chr.16: 86567642-86567642 on Build GRCh38
rs758886097 Chr.16: 86567591-86567591 on Build GRCh38
rs370541542 Chr.16: 86567870-86567870 on Build GRCh38
rs201027957 Chr.16: 86567883-86567883 on Build GRCh38
rs749722544 Chr.16: 86567598-86567598 on Build GRCh38
rs890746998 Chr.16: 86567744-86567744 on Build GRCh38
rs779821571 Chr.16: 86567945-86567945 on Build GRCh38
rs763401027 Chr.16: 86567930-86567930 on Build GRCh38
rs1048453237 Chr.16: 86567731-86567731 on Build GRCh38
rs868831268 Chr.16: 86567852-86567852 on Build GRCh38
rs762735481 Chr.16: 86567656-86567656 on Build GRCh38
rs754147232 Chr.16: 86567694-86567694 on Build GRCh38
rs771775997 Chr.16: 86567916-86567916 on Build GRCh38
rs200766961 Chr.16: 86567949-86567949 on Build GRCh38
rs772764204 Chr.16: 86567881-86567881 on Build GRCh38
rs199772307 Chr.16: 86567685-86567685 on Build GRCh38
rs774078804 Chr.16: 86567874-86567874 on Build GRCh38
rs768803682 Chr.16: 86567602-86567602 on Build GRCh38
rs755362690 Chr.16: 86567704-86567704 on Build GRCh38
rs776105836 Chr.16: 86567929-86567929 on Build GRCh38
rs768532058 Chr.16: 86567912-86567912 on Build GRCh38
rs201924901 Chr.16: 86567879-86567879 on Build GRCh38
rs758005564 Chr.16: 86567578-86567578 on Build GRCh38
rs528105097 Chr.16: 86567884-86567884 on Build GRCh38
rs371100302 Chr.16: 86567620-86567620 on Build GRCh38
rs779286561 Chr.16: 86567705-86567705 on Build GRCh38
rs901535122 Chr.16: 86567972-86567972 on Build GRCh38
rs929372985 Chr.16: 86567905-86567905 on Build GRCh38
rs748344148 Chr.16: 86567710-86567710 on Build GRCh38
rs138472501 Chr.16: 86567566-86567566 on Build GRCh38
rs774058810 Chr.16: 86567913-86567913 on Build GRCh38
rs121909107 Chr.16: 86567697-86567697 on Build GRCh38
rs1032072257 Chr.16: 86567788-86567788 on Build GRCh38
rs201189554 Chr.16: 86567775-86567775 on Build GRCh38
rs761192494 Chr.16: 86567878-86567878 on Build GRCh38
rs1040053613 Chr.16: 86567776-86567776 on Build GRCh38
rs770715665 Chr.16: 86567740-86567740 on Build GRCh38
rs563038940 Chr.16: 86567806-86567806 on Build GRCh38
rs761402501 Chr.16: 86567777-86567777 on Build GRCh38
rs1039275268 Chr.16: 86567925-86567925 on Build GRCh38
rs78018668 Chr.16: 86567907-86567907 on Build GRCh38
rs764985786 Chr.16: 86567560-86567560 on Build GRCh38
rs376293282 Chr.16: 86567785-86567785 on Build GRCh38
rs757389047 Chr.16: 86567848-86567848 on Build GRCh38
rs377509198 Chr.16: 86567627-86567627 on Build GRCh38
rs756695162 Chr.16: 86567593-86567593 on Build GRCh38
rs778068967 Chr.16: 86567836-86567836 on Build GRCh38
rs886038560 Chr.16: 86567686-86567686 on Build GRCh38
rs144326380 Chr.16: 86567610-86567610 on Build GRCh38
rs148359559 Chr.16: 86567659-86567659 on Build GRCh38
rs375618253 Chr.16: 86567689-86567689 on Build GRCh38
rs41312258 Chr.16: 86567761-86567761 on Build GRCh38
rs770965792 Chr.16: 86567611-86567611 on Build GRCh38
rs970711015 Chr.16: 86567859-86567859 on Build GRCh38
rs200483763 Chr.16: 86567901-86567901 on Build GRCh38
rs375500879 Chr.16: 86567693-86567693 on Build GRCh38
rs766826259 Chr.16: 86567780-86567780 on Build GRCh38
rs777707890 Chr.16: 86567732-86567732 on Build GRCh38
rs951933870 Chr.16: 86567862-86567864 on Build GRCh38
rs550350665 Chr.16: 86567939-86567939 on Build GRCh38
rs745779118 Chr.16: 86567861-86567861 on Build GRCh38
rs926624500 Chr.16: 86567614-86567614 on Build GRCh38
rs747011789 Chr.16: 86567844-86567844 on Build GRCh38
rs200316659 Chr.16: 86567932-86567932 on Build GRCh38
rs143108216 Chr.16: 86567608-86567608 on Build GRCh38
rs764181783 Chr.16: 86567897-86567897 on Build GRCh38
rs79452610 Chr.16: 86567770-86567770 on Build GRCh38
rs764933050 Chr.16: 86567635-86567635 on Build GRCh38
rs920777360 Chr.16: 86567653-86567653 on Build GRCh38
rs763490810 Chr.16: 86567663-86567663 on Build GRCh38
rs759364638 Chr.16: 86567751-86567751 on Build GRCh38
rs201456476 Chr.16: 86567940-86567940 on Build GRCh38
rs543303165 Chr.16: 86567796-86567796 on Build GRCh38
rs530313870 Chr.16: 86567926-86567926 on Build GRCh38
rs747819833 Chr.16: 86567915-86567915 on Build GRCh38
rs371246110 Chr.16: 86567629-86567629 on Build GRCh38
rs758812819 Chr.16: 86567896-86567896 on Build GRCh38
rs1003431373 Chr.16: 86567995-86567995 on Build GRCh38
rs104894516 Chr.16: 86567632-86567632 on Build GRCh38
rs886038561 Chr.16: 86567746-86567746 on Build GRCh38
rs746013945 Chr.16: 86567900-86567900 on Build GRCh38
rs751882462 Chr.16: 86567938-86567938 on Build GRCh38
rs1033516075 Chr.16: 86568004-86568004 on Build GRCh38
rs748182598 Chr.16: 86567605-86567605 on Build GRCh38
rs121909106 Chr.16: 86567709-86567709 on Build GRCh38
rs747115659 Chr.16: 86567737-86567737 on Build GRCh38
rs1020830836 Chr.16: 86567592-86567592 on Build GRCh38
rs545598387 Chr.16: 86567873-86567873 on Build GRCh38
rs1009368067 Chr.16: 86567849-86567849 on Build GRCh38
rs776500374 Chr.16: 86567613-86567613 on Build GRCh38
rs781409456 Chr.16: 86567899-86567899 on Build GRCh38
rs751140878 Chr.16: 86567668-86567668 on Build GRCh38
rs199935844 Chr.16: 86567962-86567962 on Build GRCh38
rs201833900 Chr.16: 86567754-86567754 on Build GRCh38
rs754888314 Chr.16: 86567956-86567956 on Build GRCh38
rs763459418 Chr.16: 86567987-86567987 on Build GRCh38
rs372749779 Chr.16: 86567959-86567959 on Build GRCh38
rs776026772 Chr.16: 86567985-86567985 on Build GRCh38
rs781696493 Chr.16: 86567581-86567581 on Build GRCh38
rs771702369 Chr.16: 86567961-86567961 on Build GRCh38
rs374000899 Chr.16: 86567594-86567594 on Build GRCh38
rs944950685 Chr.16: 86567752-86567752 on Build GRCh38
rs79883807 Chr.16: 86567983-86567983 on Build GRCh38
rs752945180 Chr.16: 86567895-86567895 on Build GRCh38

View Full Product Details