Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 86578417-86578918 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOXL1
Entrez Gene ID
2300
Gene Name
forkhead box L1
Gene Aliases
FKH6, FKHL11, FREAC7
UniGene
Hs.533830
Gene Chromosome Location
Chr.16: 86578509-86581698 on Build GRCh38
Transcripts
NM_005250.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
108486 c.128G>T p.R43L Chr.16: 86578851-86578851 on Build GRCh38
3513108 c.174C>T p.I58I Chr.16: 86578897-86578897 on Build GRCh38
1128847 c.44C>T p.S15L Chr.16: 86578767-86578767 on Build GRCh38
5149000 c.58C>G p.L20V Chr.16: 86578781-86578781 on Build GRCh38
6666376 c.98delC p.A35fs*4 Chr.16: 86578821-86578821 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769589179 Chr.16: 86578751-86578751 on Build GRCh38
rs145237316 Chr.16: 86578861-86578861 on Build GRCh38
rs374262221 Chr.16: 86578756-86578756 on Build GRCh38
rs1056333251 Chr.16: 86578763-86578763 on Build GRCh38
rs747832608 Chr.16: 86578846-86578846 on Build GRCh38
rs779829411 Chr.16: 86578833-86578833 on Build GRCh38
rs949902867 Chr.16: 86578671-86578671 on Build GRCh38
rs748909977 Chr.16: 86578755-86578755 on Build GRCh38
rs758564491 Chr.16: 86578701-86578701 on Build GRCh38
rs955044602 Chr.16: 86578489-86578489 on Build GRCh38
rs902270198 Chr.16: 86578739-86578739 on Build GRCh38
rs1015117129 Chr.16: 86578566-86578568 on Build GRCh38
rs1023994643 Chr.16: 86578852-86578852 on Build GRCh38
rs759986631 Chr.16: 86578777-86578777 on Build GRCh38
rs755415298 Chr.16: 86578687-86578687 on Build GRCh38
rs769907243 Chr.16: 86578692-86578693 on Build GRCh38
rs751668014 Chr.16: 86578817-86578817 on Build GRCh38
rs151159060 Chr.16: 86578782-86578782 on Build GRCh38
rs987683142 Chr.16: 86578565-86578565 on Build GRCh38
rs766934472 Chr.16: 86578675-86578675 on Build GRCh38
rs536150721 Chr.16: 86578472-86578472 on Build GRCh38
rs376367215 Chr.16: 86578679-86578679 on Build GRCh38
rs140270972 Chr.16: 86578707-86578707 on Build GRCh38
rs996233593 Chr.16: 86578787-86578787 on Build GRCh38
rs750324359 Chr.16: 86578823-86578823 on Build GRCh38
rs143461122 Chr.16: 86578851-86578851 on Build GRCh38
rs781327502 Chr.16: 86578730-86578730 on Build GRCh38
rs962143675 Chr.16: 86578476-86578476 on Build GRCh38
rs893805479 Chr.16: 86578728-86578728 on Build GRCh38
rs377725521 Chr.16: 86578818-86578818 on Build GRCh38
rs777845014 Chr.16: 86578705-86578705 on Build GRCh38
rs920261694 Chr.16: 86578624-86578624 on Build GRCh38
rs765554217 Chr.16: 86578690-86578690 on Build GRCh38
rs772725671 Chr.16: 86578850-86578850 on Build GRCh38
rs779734117 Chr.16: 86578754-86578754 on Build GRCh38
rs756015839 Chr.16: 86578825-86578825 on Build GRCh38
rs574929113 Chr.16: 86578661-86578661 on Build GRCh38
rs193108861 Chr.16: 86578492-86578492 on Build GRCh38
rs547433687 Chr.16: 86578602-86578602 on Build GRCh38
rs777086575 Chr.16: 86578776-86578776 on Build GRCh38
rs1014891780 Chr.16: 86578864-86578864 on Build GRCh38
rs763233943 Chr.16: 86578793-86578793 on Build GRCh38
rs757247553 Chr.16: 86578716-86578716 on Build GRCh38
rs773843053 Chr.16: 86578765-86578765 on Build GRCh38
rs973021501 Chr.16: 86578606-86578606 on Build GRCh38
rs762354526 Chr.16: 86578537-86578537 on Build GRCh38
rs763277307 Chr.16: 86578853-86578853 on Build GRCh38
rs752917409 Chr.16: 86578696-86578696 on Build GRCh38
rs367924573 Chr.16: 86578867-86578867 on Build GRCh38
rs768338015 Chr.16: 86578835-86578835 on Build GRCh38
rs937057187 Chr.16: 86578642-86578642 on Build GRCh38
rs764468898 Chr.16: 86578809-86578809 on Build GRCh38
rs781452561 Chr.16: 86578821-86578821 on Build GRCh38
rs923401593 Chr.16: 86578493-86578493 on Build GRCh38
rs765582433 Chr.16: 86578781-86578781 on Build GRCh38
rs991638685 Chr.16: 86578656-86578656 on Build GRCh38
rs759726036 Chr.16: 86578452-86578452 on Build GRCh38
rs142168655 Chr.16: 86578772-86578772 on Build GRCh38
rs140310025 Chr.16: 86578845-86578845 on Build GRCh38
rs1041579024 Chr.16: 86578674-86578674 on Build GRCh38
rs200674245 Chr.16: 86578899-86578899 on Build GRCh38
rs932253835 Chr.16: 86578572-86578572 on Build GRCh38
rs914811945 Chr.16: 86578663-86578663 on Build GRCh38
rs771526662 Chr.16: 86578847-86578847 on Build GRCh38
rs370331025 Chr.16: 86578855-86578855 on Build GRCh38
rs771400140 Chr.16: 86578775-86578775 on Build GRCh38
rs749099997 Chr.16: 86578834-86578834 on Build GRCh38
rs374067376 Chr.16: 86578865-86578865 on Build GRCh38
rs200593453 Chr.16: 86578863-86578863 on Build GRCh38
rs745627594 Chr.16: 86578738-86578738 on Build GRCh38
rs534992920 Chr.16: 86578566-86578566 on Build GRCh38

View Full Product Details